Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228209546-228210044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf145
Entrez Gene ID
574407
Gene Name
chromosome 1 open reading frame 145
Gene Aliases
-
UniGene
Hs.650039
Gene Chromosome Location
Chr.1: 228203506-228213664 on Build GRCh38
Transcripts
NR_073154.1 NR_073155.1 NM_001098623.2 NM_001271223.2 NM_052843.3 XM_005273287.4 XM_005273291.4 XM_005273298.4 XM_005273307.4 XM_006711822.3 XM_006711823.3 XM_006711827.3 XM_006711829.3 XM_011544297.2 XM_011544299.2 XM_017002443.1 XM_017002444.1 XM_017002445.1 XM_017002446.1 XM_017002447.1 XM_017002448.1 XM_017002449.1 XM_017002450.1 XM_017002451.1 XM_017002452.1 XM_017002453.1 XM_017002454.1 XM_017002455.1 XM_017002456.1 XM_017002457.1 XM_017002458.1 XM_017002459.1 XM_017002460.1 XM_017002461.1 XM_017002462.1 XM_017002463.1 XM_017002464.1 XM_017002465.1 XM_017002466.1 XM_017002467.1 XM_017002468.1 XM_017002469.1 XM_017002470.1 XM_017002471.1 XM_017002472.1 XM_017002473.1

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
UniGene
Hs.650039
Gene Chromosome Location
Chr.1: 228208013-228381431 on Build GRCh38
Transcripts
NR_073154.1 NR_073155.1 NM_001098623.2 NM_001271223.2 NM_052843.3 XM_005273287.4 XM_005273291.4 XM_005273298.4 XM_005273307.4 XM_006711822.3 XM_006711823.3 XM_006711827.3 XM_006711829.3 XM_011544297.2 XM_011544299.2 XM_017002443.1 XM_017002444.1 XM_017002445.1 XM_017002446.1 XM_017002447.1 XM_017002448.1 XM_017002449.1 XM_017002450.1 XM_017002451.1 XM_017002452.1 XM_017002453.1 XM_017002454.1 XM_017002455.1 XM_017002456.1 XM_017002457.1 XM_017002458.1 XM_017002459.1 XM_017002460.1 XM_017002461.1 XM_017002462.1 XM_017002463.1 XM_017002464.1 XM_017002465.1 XM_017002466.1 XM_017002467.1 XM_017002468.1 XM_017002469.1 XM_017002470.1 XM_017002471.1 XM_017002472.1 XM_017002473.1

SNP Detail

SNP ID Chromosome Location
rs575298706 Chr.1: 228209886-228209886 on Build GRCh38
rs1000274958 Chr.1: 228210004-228210004 on Build GRCh38
rs36002902 Chr.1: 228209621-228209622 on Build GRCh38
rs968795984 Chr.1: 228209694-228209694 on Build GRCh38
rs951877600 Chr.1: 228209927-228209927 on Build GRCh38
rs1033814968 Chr.1: 228209982-228209982 on Build GRCh38
rs1048081759 Chr.1: 228209880-228209880 on Build GRCh38
rs1050263657 Chr.1: 228209775-228209775 on Build GRCh38
rs889800127 Chr.1: 228209777-228209777 on Build GRCh38
rs564580742 Chr.1: 228209824-228209824 on Build GRCh38
rs188620507 Chr.1: 228209773-228209773 on Build GRCh38
rs140172972 Chr.1: 228209850-228209850 on Build GRCh38
rs143826126 Chr.1: 228209855-228209855 on Build GRCh38
rs1020410416 Chr.1: 228209962-228209962 on Build GRCh38
rs949261900 Chr.1: 228209776-228209776 on Build GRCh38
rs560060769 Chr.1: 228209997-228209997 on Build GRCh38
rs571814155 Chr.1: 228209735-228209735 on Build GRCh38
rs372934787 Chr.1: 228209602-228209602 on Build GRCh38
rs386369893 Chr.1: 228209628-228209629 on Build GRCh38
rs925430384 Chr.1: 228209772-228209772 on Build GRCh38
rs1020799302 Chr.1: 228209660-228209660 on Build GRCh38
rs557944739 Chr.1: 228209919-228209919 on Build GRCh38
rs540115476 Chr.1: 228209966-228209966 on Build GRCh38
rs796510815 Chr.1: 228209915-228209915 on Build GRCh38
rs1034380831 Chr.1: 228210007-228210007 on Build GRCh38
rs904643697 Chr.1: 228210003-228210003 on Build GRCh38
rs146363495 Chr.1: 228209959-228209959 on Build GRCh38
rs1010377441 Chr.1: 228209612-228209612 on Build GRCh38
rs371704045 Chr.1: 228209633-228209634 on Build GRCh38
rs992432454 Chr.1: 228209605-228209605 on Build GRCh38
rs531767256 Chr.1: 228209585-228209585 on Build GRCh38
rs1034349735 Chr.1: 228209717-228209717 on Build GRCh38
rs1042551611 Chr.1: 228209755-228209755 on Build GRCh38
rs111238500 Chr.1: 228209898-228209898 on Build GRCh38
rs901319434 Chr.1: 228209607-228209607 on Build GRCh38
rs971848145 Chr.1: 228209572-228209572 on Build GRCh38
rs137941745 Chr.1: 228209581-228209581 on Build GRCh38
rs368180628 Chr.1: 228209633-228209633 on Build GRCh38
rs756003336 Chr.1: 228209890-228209890 on Build GRCh38
rs758102887 Chr.1: 228209743-228209743 on Build GRCh38
rs566474373 Chr.1: 228209849-228209849 on Build GRCh38
rs534493303 Chr.1: 228209752-228209752 on Build GRCh38
rs926346965 Chr.1: 228209594-228209594 on Build GRCh38
rs139885646 Chr.1: 228210018-228210018 on Build GRCh38
rs569020787 Chr.1: 228209584-228209584 on Build GRCh38
rs1040310257 Chr.1: 228209842-228209842 on Build GRCh38
rs1026031066 Chr.1: 228209581-228209581 on Build GRCh38
rs901394337 Chr.1: 228209931-228209931 on Build GRCh38
rs978759752 Chr.1: 228209699-228209699 on Build GRCh38
rs1041712672 Chr.1: 228209930-228209930 on Build GRCh38

View Full Product Details