Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68033758-68034255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4691
Entrez Gene ID
100616403
Gene Name
microRNA 4691
Gene Aliases
mir-4691
UniGene
Gene Chromosome Location
Chr.11: 68033897-68033981 on Build GRCh38
Transcripts
NR_039840.1 NM_002496.3 XM_005274013.1 XM_005274014.2 XM_011545053.2

Gene Symbol
NDUFS8
Entrez Gene ID
4728
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Gene Aliases
CI-23k, CI23KD, TYKY
UniGene
Hs.90443
Gene Chromosome Location
Chr.11: 68030617-68036647 on Build GRCh38
Transcripts
NR_039840.1 NM_002496.3 XM_005274013.1 XM_005274014.2 XM_011545053.2

SNP Detail

SNP ID Chromosome Location
rs1000627782 Chr.11: 68034042-68034042 on Build GRCh38
rs140551363 Chr.11: 68034151-68034151 on Build GRCh38
rs1034544636 Chr.11: 68034083-68034083 on Build GRCh38
rs571275015 Chr.11: 68033811-68033811 on Build GRCh38
rs889678479 Chr.11: 68033869-68033869 on Build GRCh38
rs1025183309 Chr.11: 68033782-68033782 on Build GRCh38
rs762429709 Chr.11: 68034103-68034103 on Build GRCh38
rs980138058 Chr.11: 68033958-68033958 on Build GRCh38
rs1024145316 Chr.11: 68034072-68034072 on Build GRCh38
rs926831556 Chr.11: 68033806-68033806 on Build GRCh38
rs925998540 Chr.11: 68033924-68033924 on Build GRCh38
rs978898415 Chr.11: 68034114-68034114 on Build GRCh38
rs745400074 Chr.11: 68033859-68033859 on Build GRCh38
rs764505048 Chr.11: 68034112-68034112 on Build GRCh38
rs993161365 Chr.11: 68034048-68034048 on Build GRCh38
rs987304905 Chr.11: 68033817-68033817 on Build GRCh38
rs544391827 Chr.11: 68033985-68033985 on Build GRCh38
rs181585480 Chr.11: 68033953-68033953 on Build GRCh38
rs984461832 Chr.11: 68034107-68034107 on Build GRCh38
rs760169770 Chr.11: 68033863-68033863 on Build GRCh38
rs561901609 Chr.11: 68034147-68034147 on Build GRCh38
rs951729026 Chr.11: 68034095-68034095 on Build GRCh38
rs12803743 Chr.11: 68034029-68034029 on Build GRCh38
rs3115546 Chr.11: 68034050-68034050 on Build GRCh38
rs202187634 Chr.11: 68033803-68033803 on Build GRCh38
rs1032959227 Chr.11: 68033931-68033931 on Build GRCh38
rs1037196162 Chr.11: 68034019-68034019 on Build GRCh38
rs868730847 Chr.11: 68034013-68034013 on Build GRCh38
rs988684976 Chr.11: 68033946-68033946 on Build GRCh38
rs961614905 Chr.11: 68034119-68034119 on Build GRCh38
rs536528815 Chr.11: 68033890-68033890 on Build GRCh38
rs934181720 Chr.11: 68034148-68034148 on Build GRCh38
rs1037059033 Chr.11: 68033836-68033836 on Build GRCh38
rs3133267 Chr.11: 68033815-68033815 on Build GRCh38
rs971533362 Chr.11: 68034074-68034074 on Build GRCh38
rs371402717 Chr.11: 68033973-68033973 on Build GRCh38
rs1028327203 Chr.11: 68033865-68033865 on Build GRCh38
rs769304760 Chr.11: 68034002-68034002 on Build GRCh38
rs889780209 Chr.11: 68034040-68034040 on Build GRCh38
rs745651494 Chr.11: 68033887-68033887 on Build GRCh38
rs139012228 Chr.11: 68033980-68033980 on Build GRCh38
rs955840807 Chr.11: 68033937-68033937 on Build GRCh38
rs1000131766 Chr.11: 68033906-68033906 on Build GRCh38
rs1053332851 Chr.11: 68033933-68033933 on Build GRCh38
rs971176286 Chr.11: 68033956-68033956 on Build GRCh38
rs901305002 Chr.11: 68033840-68033840 on Build GRCh38
rs910343335 Chr.11: 68033845-68033845 on Build GRCh38
rs949700295 Chr.11: 68033780-68033780 on Build GRCh38
rs933180658 Chr.11: 68033816-68033816 on Build GRCh38
rs917212539 Chr.11: 68034160-68034160 on Build GRCh38
rs926999320 Chr.11: 68033960-68033960 on Build GRCh38
rs575166621 Chr.11: 68033854-68033854 on Build GRCh38
rs998586074 Chr.11: 68033858-68033858 on Build GRCh38
rs191156204 Chr.11: 68034053-68034053 on Build GRCh38
rs1050162576 Chr.11: 68034036-68034036 on Build GRCh38
rs984325988 Chr.11: 68033976-68033976 on Build GRCh38
rs901231644 Chr.11: 68034023-68034023 on Build GRCh38
rs763431787 Chr.11: 68034104-68034104 on Build GRCh38
rs1010037050 Chr.11: 68034071-68034071 on Build GRCh38
rs1001306592 Chr.11: 68034075-68034075 on Build GRCh38
rs943479203 Chr.11: 68033855-68033855 on Build GRCh38
rs950313744 Chr.11: 68034191-68034191 on Build GRCh38
rs879846379 Chr.11: 68034030-68034030 on Build GRCh38
rs552711641 Chr.11: 68034126-68034126 on Build GRCh38
rs538378336 Chr.11: 68033813-68033813 on Build GRCh38
rs534165176 Chr.11: 68033787-68033787 on Build GRCh38
rs370871461 Chr.11: 68034210-68034210 on Build GRCh38
rs114255089 Chr.11: 68033947-68033947 on Build GRCh38
rs936007158 Chr.11: 68034188-68034188 on Build GRCh38
rs370147778 Chr.11: 68033810-68033810 on Build GRCh38
rs562882705 Chr.11: 68034026-68034026 on Build GRCh38
rs534415390 Chr.11: 68034141-68034141 on Build GRCh38
rs527983870 Chr.11: 68034115-68034115 on Build GRCh38
rs376505677 Chr.11: 68033997-68033997 on Build GRCh38
rs906004623 Chr.11: 68033974-68033974 on Build GRCh38
rs186309417 Chr.11: 68033983-68033983 on Build GRCh38
rs1049816727 Chr.11: 68034150-68034150 on Build GRCh38
rs910127807 Chr.11: 68034113-68034113 on Build GRCh38
rs969165338 Chr.11: 68033798-68033798 on Build GRCh38

View Full Product Details