Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106683568-106684006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100287944
Entrez Gene ID
100287944
Gene Name
uncharacterized LOC100287944
Gene Aliases
-
UniGene
Hs.742087
Gene Chromosome Location
Chr.12: 106496410-106774831 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

Gene Symbol
LOC100505978
Entrez Gene ID
100505978
Gene Name
uncharacterized LOC100505978
Gene Aliases
-
UniGene
Hs.662583
Gene Chromosome Location
Chr.12: 106680758-106684700 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

Gene Symbol
RFX4
Entrez Gene ID
5992
Gene Name
regulatory factor X4
Gene Aliases
NYD-SP10
UniGene
Hs.388827
Gene Chromosome Location
Chr.12: 106582907-106762804 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

SNP Detail

SNP ID Chromosome Location
rs753535357 Chr.12: 106683795-106683795 on Build GRCh38
rs568841472 Chr.12: 106683903-106683903 on Build GRCh38
rs944778999 Chr.12: 106683734-106683734 on Build GRCh38
rs1031579452 Chr.12: 106683777-106683777 on Build GRCh38
rs908113801 Chr.12: 106683886-106683886 on Build GRCh38
rs1055932235 Chr.12: 106683919-106683919 on Build GRCh38
rs970140256 Chr.12: 106683815-106683816 on Build GRCh38
rs1045382058 Chr.12: 106683890-106683890 on Build GRCh38
rs767853942 Chr.12: 106683696-106683696 on Build GRCh38
rs192309074 Chr.12: 106683660-106683660 on Build GRCh38
rs952036038 Chr.12: 106683877-106683877 on Build GRCh38
rs921824762 Chr.12: 106683742-106683742 on Build GRCh38
rs1033886723 Chr.12: 106683655-106683655 on Build GRCh38
rs956106910 Chr.12: 106683794-106683794 on Build GRCh38
rs904218037 Chr.12: 106683978-106683978 on Build GRCh38
rs1049000091 Chr.12: 106683796-106683796 on Build GRCh38
rs930453969 Chr.12: 106683747-106683747 on Build GRCh38
rs550554264 Chr.12: 106683866-106683871 on Build GRCh38
rs1048874248 Chr.12: 106683713-106683725 on Build GRCh38
rs182750463 Chr.12: 106683887-106683887 on Build GRCh38
rs989775007 Chr.12: 106683684-106683684 on Build GRCh38
rs796506949 Chr.12: 106683707-106683707 on Build GRCh38
rs7309351 Chr.12: 106683654-106683654 on Build GRCh38
rs1049486662 Chr.12: 106683929-106683929 on Build GRCh38
rs1024819746 Chr.12: 106683811-106683811 on Build GRCh38
rs558313216 Chr.12: 106683597-106683597 on Build GRCh38
rs966925090 Chr.12: 106683701-106683701 on Build GRCh38
rs150727859 Chr.12: 106683885-106683885 on Build GRCh38
rs12425370 Chr.12: 106683663-106683663 on Build GRCh38
rs756427941 Chr.12: 106683725-106683725 on Build GRCh38
rs778547565 Chr.12: 106683907-106683907 on Build GRCh38
rs908067042 Chr.12: 106683635-106683635 on Build GRCh38
rs980091852 Chr.12: 106683840-106683840 on Build GRCh38
rs900879409 Chr.12: 106683692-106683692 on Build GRCh38
rs573855846 Chr.12: 106683726-106683726 on Build GRCh38
rs762450394 Chr.12: 106683664-106683664 on Build GRCh38
rs935645246 Chr.12: 106683980-106683980 on Build GRCh38
rs141560549 Chr.12: 106683737-106683737 on Build GRCh38
rs141024361 Chr.12: 106683714-106683714 on Build GRCh38

View Full Product Details