Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 91351075-91351574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR17HG
Entrez Gene ID
407975
Gene Name
miR-17-92a-1 cluster host gene
Gene Aliases
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92
UniGene
Hs.24115
Gene Chromosome Location
Chr.13: 91347820-91354575 on Build GRCh38
Transcripts
NR_027349.1 NR_027350.1 NR_029490.1 NR_029492.1 NR_029508.1

Gene Symbol
MIR19B1
Entrez Gene ID
406980
Gene Name
microRNA 19b-1
Gene Aliases
MIR19B, MIRN19B1, miR-19b-1, miRNA19B1
UniGene
Hs.24115
Gene Chromosome Location
Chr.13: 91351192-91351278 on Build GRCh38
Transcripts
NR_027349.1 NR_027350.1 NR_029490.1 NR_029492.1 NR_029508.1

Gene Symbol
MIR20A
Entrez Gene ID
406982
Gene Name
microRNA 20a
Gene Aliases
MIR20, MIRN20, MIRN20A, hsa-mir-20, hsa-mir-20a, miR-20, miRNA20A, mir-20a
UniGene
Hs.24115
Gene Chromosome Location
Chr.13: 91351065-91351135 on Build GRCh38
Transcripts
NR_027349.1 NR_027350.1 NR_029490.1 NR_029492.1 NR_029508.1

Gene Symbol
MIR92A1
Entrez Gene ID
407048
Gene Name
microRNA 92a-1
Gene Aliases
MIR92-1, MIRN92-1, MIRN92A-1, MIRN92A1, miRNA92-1, mir-92a-1
UniGene
Hs.24115
Gene Chromosome Location
Chr.13: 91351314-91351391 on Build GRCh38
Transcripts
NR_027349.1 NR_027350.1 NR_029490.1 NR_029492.1 NR_029508.1

SNP Detail

SNP ID Chromosome Location
rs930546473 Chr.13: 91351471-91351471 on Build GRCh38
rs190933807 Chr.13: 91351441-91351441 on Build GRCh38
rs896163168 Chr.13: 91351546-91351546 on Build GRCh38
rs374040259 Chr.13: 91351357-91351357 on Build GRCh38
rs747974417 Chr.13: 91351365-91351366 on Build GRCh38
rs776965935 Chr.13: 91351236-91351236 on Build GRCh38
rs759748416 Chr.13: 91351115-91351115 on Build GRCh38
rs777705064 Chr.13: 91351368-91351369 on Build GRCh38
rs764242654 Chr.13: 91351301-91351301 on Build GRCh38
rs767623695 Chr.13: 91351316-91351316 on Build GRCh38
rs377632383 Chr.13: 91351258-91351258 on Build GRCh38
rs913705730 Chr.13: 91351514-91351514 on Build GRCh38
rs374218049 Chr.13: 91351201-91351201 on Build GRCh38
rs377119994 Chr.13: 91351181-91351181 on Build GRCh38
rs771404053 Chr.13: 91351348-91351348 on Build GRCh38
rs754002158 Chr.13: 91351113-91351113 on Build GRCh38
rs904997584 Chr.13: 91351480-91351483 on Build GRCh38
rs769561768 Chr.13: 91351170-91351170 on Build GRCh38
rs190300180 Chr.13: 91351535-91351535 on Build GRCh38
rs957869044 Chr.13: 91351446-91351446 on Build GRCh38
rs994661435 Chr.13: 91351116-91351116 on Build GRCh38
rs989358175 Chr.13: 91351506-91351506 on Build GRCh38
rs777162203 Chr.13: 91351352-91351352 on Build GRCh38
rs758544671 Chr.13: 91351155-91351155 on Build GRCh38
rs757057109 Chr.13: 91351164-91351164 on Build GRCh38
rs897783443 Chr.13: 91351470-91351470 on Build GRCh38
rs945273726 Chr.13: 91351175-91351175 on Build GRCh38
rs766752478 Chr.13: 91351105-91351105 on Build GRCh38
rs370270260 Chr.13: 91351347-91351347 on Build GRCh38
rs780223781 Chr.13: 91351353-91351353 on Build GRCh38
rs1048918150 Chr.13: 91351477-91351477 on Build GRCh38
rs887397729 Chr.13: 91351327-91351327 on Build GRCh38
rs758563737 Chr.13: 91351290-91351291 on Build GRCh38
rs367923750 Chr.13: 91351375-91351375 on Build GRCh38
rs759942548 Chr.13: 91351239-91351239 on Build GRCh38
rs185444885 Chr.13: 91351519-91351519 on Build GRCh38
rs764286076 Chr.13: 91351404-91351404 on Build GRCh38
rs554016147 Chr.13: 91351543-91351543 on Build GRCh38
rs757259599 Chr.13: 91351307-91351307 on Build GRCh38
rs371396932 Chr.13: 91351321-91351321 on Build GRCh38
rs779800151 Chr.13: 91351173-91351173 on Build GRCh38
rs770213461 Chr.13: 91351355-91351355 on Build GRCh38
rs778492035 Chr.13: 91351342-91351342 on Build GRCh38
rs747844782 Chr.13: 91351443-91351443 on Build GRCh38
rs774562990 Chr.13: 91351421-91351421 on Build GRCh38
rs372889756 Chr.13: 91351176-91351176 on Build GRCh38
rs980850309 Chr.13: 91351108-91351108 on Build GRCh38
rs537481270 Chr.13: 91351464-91351464 on Build GRCh38
rs371637883 Chr.13: 91351430-91351430 on Build GRCh38
rs762890626 Chr.13: 91351283-91351283 on Build GRCh38
rs72631821 Chr.13: 91351334-91351334 on Build GRCh38
rs765344673 Chr.13: 91351241-91351241 on Build GRCh38
rs369660739 Chr.13: 91351189-91351189 on Build GRCh38
rs182719017 Chr.13: 91351442-91351442 on Build GRCh38
rs745728027 Chr.13: 91351167-91351167 on Build GRCh38
rs750420137 Chr.13: 91351432-91351432 on Build GRCh38
rs945350331 Chr.13: 91351536-91351536 on Build GRCh38
rs568111799 Chr.13: 91351531-91351531 on Build GRCh38
rs755896235 Chr.13: 91351367-91351368 on Build GRCh38
rs1035144931 Chr.13: 91351516-91351516 on Build GRCh38
rs9589207 Chr.13: 91351335-91351335 on Build GRCh38
rs533461664 Chr.13: 91351418-91351418 on Build GRCh38
rs752695760 Chr.13: 91351148-91351148 on Build GRCh38
rs376270162 Chr.13: 91351197-91351197 on Build GRCh38
rs751473041 Chr.13: 91351305-91351305 on Build GRCh38
rs752994578 Chr.13: 91351256-91351256 on Build GRCh38
rs756016604 Chr.13: 91351322-91351322 on Build GRCh38
rs778027764 Chr.13: 91351512-91351516 on Build GRCh38
rs533088983 Chr.13: 91351168-91351171 on Build GRCh38
rs780955709 Chr.13: 91351300-91351300 on Build GRCh38
rs368984000 Chr.13: 91351165-91351165 on Build GRCh38
rs185831554 Chr.13: 91351102-91351102 on Build GRCh38
rs746189994 Chr.13: 91351354-91351354 on Build GRCh38
rs986596758 Chr.13: 91351402-91351402 on Build GRCh38
rs751535398 Chr.13: 91351157-91351157 on Build GRCh38
rs762035134 Chr.13: 91351422-91351422 on Build GRCh38
rs926478042 Chr.13: 91351112-91351112 on Build GRCh38
rs750642161 Chr.13: 91351269-91351270 on Build GRCh38
rs779808798 Chr.13: 91351172-91351172 on Build GRCh38
rs1002349918 Chr.13: 91351188-91351188 on Build GRCh38
rs749184556 Chr.13: 91351419-91351422 on Build GRCh38
rs771220050 Chr.13: 91351229-91351229 on Build GRCh38
rs765553584 Chr.13: 91351141-91351141 on Build GRCh38
rs1001980789 Chr.13: 91351489-91351489 on Build GRCh38
rs763238033 Chr.13: 91351358-91351358 on Build GRCh38
rs374824450 Chr.13: 91351156-91351156 on Build GRCh38
rs761184964 Chr.13: 91351203-91351203 on Build GRCh38
rs770733281 Chr.13: 91351297-91351297 on Build GRCh38

View Full Product Details