Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23296643-23297176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3E
Entrez Gene ID
90673
Gene Name
protein phosphatase 1 regulatory subunit 3E
Gene Aliases
-
UniGene
Hs.601513
Gene Chromosome Location
Chr.14: 23295921-23302848 on Build GRCh38
Transcripts
NM_001276318.1

SNP Detail

SNP ID Chromosome Location
rs978975663 Chr.14: 23296913-23296913 on Build GRCh38
rs1022774632 Chr.14: 23296704-23296704 on Build GRCh38
rs921708540 Chr.14: 23297126-23297126 on Build GRCh38
rs940614789 Chr.14: 23296979-23296979 on Build GRCh38
rs998058953 Chr.14: 23297103-23297103 on Build GRCh38
rs1065176 Chr.14: 23296965-23296965 on Build GRCh38
rs187495227 Chr.14: 23296827-23296827 on Build GRCh38
rs886378002 Chr.14: 23296906-23296906 on Build GRCh38
rs957211440 Chr.14: 23297067-23297067 on Build GRCh38
rs1013421120 Chr.14: 23296688-23296688 on Build GRCh38
rs968166014 Chr.14: 23296798-23296798 on Build GRCh38
rs191648830 Chr.14: 23297025-23297025 on Build GRCh38
rs184970544 Chr.14: 23297145-23297145 on Build GRCh38
rs751064924 Chr.14: 23296775-23296775 on Build GRCh38
rs934908232 Chr.14: 23296938-23296938 on Build GRCh38
rs765288932 Chr.14: 23297111-23297111 on Build GRCh38
rs569605734 Chr.14: 23296991-23296991 on Build GRCh38
rs570785800 Chr.14: 23296868-23296868 on Build GRCh38
rs896451418 Chr.14: 23296989-23296989 on Build GRCh38
rs78507263 Chr.14: 23297136-23297136 on Build GRCh38
rs988923884 Chr.14: 23296824-23296824 on Build GRCh38
rs924854406 Chr.14: 23296917-23296917 on Build GRCh38
rs974450846 Chr.14: 23296864-23296864 on Build GRCh38
rs1012306475 Chr.14: 23296990-23296990 on Build GRCh38
rs897041737 Chr.14: 23297079-23297079 on Build GRCh38
rs1057115851 Chr.14: 23296947-23296947 on Build GRCh38
rs549998546 Chr.14: 23296912-23296912 on Build GRCh38
rs1035006995 Chr.14: 23296699-23296699 on Build GRCh38
rs944446929 Chr.14: 23296854-23296854 on Build GRCh38
rs1001014738 Chr.14: 23297043-23297043 on Build GRCh38
rs888290946 Chr.14: 23296714-23296714 on Build GRCh38
rs752591702 Chr.14: 23297134-23297134 on Build GRCh38
rs1005414927 Chr.14: 23296761-23296761 on Build GRCh38
rs895870770 Chr.14: 23296666-23296666 on Build GRCh38
rs976377601 Chr.14: 23296822-23296822 on Build GRCh38
rs985631403 Chr.14: 23296847-23296847 on Build GRCh38
rs1065175 Chr.14: 23297002-23297002 on Build GRCh38
rs903936753 Chr.14: 23297040-23297040 on Build GRCh38
rs41285524 Chr.14: 23296879-23296879 on Build GRCh38
rs760828089 Chr.14: 23296806-23296806 on Build GRCh38
rs368538940 Chr.14: 23296968-23296971 on Build GRCh38
rs1019029781 Chr.14: 23297107-23297107 on Build GRCh38
rs773584875 Chr.14: 23296805-23296805 on Build GRCh38
rs896600875 Chr.14: 23296700-23296700 on Build GRCh38
rs1023764921 Chr.14: 23296735-23296735 on Build GRCh38
rs772326308 Chr.14: 23296685-23296685 on Build GRCh38
rs1012275511 Chr.14: 23296731-23296731 on Build GRCh38
rs771191492 Chr.14: 23296878-23296878 on Build GRCh38
rs985067364 Chr.14: 23297141-23297141 on Build GRCh38
rs539313465 Chr.14: 23296814-23296814 on Build GRCh38
rs1056900614 Chr.14: 23296988-23296988 on Build GRCh38
rs534344703 Chr.14: 23297147-23297147 on Build GRCh38
rs888564415 Chr.14: 23297021-23297021 on Build GRCh38
rs1005320428 Chr.14: 23296678-23296678 on Build GRCh38
rs987037821 Chr.14: 23297092-23297092 on Build GRCh38
rs1031166522 Chr.14: 23297064-23297064 on Build GRCh38
rs146615313 Chr.14: 23296803-23296803 on Build GRCh38
rs979669276 Chr.14: 23296801-23296801 on Build GRCh38
rs1005798087 Chr.14: 23297041-23297041 on Build GRCh38
rs974502881 Chr.14: 23297116-23297116 on Build GRCh38

View Full Product Details