Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 6732511-6733025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00487
Entrez Gene ID
400941
Gene Name
long intergenic non-protein coding RNA 487
Gene Aliases
-
UniGene
Hs.446195
Gene Chromosome Location
Chr.2: 6729168-6770311 on Build GRCh38
Transcripts
NR_038369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50819 c.119G>C p.R40T Chr.2: 6732788-6732788 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs947348520 Chr.2: 6732705-6732705 on Build GRCh38
rs985000784 Chr.2: 6732724-6732724 on Build GRCh38
rs951549605 Chr.2: 6732628-6732628 on Build GRCh38
rs745699866 Chr.2: 6732823-6732823 on Build GRCh38
rs960462449 Chr.2: 6732942-6732942 on Build GRCh38
rs918996092 Chr.2: 6732619-6732619 on Build GRCh38
rs541354174 Chr.2: 6732833-6732833 on Build GRCh38
rs558991503 Chr.2: 6732746-6732746 on Build GRCh38
rs1007853469 Chr.2: 6732539-6732539 on Build GRCh38
rs373975515 Chr.2: 6732652-6732652 on Build GRCh38
rs376093190 Chr.2: 6732725-6732725 on Build GRCh38
rs10204406 Chr.2: 6732847-6732847 on Build GRCh38
rs1010034363 Chr.2: 6732676-6732676 on Build GRCh38
rs776542459 Chr.2: 6732772-6732772 on Build GRCh38
rs572745838 Chr.2: 6732792-6732792 on Build GRCh38
rs973189460 Chr.2: 6732590-6732590 on Build GRCh38
rs777201108 Chr.2: 6732717-6732717 on Build GRCh38
rs1001957226 Chr.2: 6732694-6732694 on Build GRCh38
rs112793812 Chr.2: 6732732-6732732 on Build GRCh38
rs890244693 Chr.2: 6732774-6732774 on Build GRCh38
rs978722544 Chr.2: 6733001-6733001 on Build GRCh38
rs1006137331 Chr.2: 6732997-6732998 on Build GRCh38
rs889731902 Chr.2: 6732607-6732607 on Build GRCh38
rs991771138 Chr.2: 6732956-6732956 on Build GRCh38
rs1045545028 Chr.2: 6732884-6732884 on Build GRCh38
rs1026108633 Chr.2: 6732835-6732835 on Build GRCh38
rs907302974 Chr.2: 6732886-6732886 on Build GRCh38
rs879615754 Chr.2: 6732998-6732998 on Build GRCh38
rs915934073 Chr.2: 6732700-6732700 on Build GRCh38
rs769605279 Chr.2: 6732826-6732826 on Build GRCh38
rs186956192 Chr.2: 6732641-6732641 on Build GRCh38
rs10166762 Chr.2: 6732685-6732685 on Build GRCh38
rs922363897 Chr.2: 6732775-6732775 on Build GRCh38
rs1034347071 Chr.2: 6732940-6732940 on Build GRCh38
rs933737570 Chr.2: 6732778-6732778 on Build GRCh38
rs992113126 Chr.2: 6732698-6732698 on Build GRCh38
rs1001612624 Chr.2: 6732929-6732929 on Build GRCh38
rs1017909294 Chr.2: 6732820-6732820 on Build GRCh38
rs553971544 Chr.2: 6732573-6732573 on Build GRCh38
rs370594391 Chr.2: 6732624-6732624 on Build GRCh38
rs531705998 Chr.2: 6732965-6732965 on Build GRCh38
rs982946098 Chr.2: 6732660-6732660 on Build GRCh38
rs370073860 Chr.2: 6732728-6732728 on Build GRCh38
rs370619956 Chr.2: 6732649-6732649 on Build GRCh38
rs769363428 Chr.2: 6732714-6732714 on Build GRCh38
rs1035863467 Chr.2: 6732872-6732872 on Build GRCh38
rs963349923 Chr.2: 6732575-6732575 on Build GRCh38
rs376794086 Chr.2: 6732541-6732541 on Build GRCh38
rs752879335 Chr.2: 6732814-6732814 on Build GRCh38
rs191341882 Chr.2: 6732681-6732681 on Build GRCh38
rs1017273284 Chr.2: 6732555-6732555 on Build GRCh38
rs1049382034 Chr.2: 6732604-6732604 on Build GRCh38
rs368616466 Chr.2: 6732998-6732998 on Build GRCh38
rs981824645 Chr.2: 6732868-6732868 on Build GRCh38
rs747787396 Chr.2: 6732632-6732632 on Build GRCh38
rs985153532 Chr.2: 6732780-6732780 on Build GRCh38
rs141282471 Chr.2: 6732883-6732883 on Build GRCh38
rs1054377969 Chr.2: 6732830-6732830 on Build GRCh38
rs182836095 Chr.2: 6732846-6732846 on Build GRCh38
rs577223977 Chr.2: 6732655-6732655 on Build GRCh38
rs1054326051 Chr.2: 6732637-6732637 on Build GRCh38
rs898860064 Chr.2: 6732824-6732824 on Build GRCh38
rs1007493046 Chr.2: 6732795-6732795 on Build GRCh38

View Full Product Details