Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49280203-49280701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORD12
Entrez Gene ID
692057
Gene Name
small nucleolar RNA, C/D box 12
Gene Aliases
HBII-99, MIR1259, MIRN1259
UniGene
Gene Chromosome Location
Chr.20: 49280683-49280772 on Build GRCh38
Transcripts
NR_003030.1 NR_003695.1 NR_003604.2 NR_003605.1 NR_003606.2 NR_036658.1 NR_036659.1

Gene Symbol
SNORD12B
Entrez Gene ID
100113393
Gene Name
small nucleolar RNA, C/D box 12B
Gene Aliases
-
UniGene
Hs.711216
Gene Chromosome Location
Chr.20: 49280313-49280415 on Build GRCh38
Transcripts
NR_003030.1 NR_003695.1 NR_003604.2 NR_003605.1 NR_003606.2 NR_036658.1 NR_036659.1

Gene Symbol
ZFAS1
Entrez Gene ID
441951
Gene Name
ZNFX1 antisense RNA 1
Gene Aliases
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1
UniGene
Hs.356766
Gene Chromosome Location
Chr.20: 49278178-49289260 on Build GRCh38
Transcripts
NR_003030.1 NR_003695.1 NR_003604.2 NR_003605.1 NR_003606.2 NR_036658.1 NR_036659.1

SNP Detail

SNP ID Chromosome Location
rs775155140 Chr.20: 49280649-49280649 on Build GRCh38
rs992112191 Chr.20: 49280348-49280348 on Build GRCh38
rs751303900 Chr.20: 49280262-49280262 on Build GRCh38
rs1036081654 Chr.20: 49280477-49280477 on Build GRCh38
rs898487517 Chr.20: 49280534-49280534 on Build GRCh38
rs749223334 Chr.20: 49280468-49280468 on Build GRCh38
rs769045594 Chr.20: 49280434-49280434 on Build GRCh38
rs765883348 Chr.20: 49280232-49280232 on Build GRCh38
rs753333943 Chr.20: 49280347-49280347 on Build GRCh38
rs770659835 Chr.20: 49280290-49280290 on Build GRCh38
rs113003243 Chr.20: 49280484-49280484 on Build GRCh38
rs902691534 Chr.20: 49280592-49280592 on Build GRCh38
rs544703254 Chr.20: 49280337-49280337 on Build GRCh38
rs780654382 Chr.20: 49280354-49280354 on Build GRCh38
rs771127237 Chr.20: 49280644-49280644 on Build GRCh38
rs767295852 Chr.20: 49280368-49280370 on Build GRCh38
rs185296829 Chr.20: 49280301-49280301 on Build GRCh38
rs776727899 Chr.20: 49280308-49280308 on Build GRCh38
rs374543410 Chr.20: 49280371-49280371 on Build GRCh38
rs536091598 Chr.20: 49280556-49280556 on Build GRCh38
rs1013912137 Chr.20: 49280531-49280531 on Build GRCh38
rs949927568 Chr.20: 49280281-49280281 on Build GRCh38
rs747498452 Chr.20: 49280356-49280356 on Build GRCh38
rs563705960 Chr.20: 49280661-49280663 on Build GRCh38
rs774652914 Chr.20: 49280662-49280662 on Build GRCh38
rs767500565 Chr.20: 49280327-49280327 on Build GRCh38
rs891989456 Chr.20: 49280270-49280270 on Build GRCh38
rs755973388 Chr.20: 49280403-49280403 on Build GRCh38
rs776690361 Chr.20: 49280663-49280663 on Build GRCh38
rs760515326 Chr.20: 49280652-49280652 on Build GRCh38
rs1001655437 Chr.20: 49280476-49280476 on Build GRCh38
rs905839549 Chr.20: 49280305-49280305 on Build GRCh38
rs780300199 Chr.20: 49280416-49280416 on Build GRCh38
rs535286675 Chr.20: 49280275-49280275 on Build GRCh38
rs756816625 Chr.20: 49280406-49280406 on Build GRCh38
rs761812392 Chr.20: 49280360-49280360 on Build GRCh38
rs1133114 Chr.20: 49280569-49280569 on Build GRCh38
rs988990065 Chr.20: 49280608-49280608 on Build GRCh38
rs755566114 Chr.20: 49280279-49280279 on Build GRCh38
rs943754352 Chr.20: 49280594-49280594 on Build GRCh38
rs556374583 Chr.20: 49280359-49280359 on Build GRCh38
rs376957272 Chr.20: 49280285-49280285 on Build GRCh38
rs991144188 Chr.20: 49280653-49280653 on Build GRCh38
rs368380129 Chr.20: 49280372-49280372 on Build GRCh38
rs749900503 Chr.20: 49280671-49280671 on Build GRCh38
rs188440884 Chr.20: 49280667-49280667 on Build GRCh38
rs1021699373 Chr.20: 49280286-49280286 on Build GRCh38
rs750103183 Chr.20: 49280377-49280377 on Build GRCh38
rs367833440 Chr.20: 49280593-49280593 on Build GRCh38
rs777625994 Chr.20: 49280454-49280454 on Build GRCh38
rs746295209 Chr.20: 49280352-49280352 on Build GRCh38
rs957017618 Chr.20: 49280622-49280622 on Build GRCh38
rs554061974 Chr.20: 49280278-49280278 on Build GRCh38
rs370860139 Chr.20: 49280414-49280414 on Build GRCh38
rs527995911 Chr.20: 49280609-49280609 on Build GRCh38
rs112684977 Chr.20: 49280483-49280483 on Build GRCh38
rs568285026 Chr.20: 49280261-49280261 on Build GRCh38
rs944272051 Chr.20: 49280410-49280410 on Build GRCh38
rs754919768 Chr.20: 49280271-49280271 on Build GRCh38
rs749002161 Chr.20: 49280641-49280641 on Build GRCh38
rs547936595 Chr.20: 49280541-49280541 on Build GRCh38
rs781719579 Chr.20: 49280435-49280435 on Build GRCh38
rs915853699 Chr.20: 49280446-49280447 on Build GRCh38
rs751307159 Chr.20: 49280267-49280267 on Build GRCh38
rs765382850 Chr.20: 49280312-49280312 on Build GRCh38
rs559013144 Chr.20: 49280309-49280309 on Build GRCh38
rs376973398 Chr.20: 49280467-49280468 on Build GRCh38
rs1011616111 Chr.20: 49280277-49280277 on Build GRCh38
rs538942709 Chr.20: 49280233-49280233 on Build GRCh38
rs763999183 Chr.20: 49280660-49280660 on Build GRCh38
rs778707314 Chr.20: 49280351-49280351 on Build GRCh38
rs751359007 Chr.20: 49280316-49280316 on Build GRCh38
rs1026703433 Chr.20: 49280562-49280562 on Build GRCh38
rs985253185 Chr.20: 49280580-49280580 on Build GRCh38
rs778762269 Chr.20: 49280408-49280408 on Build GRCh38
rs912213537 Chr.20: 49280590-49280590 on Build GRCh38
rs759319180 Chr.20: 49280317-49280317 on Build GRCh38
rs772081385 Chr.20: 49280498-49280498 on Build GRCh38
rs374810588 Chr.20: 49280670-49280670 on Build GRCh38
rs749534566 Chr.20: 49280635-49280635 on Build GRCh38
rs747338510 Chr.20: 49280425-49280425 on Build GRCh38
rs1015525926 Chr.20: 49280539-49280539 on Build GRCh38
rs748282993 Chr.20: 49280451-49280451 on Build GRCh38
rs774871572 Chr.20: 49280298-49280298 on Build GRCh38
rs756976100 Chr.20: 49280349-49280349 on Build GRCh38
rs760544869 Chr.20: 49280401-49280401 on Build GRCh38
rs761452858 Chr.20: 49280668-49280668 on Build GRCh38
rs539709636 Chr.20: 49280306-49280306 on Build GRCh38
rs757223176 Chr.20: 49280279-49280280 on Build GRCh38
rs944194386 Chr.20: 49280237-49280237 on Build GRCh38
rs748807648 Chr.20: 49280547-49280548 on Build GRCh38
rs760861632 Chr.20: 49280307-49280307 on Build GRCh38
rs577263499 Chr.20: 49280333-49280333 on Build GRCh38
rs747388235 Chr.20: 49280252-49280252 on Build GRCh38
rs116993790 Chr.20: 49280370-49280370 on Build GRCh38
rs891410546 Chr.20: 49280554-49280554 on Build GRCh38
rs764037452 Chr.20: 49280266-49280288 on Build GRCh38
rs961159105 Chr.20: 49280540-49280540 on Build GRCh38
rs752554628 Chr.20: 49280328-49280328 on Build GRCh38
rs578258967 Chr.20: 49280364-49280364 on Build GRCh38
rs779505060 Chr.20: 49280470-49280471 on Build GRCh38

View Full Product Details