Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 16070416-16070910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR99AHG
Entrez Gene ID
388815
Gene Name
mir-99a-let-7c cluster host gene
Gene Aliases
C21orf34, C21orf35, LINC00478, MONC
UniGene
Hs.473394
Gene Chromosome Location
Chr.21: 16070488-16631727 on Build GRCh38
Transcripts
NR_027790.3 NR_136541.1 NR_136542.1

SNP Detail

SNP ID Chromosome Location
rs1014337451 Chr.21: 16070449-16070449 on Build GRCh38
rs976520054 Chr.21: 16070809-16070809 on Build GRCh38
rs187416164 Chr.21: 16070792-16070792 on Build GRCh38
rs1039600327 Chr.21: 16070604-16070604 on Build GRCh38
rs528902900 Chr.21: 16070658-16070658 on Build GRCh38
rs17307260 Chr.21: 16070725-16070725 on Build GRCh38
rs890440499 Chr.21: 16070539-16070539 on Build GRCh38
rs1051822789 Chr.21: 16070728-16070728 on Build GRCh38
rs956463987 Chr.21: 16070639-16070639 on Build GRCh38
rs1015487177 Chr.21: 16070786-16070786 on Build GRCh38
rs1045854941 Chr.21: 16070704-16070704 on Build GRCh38
rs556183520 Chr.21: 16070600-16070600 on Build GRCh38
rs908357609 Chr.21: 16070552-16070552 on Build GRCh38
rs182410710 Chr.21: 16070689-16070689 on Build GRCh38
rs1000695450 Chr.21: 16070622-16070622 on Build GRCh38
rs763586934 Chr.21: 16070729-16070729 on Build GRCh38
rs929412036 Chr.21: 16070527-16070527 on Build GRCh38
rs1032617560 Chr.21: 16070628-16070628 on Build GRCh38
rs544412806 Chr.21: 16070555-16070555 on Build GRCh38
rs942173966 Chr.21: 16070481-16070481 on Build GRCh38
rs55954827 Chr.21: 16070677-16070677 on Build GRCh38
rs374186144 Chr.21: 16070649-16070649 on Build GRCh38
rs970067120 Chr.21: 16070484-16070484 on Build GRCh38
rs919444829 Chr.21: 16070698-16070698 on Build GRCh38
rs553727435 Chr.21: 16070523-16070523 on Build GRCh38
rs1053727856 Chr.21: 16070871-16070871 on Build GRCh38
rs564317367 Chr.21: 16070573-16070573 on Build GRCh38
rs905936599 Chr.21: 16070711-16070711 on Build GRCh38
rs980061957 Chr.21: 16070495-16070495 on Build GRCh38
rs2823596 Chr.21: 16070697-16070697 on Build GRCh38
rs145261259 Chr.21: 16070775-16070775 on Build GRCh38
rs763187544 Chr.21: 16070490-16070490 on Build GRCh38
rs778274851 Chr.21: 16070617-16070617 on Build GRCh38
rs553414500 Chr.21: 16070773-16070773 on Build GRCh38
rs1051751602 Chr.21: 16070535-16070535 on Build GRCh38
rs899733986 Chr.21: 16070762-16070762 on Build GRCh38
rs943345992 Chr.21: 16070734-16070734 on Build GRCh38
rs575471803 Chr.21: 16070503-16070503 on Build GRCh38
rs1038315720 Chr.21: 16070522-16070522 on Build GRCh38
rs926886065 Chr.21: 16070451-16070451 on Build GRCh38
rs540092512 Chr.21: 16070742-16070742 on Build GRCh38
rs533241136 Chr.21: 16070602-16070602 on Build GRCh38
rs1024369444 Chr.21: 16070477-16070477 on Build GRCh38
rs904813239 Chr.21: 16070672-16070673 on Build GRCh38
rs555840705 Chr.21: 16070850-16070850 on Build GRCh38
rs980408431 Chr.21: 16070659-16070659 on Build GRCh38
rs1009706953 Chr.21: 16070646-16070646 on Build GRCh38
rs976488773 Chr.21: 16070618-16070618 on Build GRCh38
rs546609929 Chr.21: 16070605-16070605 on Build GRCh38
rs550741147 Chr.21: 16070710-16070710 on Build GRCh38
rs1039086643 Chr.21: 16070745-16070745 on Build GRCh38
rs199514533 Chr.21: 16070660-16070661 on Build GRCh38

View Full Product Details