Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46090881-46091385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR3619
Entrez Gene ID
100500828
Gene Name
microRNA 3619
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.22: 46091044-46091126 on Build GRCh38
Transcripts
NR_037413.1 NR_027033.2 NR_110479.1

Gene Symbol
MIRLET7BHG
Entrez Gene ID
400931
Gene Name
MIRLET7B host gene
Gene Aliases
linc-Ppara
UniGene
Hs.235838
Gene Chromosome Location
Chr.22: 46085997-46113928 on Build GRCh38
Transcripts
NR_037413.1 NR_027033.2 NR_110479.1

SNP Detail

SNP ID Chromosome Location
rs772303061 Chr.22: 46091238-46091238 on Build GRCh38
rs751374305 Chr.22: 46091014-46091014 on Build GRCh38
rs113999222 Chr.22: 46090962-46090962 on Build GRCh38
rs964489151 Chr.22: 46091279-46091279 on Build GRCh38
rs542476403 Chr.22: 46091260-46091260 on Build GRCh38
rs533405291 Chr.22: 46091280-46091280 on Build GRCh38
rs950899694 Chr.22: 46091301-46091301 on Build GRCh38
rs911137261 Chr.22: 46090930-46090930 on Build GRCh38
rs896846115 Chr.22: 46091177-46091177 on Build GRCh38
rs769715219 Chr.22: 46091355-46091355 on Build GRCh38
rs190040274 Chr.22: 46091218-46091218 on Build GRCh38
rs150279879 Chr.22: 46091103-46091103 on Build GRCh38
rs564505981 Chr.22: 46090960-46090960 on Build GRCh38
rs181112796 Chr.22: 46091267-46091267 on Build GRCh38
rs566266924 Chr.22: 46091092-46091092 on Build GRCh38
rs1018500730 Chr.22: 46091243-46091243 on Build GRCh38
rs564813794 Chr.22: 46091268-46091268 on Build GRCh38
rs60814799 Chr.22: 46091038-46091038 on Build GRCh38
rs1017245891 Chr.22: 46091274-46091274 on Build GRCh38
rs752680985 Chr.22: 46091325-46091325 on Build GRCh38
rs1026726145 Chr.22: 46091046-46091046 on Build GRCh38
rs917510288 Chr.22: 46091338-46091338 on Build GRCh38
rs1045199013 Chr.22: 46091125-46091125 on Build GRCh38
rs1018231890 Chr.22: 46091083-46091083 on Build GRCh38
rs755584928 Chr.22: 46091082-46091082 on Build GRCh38
rs564468798 Chr.22: 46091270-46091270 on Build GRCh38
rs569430328 Chr.22: 46091277-46091277 on Build GRCh38
rs759357010 Chr.22: 46091210-46091210 on Build GRCh38
rs1021064628 Chr.22: 46091068-46091068 on Build GRCh38
rs969419018 Chr.22: 46091294-46091294 on Build GRCh38
rs893943221 Chr.22: 46090964-46090964 on Build GRCh38
rs375724956 Chr.22: 46091158-46091158 on Build GRCh38
rs766999686 Chr.22: 46091063-46091063 on Build GRCh38
rs906761777 Chr.22: 46091008-46091008 on Build GRCh38
rs965644264 Chr.22: 46091087-46091087 on Build GRCh38
rs1009486096 Chr.22: 46091057-46091057 on Build GRCh38
rs1048194632 Chr.22: 46091199-46091199 on Build GRCh38
rs989559816 Chr.22: 46091107-46091107 on Build GRCh38
rs893624185 Chr.22: 46091233-46091233 on Build GRCh38
rs149416652 Chr.22: 46091362-46091362 on Build GRCh38
rs1035738129 Chr.22: 46091034-46091034 on Build GRCh38
rs76597937 Chr.22: 46091358-46091358 on Build GRCh38
rs987222533 Chr.22: 46090916-46090916 on Build GRCh38
rs761379155 Chr.22: 46091045-46091045 on Build GRCh38
rs943528935 Chr.22: 46090929-46090929 on Build GRCh38
rs910742039 Chr.22: 46090920-46090920 on Build GRCh38
rs998476744 Chr.22: 46091263-46091263 on Build GRCh38
rs7410377 Chr.22: 46091131-46091131 on Build GRCh38
rs748885940 Chr.22: 46091112-46091112 on Build GRCh38
rs112564068 Chr.22: 46091266-46091266 on Build GRCh38
rs77983430 Chr.22: 46091209-46091209 on Build GRCh38
rs571242189 Chr.22: 46091071-46091071 on Build GRCh38
rs7410618 Chr.22: 46091336-46091336 on Build GRCh38
rs1045329807 Chr.22: 46090980-46090980 on Build GRCh38
rs965326321 Chr.22: 46091254-46091254 on Build GRCh38
rs923899071 Chr.22: 46091099-46091099 on Build GRCh38
rs9615403 Chr.22: 46090906-46090906 on Build GRCh38
rs537220874 Chr.22: 46091332-46091332 on Build GRCh38
rs952562902 Chr.22: 46091075-46091075 on Build GRCh38
rs79348799 Chr.22: 46090961-46090961 on Build GRCh38
rs779269626 Chr.22: 46091130-46091130 on Build GRCh38
rs796389299 Chr.22: 46090971-46090971 on Build GRCh38
rs928086284 Chr.22: 46091334-46091334 on Build GRCh38
rs944075249 Chr.22: 46090932-46090932 on Build GRCh38
rs138879946 Chr.22: 46091192-46091192 on Build GRCh38
rs752698667 Chr.22: 46091074-46091074 on Build GRCh38
rs772649770 Chr.22: 46091079-46091079 on Build GRCh38
rs920848685 Chr.22: 46090934-46090934 on Build GRCh38
rs141392150 Chr.22: 46091033-46091033 on Build GRCh38
rs73888764 Chr.22: 46091116-46091116 on Build GRCh38
rs1042322750 Chr.22: 46091245-46091245 on Build GRCh38
rs939577435 Chr.22: 46091136-46091136 on Build GRCh38
rs1017369743 Chr.22: 46091090-46091090 on Build GRCh38
rs746816422 Chr.22: 46090954-46090954 on Build GRCh38

View Full Product Details