Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4749400-4749944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGOT
Entrez Gene ID
100126791
Gene Name
eosinophil granule ontogeny transcript (non-protein coding)
Gene Aliases
EGO, NCRNA00190
UniGene
Hs.663059
Gene Chromosome Location
Chr.3: 4749192-4751590 on Build GRCh38
Transcripts
NR_004428.1 NM_001099952.2 NM_001168272.1 NM_002222.5 XM_005265109.2 XM_005265110.2 XM_006713131.2 XM_011533681.1 XM_011533682.2 XM_011533683.2 XM_011533684.1 XM_011533685.1 XM_011533686.1 XM_011533687.1 XM_011533688.1 XM_011533690.1 XM_011533691.1 XM_017006357.1

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
UniGene
Hs.567295
Gene Chromosome Location
Chr.3: 4493348-4847840 on Build GRCh38
Transcripts
NR_004428.1 NM_001099952.2 NM_001168272.1 NM_002222.5 XM_005265109.2 XM_005265110.2 XM_006713131.2 XM_011533681.1 XM_011533682.2 XM_011533683.2 XM_011533684.1 XM_011533685.1 XM_011533686.1 XM_011533687.1 XM_011533688.1 XM_011533690.1 XM_011533691.1 XM_017006357.1

SNP Detail

SNP ID Chromosome Location
rs886679565 Chr.3: 4749566-4749566 on Build GRCh38
rs1040775211 Chr.3: 4749531-4749531 on Build GRCh38
rs922357495 Chr.3: 4749762-4749762 on Build GRCh38
rs1034306300 Chr.3: 4749502-4749502 on Build GRCh38
rs1001668295 Chr.3: 4749473-4749473 on Build GRCh38
rs574782610 Chr.3: 4749552-4749552 on Build GRCh38
rs538012010 Chr.3: 4749567-4749567 on Build GRCh38
rs1045973461 Chr.3: 4749754-4749754 on Build GRCh38
rs753696086 Chr.3: 4749459-4749459 on Build GRCh38
rs1025822731 Chr.3: 4749560-4749560 on Build GRCh38
rs184113636 Chr.3: 4749597-4749597 on Build GRCh38
rs1048383764 Chr.3: 4749564-4749564 on Build GRCh38
rs115806700 Chr.3: 4749826-4749826 on Build GRCh38
rs1012274548 Chr.3: 4749900-4749900 on Build GRCh38
rs1025421387 Chr.3: 4749915-4749915 on Build GRCh38
rs974457442 Chr.3: 4749618-4749618 on Build GRCh38
rs977935688 Chr.3: 4749661-4749661 on Build GRCh38
rs901978871 Chr.3: 4749543-4749543 on Build GRCh38
rs748491906 Chr.3: 4749817-4749817 on Build GRCh38
rs923900456 Chr.3: 4749431-4749431 on Build GRCh38
rs758567603 Chr.3: 4749494-4749494 on Build GRCh38
rs532328853 Chr.3: 4749617-4749617 on Build GRCh38
rs891738875 Chr.3: 4749427-4749427 on Build GRCh38
rs376615566 Chr.3: 4749507-4749507 on Build GRCh38
rs571150675 Chr.3: 4749686-4749686 on Build GRCh38
rs765781624 Chr.3: 4749737-4749737 on Build GRCh38
rs556303125 Chr.3: 4749728-4749728 on Build GRCh38
rs182099832 Chr.3: 4749430-4749430 on Build GRCh38
rs976447831 Chr.3: 4749706-4749706 on Build GRCh38
rs995484163 Chr.3: 4749535-4749535 on Build GRCh38
rs1025072798 Chr.3: 4749601-4749601 on Build GRCh38
rs576640776 Chr.3: 4749782-4749782 on Build GRCh38
rs572302482 Chr.3: 4749891-4749891 on Build GRCh38
rs995454370 Chr.3: 4749896-4749896 on Build GRCh38
rs1031281470 Chr.3: 4749668-4749668 on Build GRCh38
rs987352797 Chr.3: 4749580-4749580 on Build GRCh38
rs565866784 Chr.3: 4749679-4749679 on Build GRCh38
rs1016267491 Chr.3: 4749577-4749577 on Build GRCh38
rs942217836 Chr.3: 4749818-4749818 on Build GRCh38
rs369660582 Chr.3: 4749579-4749579 on Build GRCh38
rs1055782108 Chr.3: 4749821-4749821 on Build GRCh38
rs554629095 Chr.3: 4749657-4749657 on Build GRCh38
rs75325691 Chr.3: 4749897-4749897 on Build GRCh38
rs540702863 Chr.3: 4749568-4749568 on Build GRCh38
rs933521747 Chr.3: 4749556-4749556 on Build GRCh38
rs1047880327 Chr.3: 4749799-4749799 on Build GRCh38
rs142575313 Chr.3: 4749576-4749576 on Build GRCh38
rs929744029 Chr.3: 4749797-4749797 on Build GRCh38
rs117013927 Chr.3: 4749653-4749653 on Build GRCh38
rs147963507 Chr.3: 4749693-4749693 on Build GRCh38
rs141811745 Chr.3: 4749810-4749810 on Build GRCh38
rs942451280 Chr.3: 4749529-4749529 on Build GRCh38
rs547911569 Chr.3: 4749692-4749692 on Build GRCh38
rs79652400 Chr.3: 4749591-4749591 on Build GRCh38
rs747299287 Chr.3: 4749681-4749681 on Build GRCh38
rs536600358 Chr.3: 4749702-4749702 on Build GRCh38
rs201889933 Chr.3: 4749685-4749685 on Build GRCh38
rs748897887 Chr.3: 4749691-4749694 on Build GRCh38
rs936610271 Chr.3: 4749462-4749462 on Build GRCh38

View Full Product Details