Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 781255-781748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129917
Entrez Gene ID
100129917
Gene Name
uncharacterized LOC100129917
Gene Aliases
-
UniGene
Hs.731755
Gene Chromosome Location
Chr.4: 780149-781848 on Build GRCh38
Transcripts
NR_036511.1 NR_036512.1

SNP Detail

SNP ID Chromosome Location
rs940364499 Chr.4: 781599-781599 on Build GRCh38
rs891472189 Chr.4: 781320-781320 on Build GRCh38
rs1008787101 Chr.4: 781515-781515 on Build GRCh38
rs571840027 Chr.4: 781661-781661 on Build GRCh38
rs1046070174 Chr.4: 781454-781454 on Build GRCh38
rs908819214 Chr.4: 781591-781591 on Build GRCh38
rs79850364 Chr.4: 781440-781440 on Build GRCh38
rs544289979 Chr.4: 781608-781608 on Build GRCh38
rs1036584208 Chr.4: 781352-781352 on Build GRCh38
rs573069695 Chr.4: 781437-781437 on Build GRCh38
rs888929923 Chr.4: 781626-781626 on Build GRCh38
rs1006068070 Chr.4: 781705-781705 on Build GRCh38
rs905201972 Chr.4: 781725-781725 on Build GRCh38
rs988581450 Chr.4: 781462-781462 on Build GRCh38
rs1047138763 Chr.4: 781546-781546 on Build GRCh38
rs564726675 Chr.4: 781486-781486 on Build GRCh38
rs542870136 Chr.4: 781414-781414 on Build GRCh38
rs927633893 Chr.4: 781468-781468 on Build GRCh38
rs1026033457 Chr.4: 781399-781399 on Build GRCh38
rs930452440 Chr.4: 781504-781504 on Build GRCh38
rs906249298 Chr.4: 781323-781323 on Build GRCh38
rs991623116 Chr.4: 781550-781550 on Build GRCh38
rs753766812 Chr.4: 781633-781633 on Build GRCh38
rs766982959 Chr.4: 781533-781533 on Build GRCh38
rs1001464072 Chr.4: 781614-781614 on Build GRCh38
rs952744347 Chr.4: 781401-781401 on Build GRCh38
rs561119511 Chr.4: 781424-781424 on Build GRCh38
rs752384989 Chr.4: 781482-781482 on Build GRCh38
rs771142068 Chr.4: 781379-781379 on Build GRCh38
rs777046275 Chr.4: 781316-781316 on Build GRCh38
rs950665484 Chr.4: 781278-781278 on Build GRCh38
rs1053173884 Chr.4: 781319-781319 on Build GRCh38
rs1033435529 Chr.4: 781653-781653 on Build GRCh38
rs557532545 Chr.4: 781326-781326 on Build GRCh38
rs191900642 Chr.4: 781631-781631 on Build GRCh38
rs930401282 Chr.4: 781619-781619 on Build GRCh38
rs922894273 Chr.4: 781680-781680 on Build GRCh38
rs1008818122 Chr.4: 781393-781393 on Build GRCh38
rs34649712 Chr.4: 781630-781630 on Build GRCh38
rs910903511 Chr.4: 781662-781662 on Build GRCh38
rs920322353 Chr.4: 781615-781615 on Build GRCh38
rs765794621 Chr.4: 781361-781361 on Build GRCh38
rs994619144 Chr.4: 781542-781542 on Build GRCh38
rs971618616 Chr.4: 781580-781580 on Build GRCh38
rs1041286581 Chr.4: 781672-781672 on Build GRCh38
rs950288352 Chr.4: 781441-781441 on Build GRCh38
rs562287459 Chr.4: 781535-781535 on Build GRCh38
rs768545714 Chr.4: 781726-781726 on Build GRCh38
rs182277911 Chr.4: 781729-781729 on Build GRCh38
rs1047704720 Chr.4: 781690-781691 on Build GRCh38
rs1043145516 Chr.4: 781483-781483 on Build GRCh38
rs752276616 Chr.4: 781536-781536 on Build GRCh38
rs953297272 Chr.4: 781385-781385 on Build GRCh38
rs1050226102 Chr.4: 781505-781505 on Build GRCh38
rs1050192911 Chr.4: 781620-781620 on Build GRCh38
rs62294125 Chr.4: 781710-781710 on Build GRCh38
rs928355504 Chr.4: 781312-781312 on Build GRCh38
rs529728401 Chr.4: 781558-781558 on Build GRCh38
rs1043312548 Chr.4: 781371-781371 on Build GRCh38
rs998827627 Chr.4: 781645-781645 on Build GRCh38
rs755644067 Chr.4: 781567-781567 on Build GRCh38
rs940376419 Chr.4: 781477-781477 on Build GRCh38
rs575805721 Chr.4: 781380-781380 on Build GRCh38
rs1018913212 Chr.4: 781394-781394 on Build GRCh38
rs796872457 Chr.4: 781315-781315 on Build GRCh38
rs995110519 Chr.4: 781375-781375 on Build GRCh38
rs539083085 Chr.4: 781668-781668 on Build GRCh38
rs908999091 Chr.4: 781284-781284 on Build GRCh38
rs567822240 Chr.4: 781625-781625 on Build GRCh38
rs773443044 Chr.4: 781363-781363 on Build GRCh38
rs981643113 Chr.4: 781690-781690 on Build GRCh38
rs898946602 Chr.4: 781490-781490 on Build GRCh38
rs984731264 Chr.4: 781279-781279 on Build GRCh38
rs1015804913 Chr.4: 781699-781699 on Build GRCh38
rs549183334 Chr.4: 781611-781611 on Build GRCh38
rs35867050 Chr.4: 781670-781670 on Build GRCh38
rs981674221 Chr.4: 781581-781581 on Build GRCh38
rs994278470 Chr.4: 781396-781396 on Build GRCh38
rs1005536026 Chr.4: 781590-781590 on Build GRCh38
rs531938542 Chr.4: 781506-781506 on Build GRCh38
rs549385894 Chr.4: 781395-781420 on Build GRCh38
rs1023081127 Chr.4: 781673-781673 on Build GRCh38
rs763115839 Chr.4: 781373-781373 on Build GRCh38
rs947587773 Chr.4: 781355-781355 on Build GRCh38
rs766762194 Chr.4: 781423-781423 on Build GRCh38
rs778761934 Chr.4: 781727-781727 on Build GRCh38
rs971545141 Chr.4: 781685-781685 on Build GRCh38
rs947065077 Chr.4: 781476-781476 on Build GRCh38
rs979964382 Chr.4: 781300-781300 on Build GRCh38
rs926058397 Chr.4: 781422-781422 on Build GRCh38
rs141456974 Chr.4: 781700-781700 on Build GRCh38
rs903170385 Chr.4: 781522-781522 on Build GRCh38
rs146422431 Chr.4: 781517-781517 on Build GRCh38
rs967842739 Chr.4: 781397-781397 on Build GRCh38
rs1004650149 Chr.4: 781339-781339 on Build GRCh38
rs901244591 Chr.4: 781395-781395 on Build GRCh38
rs948530965 Chr.4: 781293-781293 on Build GRCh38

View Full Product Details