Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43042998-43043444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC153684
Entrez Gene ID
153684
Gene Name
uncharacterized LOC153684
Gene Aliases
-
UniGene
Hs.259625
Gene Chromosome Location
Chr.5: 43042134-43045268 on Build GRCh38
Transcripts
NR_015447.1 NR_104651.1

Gene Symbol
LOC648987
Entrez Gene ID
648987
Gene Name
uncharacterized LOC648987
Gene Aliases
-
UniGene
Hs.649468
Gene Chromosome Location
Chr.5: 43014729-43067439 on Build GRCh38
Transcripts
NR_015447.1 NR_104651.1

SNP Detail

SNP ID Chromosome Location
rs890746364 Chr.5: 43043094-43043094 on Build GRCh38
rs1014821418 Chr.5: 43043314-43043314 on Build GRCh38
rs946080667 Chr.5: 43043212-43043212 on Build GRCh38
rs897570905 Chr.5: 43043309-43043310 on Build GRCh38
rs545684569 Chr.5: 43043375-43043375 on Build GRCh38
rs987632790 Chr.5: 43043371-43043371 on Build GRCh38
rs749771439 Chr.5: 43043397-43043397 on Build GRCh38
rs947045864 Chr.5: 43043408-43043408 on Build GRCh38
rs905575426 Chr.5: 43043190-43043190 on Build GRCh38
rs990170388 Chr.5: 43043189-43043189 on Build GRCh38
rs549682111 Chr.5: 43043418-43043418 on Build GRCh38
rs541857429 Chr.5: 43043104-43043104 on Build GRCh38
rs963723686 Chr.5: 43043325-43043325 on Build GRCh38
rs566753439 Chr.5: 43043347-43043347 on Build GRCh38
rs973394217 Chr.5: 43043330-43043330 on Build GRCh38
rs1035516856 Chr.5: 43043282-43043282 on Build GRCh38
rs986034632 Chr.5: 43043183-43043183 on Build GRCh38
rs911359075 Chr.5: 43043184-43043184 on Build GRCh38
rs907155871 Chr.5: 43043235-43043235 on Build GRCh38
rs1052745557 Chr.5: 43043137-43043137 on Build GRCh38
rs143202736 Chr.5: 43043036-43043036 on Build GRCh38
rs1021803394 Chr.5: 43043164-43043164 on Build GRCh38
rs922880235 Chr.5: 43043123-43043123 on Build GRCh38
rs1014708592 Chr.5: 43043306-43043306 on Build GRCh38
rs912046629 Chr.5: 43043374-43043374 on Build GRCh38
rs1014615562 Chr.5: 43043052-43043052 on Build GRCh38
rs192616896 Chr.5: 43043098-43043098 on Build GRCh38
rs915466181 Chr.5: 43043399-43043399 on Build GRCh38
rs936102341 Chr.5: 43043185-43043185 on Build GRCh38
rs554508416 Chr.5: 43043078-43043078 on Build GRCh38
rs922011585 Chr.5: 43043034-43043034 on Build GRCh38
rs527628386 Chr.5: 43043354-43043354 on Build GRCh38
rs1006037902 Chr.5: 43043305-43043305 on Build GRCh38
rs913300813 Chr.5: 43043205-43043205 on Build GRCh38
rs531537418 Chr.5: 43043385-43043385 on Build GRCh38
rs1029957857 Chr.5: 43043326-43043326 on Build GRCh38
rs1001409958 Chr.5: 43043245-43043245 on Build GRCh38
rs1045726064 Chr.5: 43043229-43043229 on Build GRCh38
rs891436372 Chr.5: 43043161-43043161 on Build GRCh38
rs905647181 Chr.5: 43043419-43043419 on Build GRCh38
rs953460530 Chr.5: 43043370-43043370 on Build GRCh38
rs369509168 Chr.5: 43043080-43043080 on Build GRCh38
rs11949983 Chr.5: 43043286-43043286 on Build GRCh38
rs183611774 Chr.5: 43043384-43043384 on Build GRCh38
rs780577009 Chr.5: 43043163-43043163 on Build GRCh38
rs1004785081 Chr.5: 43043307-43043307 on Build GRCh38
rs913356192 Chr.5: 43043405-43043405 on Build GRCh38
rs937236409 Chr.5: 43043277-43043277 on Build GRCh38

View Full Product Details