Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35728042-35728577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP5
Entrez Gene ID
2289
Gene Name
FK506 binding protein 5
Gene Aliases
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35573585-35728583 on Build GRCh38
Transcripts
NM_001145775.2 NR_027117.2

Gene Symbol
LOC285847
Entrez Gene ID
285847
Gene Name
uncharacterized LOC285847
Gene Aliases
-
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35726762-35736947 on Build GRCh38
Transcripts
NM_001145775.2 NR_027117.2

SNP Detail

SNP ID Chromosome Location
rs965128822 Chr.6: 35728131-35728131 on Build GRCh38
rs536940586 Chr.6: 35728419-35728419 on Build GRCh38
rs868219508 Chr.6: 35728462-35728462 on Build GRCh38
rs1019415695 Chr.6: 35728092-35728092 on Build GRCh38
rs985081220 Chr.6: 35728262-35728262 on Build GRCh38
rs1006637180 Chr.6: 35728074-35728074 on Build GRCh38
rs543922897 Chr.6: 35728484-35728484 on Build GRCh38
rs980039252 Chr.6: 35728495-35728495 on Build GRCh38
rs960353523 Chr.6: 35728122-35728122 on Build GRCh38
rs891645225 Chr.6: 35728333-35728333 on Build GRCh38
rs950944030 Chr.6: 35728469-35728469 on Build GRCh38
rs569303390 Chr.6: 35728088-35728088 on Build GRCh38
rs903237537 Chr.6: 35728312-35728312 on Build GRCh38
rs1043365588 Chr.6: 35728470-35728470 on Build GRCh38
rs1043252265 Chr.6: 35728299-35728299 on Build GRCh38
rs938741138 Chr.6: 35728509-35728509 on Build GRCh38
rs889453242 Chr.6: 35728364-35728364 on Build GRCh38
rs947882567 Chr.6: 35728146-35728146 on Build GRCh38
rs928682843 Chr.6: 35728313-35728313 on Build GRCh38
rs900892712 Chr.6: 35728449-35728449 on Build GRCh38
rs916512187 Chr.6: 35728136-35728136 on Build GRCh38
rs971769864 Chr.6: 35728434-35728434 on Build GRCh38
rs1016511899 Chr.6: 35728477-35728477 on Build GRCh38
rs867250999 Chr.6: 35728357-35728357 on Build GRCh38
rs1023527193 Chr.6: 35728344-35728344 on Build GRCh38
rs961892062 Chr.6: 35728429-35728429 on Build GRCh38
rs985046364 Chr.6: 35728156-35728156 on Build GRCh38
rs969712753 Chr.6: 35728391-35728391 on Build GRCh38
rs953722212 Chr.6: 35728065-35728065 on Build GRCh38
rs910767462 Chr.6: 35728447-35728447 on Build GRCh38
rs1044455168 Chr.6: 35728302-35728302 on Build GRCh38
rs919448723 Chr.6: 35728251-35728251 on Build GRCh38
rs571830040 Chr.6: 35728293-35728293 on Build GRCh38
rs948754905 Chr.6: 35728294-35728294 on Build GRCh38
rs1035703500 Chr.6: 35728085-35728085 on Build GRCh38
rs923054468 Chr.6: 35728442-35728442 on Build GRCh38
rs1036104368 Chr.6: 35728214-35728214 on Build GRCh38
rs1043028761 Chr.6: 35728339-35728339 on Build GRCh38
rs530912838 Chr.6: 35728129-35728130 on Build GRCh38
rs35168508 Chr.6: 35728157-35728158 on Build GRCh38
rs868368971 Chr.6: 35728285-35728285 on Build GRCh38
rs1057208076 Chr.6: 35728531-35728531 on Build GRCh38
rs998935800 Chr.6: 35728318-35728318 on Build GRCh38
rs576488419 Chr.6: 35728465-35728465 on Build GRCh38
rs184711840 Chr.6: 35728329-35728329 on Build GRCh38
rs551967003 Chr.6: 35728252-35728252 on Build GRCh38
rs901514729 Chr.6: 35728215-35728215 on Build GRCh38
rs774698725 Chr.6: 35728194-35728194 on Build GRCh38
rs984555039 Chr.6: 35728412-35728412 on Build GRCh38
rs554512271 Chr.6: 35728331-35728331 on Build GRCh38
rs2817033 Chr.6: 35728502-35728502 on Build GRCh38
rs1050687251 Chr.6: 35728292-35728292 on Build GRCh38
rs2817034 Chr.6: 35728512-35728512 on Build GRCh38
rs1050534216 Chr.6: 35728446-35728446 on Build GRCh38
rs181279692 Chr.6: 35728195-35728195 on Build GRCh38
rs933005938 Chr.6: 35728241-35728242 on Build GRCh38
rs1011895177 Chr.6: 35728337-35728337 on Build GRCh38
rs555042472 Chr.6: 35728443-35728443 on Build GRCh38
rs779973484 Chr.6: 35728471-35728471 on Build GRCh38
rs938787403 Chr.6: 35728321-35728321 on Build GRCh38
rs572728436 Chr.6: 35728388-35728388 on Build GRCh38

View Full Product Details