Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16266256-16266803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISPD
Entrez Gene ID
729920
Gene Name
isoprenoid synthase domain containing
Gene Aliases
MDDGA7, MDDGC7, Nip, hCG_1745121
UniGene
Hs.636502
Gene Chromosome Location
Chr.7: 16087527-16422562 on Build GRCh38
Transcripts
NM_001101417.3 NM_001101426.3 XM_006715770.3 XM_011515499.2 XM_011515500.2 XM_011515508.2 XM_011515509.2 XM_017012575.1 XM_017012576.1 XM_017012577.1 XM_017012578.1 NR_038946.1 NR_038947.1

Gene Symbol
ISPD-AS1
Entrez Gene ID
100506025
Gene Name
ISPD antisense RNA 1
Gene Aliases
-
UniGene
Hs.649029
Gene Chromosome Location
Chr.7: 16210486-16270604 on Build GRCh38
Transcripts
NM_001101417.3 NM_001101426.3 XM_006715770.3 XM_011515499.2 XM_011515500.2 XM_011515508.2 XM_011515509.2 XM_017012575.1 XM_017012576.1 XM_017012577.1 XM_017012578.1 NR_038946.1 NR_038947.1

SNP Detail

SNP ID Chromosome Location
rs891058491 Chr.7: 16266732-16266732 on Build GRCh38
rs917517110 Chr.7: 16266664-16266664 on Build GRCh38
rs531165288 Chr.7: 16266470-16266470 on Build GRCh38
rs1009927189 Chr.7: 16266727-16266727 on Build GRCh38
rs1025613258 Chr.7: 16266607-16266607 on Build GRCh38
rs943338831 Chr.7: 16266353-16266353 on Build GRCh38
rs950723251 Chr.7: 16266769-16266769 on Build GRCh38
rs577455071 Chr.7: 16266742-16266742 on Build GRCh38
rs1295140 Chr.7: 16266537-16266537 on Build GRCh38
rs113338957 Chr.7: 16266702-16266702 on Build GRCh38
rs9639257 Chr.7: 16266525-16266525 on Build GRCh38
rs543808926 Chr.7: 16266350-16266350 on Build GRCh38
rs763499920 Chr.7: 16266743-16266743 on Build GRCh38
rs772932944 Chr.7: 16266583-16266583 on Build GRCh38
rs1056318578 Chr.7: 16266473-16266476 on Build GRCh38
rs559557851 Chr.7: 16266402-16266402 on Build GRCh38
rs1295141 Chr.7: 16266334-16266334 on Build GRCh38
rs1013940381 Chr.7: 16266416-16266416 on Build GRCh38
rs746734702 Chr.7: 16266340-16266340 on Build GRCh38
rs1044548130 Chr.7: 16266675-16266675 on Build GRCh38
rs537023584 Chr.7: 16266725-16266725 on Build GRCh38
rs189215088 Chr.7: 16266387-16266387 on Build GRCh38
rs924963360 Chr.7: 16266372-16266372 on Build GRCh38
rs987779360 Chr.7: 16266698-16266698 on Build GRCh38
rs372483393 Chr.7: 16266482-16266482 on Build GRCh38
rs770154587 Chr.7: 16266464-16266464 on Build GRCh38
rs542362976 Chr.7: 16266417-16266417 on Build GRCh38
rs770693507 Chr.7: 16266678-16266678 on Build GRCh38
rs950191698 Chr.7: 16266672-16266672 on Build GRCh38
rs920805871 Chr.7: 16266356-16266356 on Build GRCh38
rs973543156 Chr.7: 16266761-16266761 on Build GRCh38
rs920641463 Chr.7: 16266628-16266628 on Build GRCh38
rs929614557 Chr.7: 16266358-16266358 on Build GRCh38
rs911770795 Chr.7: 16266722-16266722 on Build GRCh38
rs1017668406 Chr.7: 16266614-16266614 on Build GRCh38
rs905568231 Chr.7: 16266677-16266677 on Build GRCh38
rs987805546 Chr.7: 16266380-16266380 on Build GRCh38
rs905050039 Chr.7: 16266463-16266463 on Build GRCh38
rs973810535 Chr.7: 16266625-16266625 on Build GRCh38
rs558941346 Chr.7: 16266731-16266731 on Build GRCh38
rs905625086 Chr.7: 16266457-16266457 on Build GRCh38
rs922467467 Chr.7: 16266312-16266312 on Build GRCh38
rs1019777144 Chr.7: 16266646-16266646 on Build GRCh38
rs748933578 Chr.7: 16266320-16266320 on Build GRCh38
rs1028049484 Chr.7: 16266768-16266768 on Build GRCh38
rs1000233560 Chr.7: 16266468-16266468 on Build GRCh38
rs995707746 Chr.7: 16266326-16266326 on Build GRCh38
rs898561793 Chr.7: 16266750-16266750 on Build GRCh38
rs554991617 Chr.7: 16266332-16266332 on Build GRCh38
rs982478233 Chr.7: 16266723-16266723 on Build GRCh38
rs985306615 Chr.7: 16266565-16266565 on Build GRCh38
rs748035726 Chr.7: 16266378-16266378 on Build GRCh38
rs374317266 Chr.7: 16266655-16266655 on Build GRCh38
rs887147996 Chr.7: 16266773-16266773 on Build GRCh38
rs558951399 Chr.7: 16266355-16266355 on Build GRCh38
rs181149347 Chr.7: 16266635-16266635 on Build GRCh38
rs866405958 Chr.7: 16266557-16266557 on Build GRCh38
rs371033650 Chr.7: 16266441-16266441 on Build GRCh38
rs1006689343 Chr.7: 16266739-16266739 on Build GRCh38
rs565920874 Chr.7: 16266638-16266638 on Build GRCh38
rs957789474 Chr.7: 16266697-16266697 on Build GRCh38
rs184867148 Chr.7: 16266694-16266694 on Build GRCh38
rs921603772 Chr.7: 16266476-16266476 on Build GRCh38
rs577488661 Chr.7: 16266373-16266373 on Build GRCh38
rs375969426 Chr.7: 16266469-16266469 on Build GRCh38
rs571800888 Chr.7: 16266406-16266406 on Build GRCh38
rs983691349 Chr.7: 16266649-16266649 on Build GRCh38
rs964827537 Chr.7: 16266622-16266622 on Build GRCh38
rs536454134 Chr.7: 16266683-16266683 on Build GRCh38
rs775720314 Chr.7: 16266714-16266714 on Build GRCh38
rs989010598 Chr.7: 16266307-16266307 on Build GRCh38
rs532465143 Chr.7: 16266621-16266621 on Build GRCh38
rs776879240 Chr.7: 16266685-16266685 on Build GRCh38
rs913601571 Chr.7: 16266701-16266701 on Build GRCh38
rs565002816 Chr.7: 16266601-16266601 on Build GRCh38
rs759722864 Chr.7: 16266696-16266696 on Build GRCh38
rs71549980 Chr.7: 16266482-16266483 on Build GRCh38
rs561312480 Chr.7: 16266710-16266710 on Build GRCh38

View Full Product Details