Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6406243-6406735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100287015
Entrez Gene ID
100287015
Gene Name
uncharacterized LOC100287015
Gene Aliases
-
UniGene
Hs.156928
Gene Chromosome Location
Chr.8: 6403556-6406548 on Build GRCh38
Transcripts
NR_040040.1 NM_001172574.1 NM_001172575.1 NM_001322042.1 NM_001322043.1 NM_024596.4 NR_136159.1 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Gene Symbol
MCPH1
Entrez Gene ID
79648
Gene Name
microcephalin 1
Gene Aliases
BRIT1, MCT
UniGene
Hs.593807
Gene Chromosome Location
Chr.8: 6406592-6648505 on Build GRCh38
Transcripts
NR_040040.1 NM_001172574.1 NM_001172575.1 NM_001322042.1 NM_001322043.1 NM_024596.4 NR_136159.1 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625699 c.22+2T>G p.? Chr.8: 6406691-6406691 on Build GRCh38
6034455 c.15C>T p.I5I Chr.8: 6406682-6406682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs984475656 Chr.8: 6406334-6406334 on Build GRCh38
rs748563799 Chr.8: 6406657-6406657 on Build GRCh38
rs542031255 Chr.8: 6406505-6406505 on Build GRCh38
rs2243755 Chr.8: 6406418-6406418 on Build GRCh38
rs1052136394 Chr.8: 6406695-6406695 on Build GRCh38
rs537445107 Chr.8: 6406466-6406466 on Build GRCh38
rs536903745 Chr.8: 6406613-6406613 on Build GRCh38
rs1028583777 Chr.8: 6406448-6406448 on Build GRCh38
rs2243675 Chr.8: 6406295-6406295 on Build GRCh38
rs1001569264 Chr.8: 6406390-6406390 on Build GRCh38
rs989349040 Chr.8: 6406524-6406524 on Build GRCh38
rs1044331477 Chr.8: 6406378-6406378 on Build GRCh38
rs559476098 Chr.8: 6406327-6406327 on Build GRCh38
rs368033201 Chr.8: 6406647-6406647 on Build GRCh38
rs2916656 Chr.8: 6406391-6406391 on Build GRCh38
rs573224960 Chr.8: 6406353-6406353 on Build GRCh38
rs376821749 Chr.8: 6406313-6406313 on Build GRCh38
rs755653944 Chr.8: 6406692-6406692 on Build GRCh38
rs534887526 Chr.8: 6406288-6406288 on Build GRCh38
rs776805571 Chr.8: 6406668-6406668 on Build GRCh38
rs971927414 Chr.8: 6406535-6406535 on Build GRCh38
rs760734024 Chr.8: 6406632-6406632 on Build GRCh38
rs1020018556 Chr.8: 6406611-6406611 on Build GRCh38
rs749562805 Chr.8: 6406397-6406397 on Build GRCh38
rs386721786 Chr.8: 6406286-6406295 on Build GRCh38
rs2305023 Chr.8: 6406609-6406609 on Build GRCh38
rs926032287 Chr.8: 6406360-6406360 on Build GRCh38
rs1043183126 Chr.8: 6406270-6406270 on Build GRCh38
rs901556758 Chr.8: 6406506-6406506 on Build GRCh38
rs895921848 Chr.8: 6406487-6406487 on Build GRCh38
rs933226810 Chr.8: 6406273-6406273 on Build GRCh38
rs768188163 Chr.8: 6406629-6406629 on Build GRCh38
rs375171907 Chr.8: 6406660-6406660 on Build GRCh38
rs140279025 Chr.8: 6406522-6406522 on Build GRCh38
rs757247746 Chr.8: 6406682-6406682 on Build GRCh38
rs1001623153 Chr.8: 6406291-6406291 on Build GRCh38
rs375905175 Chr.8: 6406504-6406504 on Build GRCh38
rs974554523 Chr.8: 6406355-6406355 on Build GRCh38
rs890156387 Chr.8: 6406605-6406605 on Build GRCh38
rs1054717109 Chr.8: 6406617-6406617 on Build GRCh38
rs954239926 Chr.8: 6406453-6406453 on Build GRCh38
rs780940062 Chr.8: 6406685-6406685 on Build GRCh38
rs757082597 Chr.8: 6406650-6406650 on Build GRCh38
rs1047218268 Chr.8: 6406554-6406554 on Build GRCh38
rs753805652 Chr.8: 6406639-6406639 on Build GRCh38
rs947296977 Chr.8: 6406469-6406469 on Build GRCh38
rs984685842 Chr.8: 6406565-6406565 on Build GRCh38
rs977400439 Chr.8: 6406263-6406263 on Build GRCh38
rs768366254 Chr.8: 6406707-6406707 on Build GRCh38
rs747298128 Chr.8: 6406711-6406711 on Build GRCh38
rs2243674 Chr.8: 6406286-6406286 on Build GRCh38
rs771277920 Chr.8: 6406663-6406663 on Build GRCh38
rs879629334 Chr.8: 6406490-6406490 on Build GRCh38
rs921471113 Chr.8: 6406265-6406265 on Build GRCh38
rs374802474 Chr.8: 6406628-6406628 on Build GRCh38
rs180999762 Chr.8: 6406275-6406275 on Build GRCh38
rs190692683 Chr.8: 6406587-6406587 on Build GRCh38
rs557425724 Chr.8: 6406315-6406315 on Build GRCh38
rs770466911 Chr.8: 6406620-6406620 on Build GRCh38
rs769260580 Chr.8: 6406627-6406627 on Build GRCh38
rs907773737 Chr.8: 6406566-6406566 on Build GRCh38
rs1010234589 Chr.8: 6406577-6406606 on Build GRCh38
rs1029117539 Chr.8: 6406508-6406508 on Build GRCh38
rs1024394582 Chr.8: 6406530-6406530 on Build GRCh38
rs372855229 Chr.8: 6406518-6406518 on Build GRCh38
rs60705475 Chr.8: 6406377-6406377 on Build GRCh38
rs905712897 Chr.8: 6406306-6406306 on Build GRCh38
rs571599428 Chr.8: 6406434-6406434 on Build GRCh38
rs778288309 Chr.8: 6406709-6406709 on Build GRCh38
rs149296889 Chr.8: 6406278-6406278 on Build GRCh38
rs988664603 Chr.8: 6406462-6406462 on Build GRCh38
rs763887070 Chr.8: 6406677-6406677 on Build GRCh38
rs745328654 Chr.8: 6406651-6406651 on Build GRCh38
rs994599408 Chr.8: 6406307-6406307 on Build GRCh38
rs577956608 Chr.8: 6406297-6406297 on Build GRCh38
rs191474064 Chr.8: 6406452-6406452 on Build GRCh38
rs375821563 Chr.8: 6406526-6406526 on Build GRCh38
rs952944691 Chr.8: 6406424-6406424 on Build GRCh38
rs930479630 Chr.8: 6406398-6406398 on Build GRCh38
rs1015931301 Chr.8: 6406427-6406427 on Build GRCh38
rs923094827 Chr.8: 6406358-6406358 on Build GRCh38
rs557033375 Chr.8: 6406477-6406477 on Build GRCh38
rs116171906 Chr.8: 6406495-6406495 on Build GRCh38
rs940536592 Chr.8: 6406575-6406575 on Build GRCh38
rs770646984 Chr.8: 6406492-6406492 on Build GRCh38
rs41302212 Chr.8: 6406319-6406319 on Build GRCh38
rs745545461 Chr.8: 6406623-6406623 on Build GRCh38
rs891773054 Chr.8: 6406277-6406277 on Build GRCh38
rs370544928 Chr.8: 6406542-6406542 on Build GRCh38
rs762645326 Chr.8: 6406455-6406455 on Build GRCh38
rs950086945 Chr.8: 6406476-6406476 on Build GRCh38
rs940505084 Chr.8: 6406500-6406500 on Build GRCh38
rs932795141 Chr.8: 6406268-6406268 on Build GRCh38
rs751243583 Chr.8: 6406681-6406681 on Build GRCh38
rs754406776 Chr.8: 6406625-6406625 on Build GRCh38
rs765495919 Chr.8: 6406675-6406675 on Build GRCh38
rs779918939 Chr.8: 6406701-6406701 on Build GRCh38
rs996217613 Chr.8: 6406444-6406444 on Build GRCh38
rs992846832 Chr.8: 6406544-6406544 on Build GRCh38
rs137972727 Chr.8: 6406369-6406369 on Build GRCh38
rs529012425 Chr.8: 6406562-6406562 on Build GRCh38
rs542825469 Chr.8: 6406326-6406326 on Build GRCh38
rs968543823 Chr.8: 6406538-6406538 on Build GRCh38
rs147133276 Chr.8: 6406384-6406384 on Build GRCh38
rs551427322 Chr.8: 6406595-6406595 on Build GRCh38
rs182865046 Chr.8: 6406618-6406618 on Build GRCh38
rs1050162308 Chr.8: 6406588-6406588 on Build GRCh38
rs549082812 Chr.8: 6406581-6406581 on Build GRCh38
rs201366514 Chr.8: 6406645-6406645 on Build GRCh38
rs749997244 Chr.8: 6406689-6406689 on Build GRCh38
rs766469448 Chr.8: 6406633-6406633 on Build GRCh38
rs903536382 Chr.8: 6406590-6406590 on Build GRCh38
rs927313293 Chr.8: 6406482-6406482 on Build GRCh38
rs954493876 Chr.8: 6406520-6406520 on Build GRCh38
rs977055674 Chr.8: 6406335-6406335 on Build GRCh38
rs984716845 Chr.8: 6406631-6406631 on Build GRCh38
rs964466462 Chr.8: 6406347-6406347 on Build GRCh38
rs528949402 Chr.8: 6406393-6406393 on Build GRCh38
rs776200803 Chr.8: 6406621-6406621 on Build GRCh38
rs1042991447 Chr.8: 6406470-6406470 on Build GRCh38
rs41312802 Chr.8: 6406603-6406603 on Build GRCh38
rs892129514 Chr.8: 6406714-6406714 on Build GRCh38
rs886265334 Chr.8: 6406416-6406416 on Build GRCh38
rs370540097 Chr.8: 6406648-6406648 on Build GRCh38
rs1024934748 Chr.8: 6406624-6406624 on Build GRCh38
rs373718276 Chr.8: 6406586-6406586 on Build GRCh38
rs908925077 Chr.8: 6406336-6406336 on Build GRCh38
rs947349222 Chr.8: 6406376-6406376 on Build GRCh38
rs553168307 Chr.8: 6406507-6406507 on Build GRCh38
rs917290285 Chr.8: 6406475-6406475 on Build GRCh38
rs759488210 Chr.8: 6406672-6406672 on Build GRCh38
rs762789401 Chr.8: 6406676-6406676 on Build GRCh38
rs576392182 Chr.8: 6406364-6406364 on Build GRCh38
rs927278151 Chr.8: 6406471-6406471 on Build GRCh38
rs988927020 Chr.8: 6406468-6406468 on Build GRCh38
rs994362032 Chr.8: 6406409-6406409 on Build GRCh38
rs185833251 Chr.8: 6406402-6406402 on Build GRCh38
rs1053188777 Chr.8: 6406365-6406365 on Build GRCh38
rs1018646813 Chr.8: 6406324-6406324 on Build GRCh38
rs755235337 Chr.8: 6406644-6406644 on Build GRCh38
rs182389021 Chr.8: 6406499-6406499 on Build GRCh38
rs187395120 Chr.8: 6406517-6406517 on Build GRCh38
rs773915060 Chr.8: 6406658-6406658 on Build GRCh38
rs1033214698 Chr.8: 6406461-6406461 on Build GRCh38
rs903515087 Chr.8: 6406509-6406509 on Build GRCh38
rs775260226 Chr.8: 6406652-6406652 on Build GRCh38
rs921493258 Chr.8: 6406344-6406344 on Build GRCh38
rs978892439 Chr.8: 6406357-6406357 on Build GRCh38
rs3020244 Chr.8: 6406432-6406432 on Build GRCh38
rs1051804911 Chr.8: 6406712-6406712 on Build GRCh38
rs115140989 Chr.8: 6406494-6406494 on Build GRCh38
rs1053416366 Chr.8: 6406368-6406368 on Build GRCh38
rs2243756 Chr.8: 6406426-6406426 on Build GRCh38
rs569795183 Chr.8: 6406262-6406262 on Build GRCh38
rs543406435 Chr.8: 6406540-6406540 on Build GRCh38
rs2916655 Chr.8: 6406552-6406552 on Build GRCh38
rs748935887 Chr.8: 6406702-6406702 on Build GRCh38

View Full Product Details