Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9901343-9901852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00599
Entrez Gene ID
157627
Gene Name
long intergenic non-protein coding RNA 599
Gene Aliases
Rncr3
UniGene
Hs.12513
Gene Chromosome Location
Chr.8: 9900064-9903329 on Build GRCh38
Transcripts
NR_024281.1

SNP Detail

SNP ID Chromosome Location
rs935568352 Chr.8: 9901427-9901428 on Build GRCh38
rs969623757 Chr.8: 9901688-9901688 on Build GRCh38
rs888299208 Chr.8: 9901513-9901513 on Build GRCh38
rs772673938 Chr.8: 9901434-9901434 on Build GRCh38
rs980617611 Chr.8: 9901611-9901611 on Build GRCh38
rs918317601 Chr.8: 9901750-9901750 on Build GRCh38
rs981006358 Chr.8: 9901705-9901705 on Build GRCh38
rs755238201 Chr.8: 9901744-9901744 on Build GRCh38
rs771399344 Chr.8: 9901524-9901524 on Build GRCh38
rs941379528 Chr.8: 9901649-9901649 on Build GRCh38
rs1026829407 Chr.8: 9901507-9901507 on Build GRCh38
rs541668384 Chr.8: 9901653-9901653 on Build GRCh38
rs915004027 Chr.8: 9901590-9901590 on Build GRCh38
rs118190748 Chr.8: 9901795-9901795 on Build GRCh38
rs929673115 Chr.8: 9901754-9901754 on Build GRCh38
rs1010583524 Chr.8: 9901761-9901761 on Build GRCh38
rs1031923548 Chr.8: 9901740-9901740 on Build GRCh38
rs1006714233 Chr.8: 9901645-9901645 on Build GRCh38
rs888404109 Chr.8: 9901773-9901773 on Build GRCh38
rs994052883 Chr.8: 9901500-9901500 on Build GRCh38
rs965839026 Chr.8: 9901525-9901525 on Build GRCh38
rs750353313 Chr.8: 9901625-9901625 on Build GRCh38
rs771434271 Chr.8: 9901477-9901477 on Build GRCh38
rs887191868 Chr.8: 9901450-9901450 on Build GRCh38
rs370712938 Chr.8: 9901718-9901718 on Build GRCh38
rs896826709 Chr.8: 9901609-9901609 on Build GRCh38
rs117422842 Chr.8: 9901438-9901438 on Build GRCh38
rs3735824 Chr.8: 9901387-9901387 on Build GRCh38
rs965431749 Chr.8: 9901418-9901418 on Build GRCh38
rs956477402 Chr.8: 9901558-9901558 on Build GRCh38
rs942616392 Chr.8: 9901781-9901781 on Build GRCh38
rs768056666 Chr.8: 9901689-9901689 on Build GRCh38
rs982971440 Chr.8: 9901369-9901369 on Build GRCh38
rs955647328 Chr.8: 9901548-9901548 on Build GRCh38
rs924252275 Chr.8: 9901423-9901423 on Build GRCh38
rs185190117 Chr.8: 9901613-9901613 on Build GRCh38
rs528777414 Chr.8: 9901415-9901415 on Build GRCh38
rs747499435 Chr.8: 9901451-9901451 on Build GRCh38
rs908470681 Chr.8: 9901818-9901818 on Build GRCh38
rs140312280 Chr.8: 9901543-9901543 on Build GRCh38
rs1031116136 Chr.8: 9901557-9901557 on Build GRCh38
rs561544129 Chr.8: 9901711-9901711 on Build GRCh38
rs567442646 Chr.8: 9901655-9901655 on Build GRCh38
rs941336550 Chr.8: 9901819-9901819 on Build GRCh38
rs989594105 Chr.8: 9901578-9901578 on Build GRCh38
rs530637254 Chr.8: 9901556-9901556 on Build GRCh38
rs1017022752 Chr.8: 9901483-9901483 on Build GRCh38
rs758747251 Chr.8: 9901411-9901411 on Build GRCh38
rs142097991 Chr.8: 9901388-9901388 on Build GRCh38
rs981339412 Chr.8: 9901571-9901571 on Build GRCh38
rs923698366 Chr.8: 9901405-9901405 on Build GRCh38
rs998007898 Chr.8: 9901820-9901820 on Build GRCh38
rs753615478 Chr.8: 9901732-9901732 on Build GRCh38
rs572567457 Chr.8: 9901651-9901651 on Build GRCh38
rs570629098 Chr.8: 9901673-9901673 on Build GRCh38
rs896778285 Chr.8: 9901493-9901493 on Build GRCh38
rs761200612 Chr.8: 9901597-9901597 on Build GRCh38
rs983071797 Chr.8: 9901810-9901810 on Build GRCh38
rs369734902 Chr.8: 9901650-9901650 on Build GRCh38
rs969149411 Chr.8: 9901772-9901772 on Build GRCh38
rs888351598 Chr.8: 9901626-9901626 on Build GRCh38
rs377352049 Chr.8: 9901670-9901670 on Build GRCh38
rs543303953 Chr.8: 9901439-9901439 on Build GRCh38
rs181675320 Chr.8: 9901376-9901376 on Build GRCh38
rs1029878316 Chr.8: 9901386-9901386 on Build GRCh38
rs908500300 Chr.8: 9901648-9901648 on Build GRCh38
rs1026776689 Chr.8: 9901392-9901392 on Build GRCh38
rs57559536 Chr.8: 9901447-9901447 on Build GRCh38
rs531039737 Chr.8: 9901594-9901594 on Build GRCh38
rs115139254 Chr.8: 9901801-9901801 on Build GRCh38
rs552804907 Chr.8: 9901382-9901382 on Build GRCh38
rs1006660339 Chr.8: 9901514-9901514 on Build GRCh38
rs386722120 Chr.8: 9901651-9901653 on Build GRCh38
rs563908575 Chr.8: 9901412-9901412 on Build GRCh38
rs575244950 Chr.8: 9901765-9901765 on Build GRCh38
rs1047055653 Chr.8: 9901644-9901644 on Build GRCh38
rs915097055 Chr.8: 9901419-9901419 on Build GRCh38
rs980586632 Chr.8: 9901790-9901790 on Build GRCh38
rs146698520 Chr.8: 9901478-9901478 on Build GRCh38
rs939310181 Chr.8: 9901446-9901446 on Build GRCh38
rs116349224 Chr.8: 9901547-9901547 on Build GRCh38
rs777070917 Chr.8: 9901490-9901490 on Build GRCh38

View Full Product Details