Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 99536444-99536942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR599
Entrez Gene ID
693184
Gene Name
microRNA 599
Gene Aliases
MIRN599, hsa-mir-599
UniGene
Gene Chromosome Location
Chr.8: 99536636-99536730 on Build GRCh38
Transcripts
NR_030329.1 NR_030596.1 NM_017890.4 NM_152564.4 XM_005250800.3 XM_005250801.4 XM_006716510.3 XM_011516848.2 XM_011516849.2 XM_011516850.2 XM_011516851.2 XM_011516852.2 XM_011516853.2 XM_011516854.2 XM_017013109.1 XM_017013110.1 XM_017013111.1 XM_017013112.1

Gene Symbol
MIR875
Entrez Gene ID
100126309
Gene Name
microRNA 875
Gene Aliases
MIRN875, hsa-mir-875
UniGene
Gene Chromosome Location
Chr.8: 99536786-99536861 on Build GRCh38
Transcripts
NR_030329.1 NR_030596.1 NM_017890.4 NM_152564.4 XM_005250800.3 XM_005250801.4 XM_006716510.3 XM_011516848.2 XM_011516849.2 XM_011516850.2 XM_011516851.2 XM_011516852.2 XM_011516853.2 XM_011516854.2 XM_017013109.1 XM_017013110.1 XM_017013111.1 XM_017013112.1

Gene Symbol
VPS13B
Entrez Gene ID
157680
Gene Name
vacuolar protein sorting 13 homolog B
Gene Aliases
CHS1, COH1
UniGene
Hs.191540
Gene Chromosome Location
Chr.8: 99007014-99877586 on Build GRCh38
Transcripts
NR_030329.1 NR_030596.1 NM_017890.4 NM_152564.4 XM_005250800.3 XM_005250801.4 XM_006716510.3 XM_011516848.2 XM_011516849.2 XM_011516850.2 XM_011516851.2 XM_011516852.2 XM_011516853.2 XM_011516854.2 XM_017013109.1 XM_017013110.1 XM_017013111.1 XM_017013112.1

SNP Detail

SNP ID Chromosome Location
rs771863090 Chr.8: 99536877-99536877 on Build GRCh38
rs769050384 Chr.8: 99536587-99536587 on Build GRCh38
rs759382255 Chr.8: 99536730-99536730 on Build GRCh38
rs770891289 Chr.8: 99536723-99536723 on Build GRCh38
rs139580121 Chr.8: 99536606-99536606 on Build GRCh38
rs201852061 Chr.8: 99536604-99536604 on Build GRCh38
rs774124620 Chr.8: 99536726-99536726 on Build GRCh38
rs774214391 Chr.8: 99536861-99536861 on Build GRCh38
rs879924428 Chr.8: 99536900-99536900 on Build GRCh38
rs777799140 Chr.8: 99536818-99536818 on Build GRCh38
rs905656948 Chr.8: 99536568-99536568 on Build GRCh38
rs754590999 Chr.8: 99536622-99536622 on Build GRCh38
rs779708931 Chr.8: 99536619-99536620 on Build GRCh38
rs759470413 Chr.8: 99536866-99536866 on Build GRCh38
rs186714401 Chr.8: 99536689-99536689 on Build GRCh38
rs761467201 Chr.8: 99536903-99536903 on Build GRCh38
rs967707574 Chr.8: 99536516-99536516 on Build GRCh38
rs949921731 Chr.8: 99536514-99536514 on Build GRCh38
rs768982184 Chr.8: 99536688-99536689 on Build GRCh38
rs746339976 Chr.8: 99536655-99536655 on Build GRCh38
rs749432375 Chr.8: 99536833-99536833 on Build GRCh38
rs767226332 Chr.8: 99536740-99536740 on Build GRCh38
rs895717173 Chr.8: 99536505-99536505 on Build GRCh38
rs1027990954 Chr.8: 99536852-99536852 on Build GRCh38
rs1009310575 Chr.8: 99536496-99536496 on Build GRCh38
rs747419225 Chr.8: 99536691-99536691 on Build GRCh38
rs1053219284 Chr.8: 99536481-99536481 on Build GRCh38
rs961812166 Chr.8: 99536676-99536676 on Build GRCh38
rs1044355227 Chr.8: 99536562-99536563 on Build GRCh38
rs752501838 Chr.8: 99536720-99536720 on Build GRCh38
rs765072944 Chr.8: 99536600-99536600 on Build GRCh38
rs770837891 Chr.8: 99536850-99536850 on Build GRCh38
rs762854608 Chr.8: 99536721-99536721 on Build GRCh38
rs760357710 Chr.8: 99536887-99536887 on Build GRCh38
rs767262593 Chr.8: 99536623-99536623 on Build GRCh38
rs556466307 Chr.8: 99536476-99536476 on Build GRCh38
rs751404076 Chr.8: 99536612-99536612 on Build GRCh38
rs755589721 Chr.8: 99536629-99536629 on Build GRCh38
rs775173592 Chr.8: 99536886-99536886 on Build GRCh38
rs16897440 Chr.8: 99536747-99536747 on Build GRCh38
rs994164426 Chr.8: 99536610-99536610 on Build GRCh38
rs867648290 Chr.8: 99536550-99536550 on Build GRCh38
rs752047399 Chr.8: 99536909-99536909 on Build GRCh38
rs534010484 Chr.8: 99536894-99536894 on Build GRCh38
rs748351737 Chr.8: 99536806-99536806 on Build GRCh38
rs181159989 Chr.8: 99536627-99536627 on Build GRCh38
rs758515132 Chr.8: 99536907-99536907 on Build GRCh38
rs762019641 Chr.8: 99536599-99536599 on Build GRCh38
rs538502143 Chr.8: 99536805-99536805 on Build GRCh38
rs753451230 Chr.8: 99536776-99536776 on Build GRCh38
rs780773850 Chr.8: 99536784-99536784 on Build GRCh38
rs1000558271 Chr.8: 99536812-99536812 on Build GRCh38
rs780547059 Chr.8: 99536690-99536690 on Build GRCh38
rs551946708 Chr.8: 99536777-99536777 on Build GRCh38
rs370517158 Chr.8: 99536753-99536753 on Build GRCh38
rs768995370 Chr.8: 99536699-99536699 on Build GRCh38
rs150575890 Chr.8: 99536541-99536541 on Build GRCh38
rs370015268 Chr.8: 99536719-99536719 on Build GRCh38
rs375364473 Chr.8: 99536706-99536706 on Build GRCh38
rs527379892 Chr.8: 99536537-99536537 on Build GRCh38
rs764855111 Chr.8: 99536905-99536905 on Build GRCh38
rs758999818 Chr.8: 99536677-99536677 on Build GRCh38
rs766111715 Chr.8: 99536609-99536609 on Build GRCh38
rs1019772625 Chr.8: 99536633-99536633 on Build GRCh38
rs781536773 Chr.8: 99536704-99536704 on Build GRCh38
rs1000509333 Chr.8: 99536543-99536543 on Build GRCh38
rs778947346 Chr.8: 99536797-99536797 on Build GRCh38
rs373553190 Chr.8: 99536651-99536651 on Build GRCh38
rs571545492 Chr.8: 99536889-99536889 on Build GRCh38
rs751961574 Chr.8: 99536791-99536791 on Build GRCh38
rs1020682664 Chr.8: 99536508-99536508 on Build GRCh38
rs776814180 Chr.8: 99536803-99536803 on Build GRCh38
rs373749237 Chr.8: 99536808-99536808 on Build GRCh38
rs200127975 Chr.8: 99536741-99536741 on Build GRCh38
rs776544630 Chr.8: 99536588-99536588 on Build GRCh38

View Full Product Details