Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 145003088-145003629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF252P-AS1
Entrez Gene ID
286103
Gene Name
ZNF252P antisense RNA 1
Gene Aliases
C8orf77
UniGene
Hs.359978
Gene Chromosome Location
Chr.8: 145002811-145006046 on Build GRCh38
Transcripts
NR_026974.1

SNP Detail

SNP ID Chromosome Location
rs549260484 Chr.8: 145003416-145003416 on Build GRCh38
rs1056687273 Chr.8: 145003108-145003108 on Build GRCh38
rs777026960 Chr.8: 145003421-145003421 on Build GRCh38
rs959819837 Chr.8: 145003554-145003554 on Build GRCh38
rs752431074 Chr.8: 145003271-145003271 on Build GRCh38
rs117777718 Chr.8: 145003166-145003166 on Build GRCh38
rs1027337132 Chr.8: 145003484-145003484 on Build GRCh38
rs750540165 Chr.8: 145003367-145003367 on Build GRCh38
rs780153654 Chr.8: 145003386-145003386 on Build GRCh38
rs769185489 Chr.8: 145003177-145003177 on Build GRCh38
rs964378634 Chr.8: 145003448-145003448 on Build GRCh38
rs527767923 Chr.8: 145003475-145003475 on Build GRCh38
rs759182169 Chr.8: 145003245-145003245 on Build GRCh38
rs540489465 Chr.8: 145003211-145003211 on Build GRCh38
rs545021684 Chr.8: 145003144-145003144 on Build GRCh38
rs904075376 Chr.8: 145003290-145003290 on Build GRCh38
rs772389349 Chr.8: 145003218-145003218 on Build GRCh38
rs567474218 Chr.8: 145003535-145003535 on Build GRCh38
rs776304499 Chr.8: 145003242-145003242 on Build GRCh38
rs756392036 Chr.8: 145003146-145003146 on Build GRCh38
rs1023842804 Chr.8: 145003113-145003113 on Build GRCh38
rs765506131 Chr.8: 145003584-145003584 on Build GRCh38
rs938011111 Chr.8: 145003390-145003390 on Build GRCh38
rs765383632 Chr.8: 145003392-145003393 on Build GRCh38
rs964034707 Chr.8: 145003164-145003164 on Build GRCh38
rs956752695 Chr.8: 145003149-145003149 on Build GRCh38
rs913761473 Chr.8: 145003320-145003320 on Build GRCh38
rs745457221 Chr.8: 145003167-145003167 on Build GRCh38
rs921062439 Chr.8: 145003107-145003107 on Build GRCh38
rs1053687131 Chr.8: 145003132-145003132 on Build GRCh38
rs192674046 Chr.8: 145003591-145003591 on Build GRCh38
rs779436265 Chr.8: 145003422-145003422 on Build GRCh38
rs913718844 Chr.8: 145003600-145003600 on Build GRCh38
rs561953723 Chr.8: 145003213-145003213 on Build GRCh38
rs118069926 Chr.8: 145003112-145003112 on Build GRCh38
rs906574029 Chr.8: 145003160-145003160 on Build GRCh38
rs745917921 Chr.8: 145003420-145003421 on Build GRCh38
rs761114854 Chr.8: 145003273-145003273 on Build GRCh38
rs938400845 Chr.8: 145003221-145003221 on Build GRCh38
rs562601991 Chr.8: 145003120-145003120 on Build GRCh38
rs778189389 Chr.8: 145003497-145003497 on Build GRCh38
rs886643706 Chr.8: 145003477-145003477 on Build GRCh38
rs921032236 Chr.8: 145003280-145003280 on Build GRCh38
rs189900127 Chr.8: 145003135-145003135 on Build GRCh38
rs77308683 Chr.8: 145003393-145003393 on Build GRCh38
rs1045711444 Chr.8: 145003268-145003268 on Build GRCh38
rs919800024 Chr.8: 145003169-145003169 on Build GRCh38
rs748727236 Chr.8: 145003217-145003217 on Build GRCh38
rs6989348 Chr.8: 145003233-145003233 on Build GRCh38
rs975092627 Chr.8: 145003279-145003279 on Build GRCh38
rs951014861 Chr.8: 145003235-145003235 on Build GRCh38
rs770757527 Chr.8: 145003109-145003109 on Build GRCh38
rs554122795 Chr.8: 145003580-145003580 on Build GRCh38
rs748480169 Chr.8: 145003430-145003430 on Build GRCh38
rs772806641 Chr.8: 145003229-145003229 on Build GRCh38
rs957869379 Chr.8: 145003285-145003286 on Build GRCh38
rs150520557 Chr.8: 145003251-145003251 on Build GRCh38
rs115055201 Chr.8: 145003404-145003404 on Build GRCh38
rs868723896 Chr.8: 145003173-145003173 on Build GRCh38
rs964991697 Chr.8: 145003527-145003527 on Build GRCh38
rs776103395 Chr.8: 145003602-145003602 on Build GRCh38
rs182411667 Chr.8: 145003194-145003194 on Build GRCh38
rs762883506 Chr.8: 145003297-145003297 on Build GRCh38
rs186685886 Chr.8: 145003342-145003342 on Build GRCh38
rs945275750 Chr.8: 145003323-145003323 on Build GRCh38
rs995425965 Chr.8: 145003229-145003229 on Build GRCh38
rs547876946 Chr.8: 145003571-145003571 on Build GRCh38
rs760018159 Chr.8: 145003128-145003128 on Build GRCh38
rs540738626 Chr.8: 145003158-145003158 on Build GRCh38
rs2294043 Chr.8: 145003236-145003236 on Build GRCh38
rs754228245 Chr.8: 145003304-145003304 on Build GRCh38
rs1046382956 Chr.8: 145003337-145003337 on Build GRCh38
rs908374941 Chr.8: 145003549-145003549 on Build GRCh38
rs992516355 Chr.8: 145003559-145003559 on Build GRCh38
rs780092505 Chr.8: 145003155-145003155 on Build GRCh38
rs756198379 Chr.8: 145003374-145003374 on Build GRCh38
rs952622649 Chr.8: 145003495-145003495 on Build GRCh38
rs138387081 Chr.8: 145003351-145003351 on Build GRCh38

View Full Product Details