Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 38377861-38378412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B7P2
Entrez Gene ID
158160
Gene Name
hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2
Gene Aliases
HSD17B7, Hsd17b_2, bA291L22.1
UniGene
Hs.652484
Gene Chromosome Location
Chr.10: 38356380-38378505 on Build GRCh38
Transcripts
NR_003086.1

SNP Detail

SNP ID Chromosome Location
rs893483376 Chr.10: 38378160-38378160 on Build GRCh38
rs1009054977 Chr.10: 38378193-38378193 on Build GRCh38
rs376528957 Chr.10: 38377905-38377905 on Build GRCh38
rs962150078 Chr.10: 38378238-38378238 on Build GRCh38
rs962468177 Chr.10: 38378011-38378011 on Build GRCh38
rs760360072 Chr.10: 38378368-38378369 on Build GRCh38
rs10827867 Chr.10: 38378388-38378388 on Build GRCh38
rs533735527 Chr.10: 38378249-38378249 on Build GRCh38
rs987938255 Chr.10: 38377915-38377915 on Build GRCh38
rs369212456 Chr.10: 38378358-38378358 on Build GRCh38
rs2804622 Chr.10: 38378075-38378075 on Build GRCh38
rs538997200 Chr.10: 38378296-38378296 on Build GRCh38
rs1029085714 Chr.10: 38377911-38377911 on Build GRCh38
rs544868327 Chr.10: 38377891-38377891 on Build GRCh38
rs916369879 Chr.10: 38378089-38378089 on Build GRCh38
rs947929585 Chr.10: 38378102-38378102 on Build GRCh38
rs935050197 Chr.10: 38378383-38378383 on Build GRCh38
rs577486479 Chr.10: 38378229-38378229 on Build GRCh38
rs768346348 Chr.10: 38378248-38378248 on Build GRCh38
rs569156594 Chr.10: 38378063-38378063 on Build GRCh38
rs552556317 Chr.10: 38378341-38378341 on Build GRCh38
rs972050898 Chr.10: 38378040-38378040 on Build GRCh38
rs540373858 Chr.10: 38377956-38377956 on Build GRCh38
rs906266912 Chr.10: 38378117-38378117 on Build GRCh38
rs2997758 Chr.10: 38377968-38377968 on Build GRCh38
rs935113977 Chr.10: 38378129-38378129 on Build GRCh38
rs553055603 Chr.10: 38378281-38378281 on Build GRCh38
rs537733234 Chr.10: 38378004-38378004 on Build GRCh38
rs1046681723 Chr.10: 38378104-38378104 on Build GRCh38
rs560241825 Chr.10: 38377967-38377967 on Build GRCh38
rs981957445 Chr.10: 38378361-38378361 on Build GRCh38
rs769889900 Chr.10: 38377928-38377928 on Build GRCh38
rs548670205 Chr.10: 38378048-38378048 on Build GRCh38
rs1029266234 Chr.10: 38378179-38378179 on Build GRCh38
rs1052013905 Chr.10: 38378146-38378146 on Build GRCh38
rs528908244 Chr.10: 38378030-38378030 on Build GRCh38
rs568588942 Chr.10: 38377988-38377988 on Build GRCh38
rs984012928 Chr.10: 38378326-38378326 on Build GRCh38
rs1019010629 Chr.10: 38377976-38377976 on Build GRCh38
rs889390721 Chr.10: 38378191-38378191 on Build GRCh38
rs953587106 Chr.10: 38377914-38377915 on Build GRCh38
rs916481843 Chr.10: 38378329-38378329 on Build GRCh38
rs754364394 Chr.10: 38378087-38378087 on Build GRCh38
rs972145497 Chr.10: 38378271-38378271 on Build GRCh38
rs551638136 Chr.10: 38378138-38378138 on Build GRCh38
rs1011086593 Chr.10: 38378164-38378164 on Build GRCh38
rs894050522 Chr.10: 38377884-38377884 on Build GRCh38
rs947992803 Chr.10: 38378352-38378352 on Build GRCh38
rs564556651 Chr.10: 38377920-38377920 on Build GRCh38
rs775115060 Chr.10: 38377973-38377973 on Build GRCh38
rs533554487 Chr.10: 38377940-38377940 on Build GRCh38
rs571149853 Chr.10: 38378139-38378139 on Build GRCh38
rs1011117764 Chr.10: 38377900-38377900 on Build GRCh38
rs748473873 Chr.10: 38378306-38378306 on Build GRCh38

View Full Product Details