Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 130106263-130106860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE7P
Entrez Gene ID
119437
Gene Name
CTAGE family member 7, pseudogene
Gene Aliases
CTAGE7, CTAGEP, bA500G10.2, rcCTAGE5
UniGene
Hs.620802
Gene Chromosome Location
Chr.10: 130106009-130108838 on Build GRCh38
Transcripts
NR_044994.1 NR_034125.1

Gene Symbol
LINC00959
Entrez Gene ID
387723
Gene Name
long intergenic non-protein coding RNA 959
Gene Aliases
-
UniGene
Hs.127394
Gene Chromosome Location
Chr.10: 130063896-130110817 on Build GRCh38
Transcripts
NR_044994.1 NR_034125.1

SNP Detail

SNP ID Chromosome Location
rs748097055 Chr.10: 130106642-130106642 on Build GRCh38
rs777553437 Chr.10: 130106491-130106492 on Build GRCh38
rs781401501 Chr.10: 130106769-130106769 on Build GRCh38
rs760096815 Chr.10: 130106349-130106349 on Build GRCh38
rs768961091 Chr.10: 130106807-130106807 on Build GRCh38
rs772614973 Chr.10: 130106818-130106818 on Build GRCh38
rs185618372 Chr.10: 130106547-130106547 on Build GRCh38
rs757344953 Chr.10: 130106676-130106676 on Build GRCh38
rs890403806 Chr.10: 130106823-130106826 on Build GRCh38
rs779282745 Chr.10: 130106808-130106808 on Build GRCh38
rs746599842 Chr.10: 130106495-130106496 on Build GRCh38
rs756893473 Chr.10: 130106510-130106510 on Build GRCh38
rs754928948 Chr.10: 130106640-130106640 on Build GRCh38
rs770287481 Chr.10: 130106504-130106508 on Build GRCh38
rs772747892 Chr.10: 130106735-130106735 on Build GRCh38
rs777976839 Chr.10: 130106538-130106538 on Build GRCh38
rs760351749 Chr.10: 130106736-130106736 on Build GRCh38
rs765897080 Chr.10: 130106401-130106402 on Build GRCh38
rs914725650 Chr.10: 130106574-130106574 on Build GRCh38
rs772763886 Chr.10: 130106822-130106822 on Build GRCh38
rs777122179 Chr.10: 130106481-130106481 on Build GRCh38
rs754293984 Chr.10: 130106323-130106323 on Build GRCh38
rs573906935 Chr.10: 130106601-130106601 on Build GRCh38
rs769180339 Chr.10: 130106718-130106719 on Build GRCh38
rs928284763 Chr.10: 130106375-130106375 on Build GRCh38
rs773427583 Chr.10: 130106302-130106302 on Build GRCh38
rs748624376 Chr.10: 130106816-130106816 on Build GRCh38
rs749539015 Chr.10: 130106794-130106794 on Build GRCh38
rs747143224 Chr.10: 130106544-130106544 on Build GRCh38
rs773851124 Chr.10: 130106570-130106570 on Build GRCh38
rs753663063 Chr.10: 130106744-130106744 on Build GRCh38
rs781203651 Chr.10: 130106814-130106814 on Build GRCh38
rs749208637 Chr.10: 130106534-130106534 on Build GRCh38
rs779736221 Chr.10: 130106525-130106525 on Build GRCh38
rs750027918 Chr.10: 130106407-130106407 on Build GRCh38
rs779976399 Chr.10: 130106445-130106445 on Build GRCh38
rs767028483 Chr.10: 130106615-130106615 on Build GRCh38
rs756946265 Chr.10: 130106400-130106400 on Build GRCh38
rs777158452 Chr.10: 130106377-130106377 on Build GRCh38
rs540710970 Chr.10: 130106781-130106781 on Build GRCh38
rs775913557 Chr.10: 130106695-130106695 on Build GRCh38
rs907602312 Chr.10: 130106758-130106758 on Build GRCh38
rs766164506 Chr.10: 130106632-130106632 on Build GRCh38
rs769961123 Chr.10: 130106692-130106692 on Build GRCh38
rs746060013 Chr.10: 130106681-130106681 on Build GRCh38
rs773195611 Chr.10: 130106467-130106467 on Build GRCh38
rs573373861 Chr.10: 130106767-130106767 on Build GRCh38
rs938394848 Chr.10: 130106390-130106390 on Build GRCh38
rs529069355 Chr.10: 130106496-130106496 on Build GRCh38
rs775883146 Chr.10: 130106506-130106507 on Build GRCh38
rs563291441 Chr.10: 130106475-130106475 on Build GRCh38
rs774330842 Chr.10: 130106720-130106721 on Build GRCh38
rs766807290 Chr.10: 130106319-130106319 on Build GRCh38
rs759182989 Chr.10: 130106745-130106745 on Build GRCh38
rs754910419 Chr.10: 130106535-130106535 on Build GRCh38
rs762330432 Chr.10: 130106734-130106734 on Build GRCh38
rs60998497 Chr.10: 130106523-130106523 on Build GRCh38
rs758823829 Chr.10: 130106344-130106345 on Build GRCh38
rs866974664 Chr.10: 130106469-130106469 on Build GRCh38
rs149953039 Chr.10: 130106792-130106792 on Build GRCh38
rs769016455 Chr.10: 130106705-130106705 on Build GRCh38
rs769614708 Chr.10: 130106433-130106433 on Build GRCh38
rs34308825 Chr.10: 130106765-130106766 on Build GRCh38
rs1052105519 Chr.10: 130106804-130106804 on Build GRCh38
rs746099679 Chr.10: 130106546-130106546 on Build GRCh38
rs749686393 Chr.10: 130106696-130106696 on Build GRCh38
rs955551236 Chr.10: 130106505-130106505 on Build GRCh38
rs763496566 Chr.10: 130106561-130106561 on Build GRCh38
rs565758972 Chr.10: 130106512-130106512 on Build GRCh38
rs190135923 Chr.10: 130106753-130106753 on Build GRCh38
rs765827934 Chr.10: 130106362-130106362 on Build GRCh38
rs772143110 Chr.10: 130106453-130106453 on Build GRCh38
rs774782318 Chr.10: 130106720-130106720 on Build GRCh38
rs553505155 Chr.10: 130106764-130106764 on Build GRCh38
rs900383673 Chr.10: 130106474-130106474 on Build GRCh38
rs763631530 Chr.10: 130106502-130106502 on Build GRCh38
rs757855468 Chr.10: 130106372-130106372 on Build GRCh38
rs767228564 Chr.10: 130106515-130106515 on Build GRCh38
rs549114830 Chr.10: 130106504-130106504 on Build GRCh38
rs970289086 Chr.10: 130106595-130106595 on Build GRCh38
rs770479166 Chr.10: 130106837-130106837 on Build GRCh38
rs760998707 Chr.10: 130106307-130106307 on Build GRCh38
rs777300203 Chr.10: 130106623-130106623 on Build GRCh38
rs745724302 Chr.10: 130106418-130106418 on Build GRCh38
rs557675159 Chr.10: 130106634-130106634 on Build GRCh38
rs549644603 Chr.10: 130106448-130106448 on Build GRCh38
rs764571845 Chr.10: 130106568-130106568 on Build GRCh38
rs537487126 Chr.10: 130106560-130106560 on Build GRCh38
rs760168717 Chr.10: 130106835-130106835 on Build GRCh38
rs765133363 Chr.10: 130106752-130106752 on Build GRCh38
rs755882843 Chr.10: 130106524-130106524 on Build GRCh38
rs755755694 Chr.10: 130106629-130106629 on Build GRCh38
rs752139887 Chr.10: 130106364-130106364 on Build GRCh38
rs751175348 Chr.10: 130106383-130106383 on Build GRCh38
rs765521236 Chr.10: 130106492-130106492 on Build GRCh38
rs1177699 Chr.10: 130106545-130106545 on Build GRCh38
rs1008210372 Chr.10: 130106832-130106832 on Build GRCh38
rs941793590 Chr.10: 130106766-130106766 on Build GRCh38
rs760946969 Chr.10: 130106473-130106473 on Build GRCh38
rs771892224 Chr.10: 130106480-130106481 on Build GRCh38
rs996499176 Chr.10: 130106479-130106479 on Build GRCh38
rs781440297 Chr.10: 130106677-130106677 on Build GRCh38
rs763388804 Chr.10: 130106501-130106501 on Build GRCh38
rs763388463 Chr.10: 130106715-130106716 on Build GRCh38
rs780947078 Chr.10: 130106410-130106410 on Build GRCh38
rs756359095 Chr.10: 130106787-130106787 on Build GRCh38
rs574237186 Chr.10: 130106741-130106741 on Build GRCh38

View Full Product Details