Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100156441-100157028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2P5
Entrez Gene ID
55592
Gene Name
golgin A2 pseudogene 5
Gene Aliases
GOLGA2B, GOLGA2L1
UniGene
Hs.524660
Gene Chromosome Location
Chr.12: 100156391-100173343 on Build GRCh38
Transcripts
NR_036632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3998633 c.414A>G p.G138G Chr.12: 100157027-100157027 on Build GRCh38
4846293 c.415C>G p.R139G Chr.12: 100157026-100157026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560647505 Chr.12: 100156849-100156849 on Build GRCh38
rs953755886 Chr.12: 100156969-100156969 on Build GRCh38
rs573381933 Chr.12: 100156736-100156736 on Build GRCh38
rs1043258356 Chr.12: 100156774-100156774 on Build GRCh38
rs1043228211 Chr.12: 100156532-100156532 on Build GRCh38
rs892443630 Chr.12: 100156735-100156735 on Build GRCh38
rs912131211 Chr.12: 100156678-100156678 on Build GRCh38
rs539257625 Chr.12: 100156581-100156581 on Build GRCh38
rs969844260 Chr.12: 100156640-100156641 on Build GRCh38
rs896402039 Chr.12: 100156791-100156791 on Build GRCh38
rs980184699 Chr.12: 100156682-100156683 on Build GRCh38
rs546093167 Chr.12: 100156859-100156859 on Build GRCh38
rs960443315 Chr.12: 100156507-100156507 on Build GRCh38
rs986709464 Chr.12: 100156971-100156971 on Build GRCh38
rs116667659 Chr.12: 100156781-100156781 on Build GRCh38
rs773323737 Chr.12: 100156548-100156549 on Build GRCh38
rs1052877359 Chr.12: 100156711-100156711 on Build GRCh38
rs543436915 Chr.12: 100156634-100156634 on Build GRCh38
rs879758155 Chr.12: 100156873-100156873 on Build GRCh38
rs979964634 Chr.12: 100156970-100156972 on Build GRCh38
rs961620797 Chr.12: 100156612-100156612 on Build GRCh38
rs973771635 Chr.12: 100156584-100156584 on Build GRCh38
rs767486761 Chr.12: 100156883-100156883 on Build GRCh38
rs150859784 Chr.12: 100156980-100156980 on Build GRCh38
rs973824167 Chr.12: 100156865-100156865 on Build GRCh38
rs1027997776 Chr.12: 100156563-100156563 on Build GRCh38
rs996567494 Chr.12: 100156550-100156550 on Build GRCh38
rs920787536 Chr.12: 100156613-100156613 on Build GRCh38
rs908170596 Chr.12: 100156546-100156546 on Build GRCh38
rs1026864136 Chr.12: 100156954-100156954 on Build GRCh38
rs557602651 Chr.12: 100156957-100156960 on Build GRCh38
rs928750594 Chr.12: 100156694-100156694 on Build GRCh38
rs34923288 Chr.12: 100156956-100156956 on Build GRCh38
rs905062570 Chr.12: 100156752-100156752 on Build GRCh38
rs576243423 Chr.12: 100156615-100156615 on Build GRCh38
rs900652858 Chr.12: 100156535-100156535 on Build GRCh38
rs565839496 Chr.12: 100156526-100156526 on Build GRCh38
rs773960610 Chr.12: 100156775-100156775 on Build GRCh38
rs148343041 Chr.12: 100156583-100156583 on Build GRCh38
rs747454393 Chr.12: 100156973-100156973 on Build GRCh38
rs35900949 Chr.12: 100156580-100156581 on Build GRCh38
rs535775134 Chr.12: 100156469-100156469 on Build GRCh38
rs1050738137 Chr.12: 100156669-100156669 on Build GRCh38
rs1011816212 Chr.12: 100156508-100156509 on Build GRCh38
rs1004106816 Chr.12: 100156808-100156808 on Build GRCh38
rs542475841 Chr.12: 100156842-100156842 on Build GRCh38
rs1028025444 Chr.12: 100156951-100156951 on Build GRCh38
rs944941271 Chr.12: 100156680-100156680 on Build GRCh38
rs890906980 Chr.12: 100156838-100156839 on Build GRCh38
rs764984332 Chr.12: 100156967-100156967 on Build GRCh38
rs539940350 Chr.12: 100156810-100156810 on Build GRCh38
rs776499031 Chr.12: 100156955-100156955 on Build GRCh38
rs761644815 Chr.12: 100156960-100156960 on Build GRCh38
rs925333038 Chr.12: 100156776-100156776 on Build GRCh38
rs744508 Chr.12: 100156684-100156684 on Build GRCh38
rs1043819233 Chr.12: 100156746-100156746 on Build GRCh38
rs1015253290 Chr.12: 100156590-100156590 on Build GRCh38
rs1002036173 Chr.12: 100156773-100156773 on Build GRCh38
rs947655824 Chr.12: 100156757-100156757 on Build GRCh38
rs982858948 Chr.12: 100156540-100156540 on Build GRCh38
rs141475210 Chr.12: 100156851-100156851 on Build GRCh38
rs995436280 Chr.12: 100156950-100156950 on Build GRCh38
rs553797584 Chr.12: 100156500-100156500 on Build GRCh38
rs1051937339 Chr.12: 100156819-100156819 on Build GRCh38

View Full Product Details