Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103866339-103866933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC41P
Entrez Gene ID
253724
Gene Name
tetratricopeptide repeat domain 41, pseudogene
Gene Aliases
GNN, GNNP
UniGene
Hs.506549
Gene Chromosome Location
Chr.12: 103843749-103930211 on Build GRCh38
Transcripts
NR_027249.1

SNP Detail

SNP ID Chromosome Location
rs1014777131 Chr.12: 103866400-103866400 on Build GRCh38
rs959422157 Chr.12: 103866544-103866544 on Build GRCh38
rs991291341 Chr.12: 103866559-103866559 on Build GRCh38
rs571506865 Chr.12: 103866437-103866437 on Build GRCh38
rs554172343 Chr.12: 103866875-103866875 on Build GRCh38
rs11111820 Chr.12: 103866738-103866738 on Build GRCh38
rs921540139 Chr.12: 103866597-103866597 on Build GRCh38
rs776203327 Chr.12: 103866429-103866430 on Build GRCh38
rs140047015 Chr.12: 103866505-103866505 on Build GRCh38
rs867223302 Chr.12: 103866540-103866540 on Build GRCh38
rs996278454 Chr.12: 103866367-103866367 on Build GRCh38
rs550994826 Chr.12: 103866499-103866499 on Build GRCh38
rs189515481 Chr.12: 103866401-103866401 on Build GRCh38
rs866782339 Chr.12: 103866360-103866360 on Build GRCh38
rs1021590594 Chr.12: 103866886-103866886 on Build GRCh38
rs977024353 Chr.12: 103866591-103866591 on Build GRCh38
rs11612613 Chr.12: 103866430-103866430 on Build GRCh38
rs931646159 Chr.12: 103866611-103866611 on Build GRCh38
rs755929530 Chr.12: 103866384-103866384 on Build GRCh38
rs1012891389 Chr.12: 103866882-103866882 on Build GRCh38
rs977527599 Chr.12: 103866525-103866525 on Build GRCh38
rs943535661 Chr.12: 103866402-103866402 on Build GRCh38
rs1035594691 Chr.12: 103866839-103866839 on Build GRCh38
rs10778292 Chr.12: 103866509-103866509 on Build GRCh38
rs777885221 Chr.12: 103866387-103866387 on Build GRCh38
rs761145097 Chr.12: 103866519-103866519 on Build GRCh38
rs557799384 Chr.12: 103866431-103866431 on Build GRCh38
rs566724280 Chr.12: 103866405-103866405 on Build GRCh38
rs2248382 Chr.12: 103866908-103866908 on Build GRCh38
rs887494455 Chr.12: 103866665-103866665 on Build GRCh38
rs573032697 Chr.12: 103866906-103866906 on Build GRCh38
rs1026768121 Chr.12: 103866469-103866469 on Build GRCh38
rs977356658 Chr.12: 103866664-103866664 on Build GRCh38
rs181816025 Chr.12: 103866593-103866593 on Build GRCh38
rs796076399 Chr.12: 103866763-103866763 on Build GRCh38
rs935781375 Chr.12: 103866366-103866366 on Build GRCh38
rs573798247 Chr.12: 103866703-103866703 on Build GRCh38
rs1022189118 Chr.12: 103866822-103866822 on Build GRCh38
rs929577430 Chr.12: 103866856-103866856 on Build GRCh38
rs1021690352 Chr.12: 103866508-103866508 on Build GRCh38
rs766189040 Chr.12: 103866423-103866423 on Build GRCh38
rs897659204 Chr.12: 103866835-103866835 on Build GRCh38
rs1050530842 Chr.12: 103866634-103866634 on Build GRCh38
rs1050168834 Chr.12: 103866374-103866374 on Build GRCh38
rs968918071 Chr.12: 103866833-103866833 on Build GRCh38
rs902103834 Chr.12: 103866424-103866424 on Build GRCh38
rs941766861 Chr.12: 103866668-103866668 on Build GRCh38
rs369016953 Chr.12: 103866883-103866883 on Build GRCh38
rs943064603 Chr.12: 103866806-103866806 on Build GRCh38
rs542718631 Chr.12: 103866715-103866715 on Build GRCh38
rs756198902 Chr.12: 103866381-103866381 on Build GRCh38
rs924787784 Chr.12: 103866534-103866534 on Build GRCh38
rs910316703 Chr.12: 103866765-103866765 on Build GRCh38
rs944251387 Chr.12: 103866557-103866557 on Build GRCh38
rs548540416 Chr.12: 103866837-103866837 on Build GRCh38
rs1024055073 Chr.12: 103866577-103866577 on Build GRCh38
rs1040565666 Chr.12: 103866808-103866808 on Build GRCh38
rs747654981 Chr.12: 103866843-103866843 on Build GRCh38
rs1003149763 Chr.12: 103866836-103866836 on Build GRCh38
rs4964466 Chr.12: 103866789-103866789 on Build GRCh38
rs769281564 Chr.12: 103866574-103866574 on Build GRCh38
rs751386633 Chr.12: 103866375-103866375 on Build GRCh38

View Full Product Details