Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52504015-52504556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPTE2P3
Entrez Gene ID
220115
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 3
Gene Aliases
TPTEps1
UniGene
Hs.528448
Gene Chromosome Location
Chr.13: 52488993-52587090 on Build GRCh38
Transcripts
NR_002793.1

SNP Detail

SNP ID Chromosome Location
rs563171990 Chr.13: 52504459-52504459 on Build GRCh38
rs1038991407 Chr.13: 52504222-52504222 on Build GRCh38
rs576750500 Chr.13: 52504482-52504482 on Build GRCh38
rs939100664 Chr.13: 52504436-52504436 on Build GRCh38
rs900032029 Chr.13: 52504223-52504223 on Build GRCh38
rs989448305 Chr.13: 52504338-52504338 on Build GRCh38
rs886748906 Chr.13: 52504139-52504139 on Build GRCh38
rs762717796 Chr.13: 52504329-52504329 on Build GRCh38
rs373902764 Chr.13: 52504229-52504229 on Build GRCh38
rs927751358 Chr.13: 52504434-52504434 on Build GRCh38
rs560457879 Chr.13: 52504275-52504275 on Build GRCh38
rs941014316 Chr.13: 52504465-52504465 on Build GRCh38
rs535664292 Chr.13: 52504053-52504053 on Build GRCh38
rs908214649 Chr.13: 52504463-52504463 on Build GRCh38
rs998873136 Chr.13: 52504311-52504311 on Build GRCh38
rs150753997 Chr.13: 52504372-52504372 on Build GRCh38
rs955469498 Chr.13: 52504282-52504282 on Build GRCh38
rs756152656 Chr.13: 52504317-52504317 on Build GRCh38
rs969274849 Chr.13: 52504356-52504356 on Build GRCh38
rs149884020 Chr.13: 52504455-52504455 on Build GRCh38
rs1031636020 Chr.13: 52504322-52504322 on Build GRCh38
rs776596626 Chr.13: 52504422-52504422 on Build GRCh38
rs574242810 Chr.13: 52504350-52504350 on Build GRCh38
rs1046806218 Chr.13: 52504438-52504438 on Build GRCh38
rs769903542 Chr.13: 52504107-52504107 on Build GRCh38
rs900064879 Chr.13: 52504531-52504531 on Build GRCh38
rs997420238 Chr.13: 52504237-52504237 on Build GRCh38
rs939025757 Chr.13: 52504098-52504098 on Build GRCh38
rs1005804174 Chr.13: 52504185-52504185 on Build GRCh38
rs555495678 Chr.13: 52504236-52504236 on Build GRCh38
rs1038654530 Chr.13: 52504477-52504477 on Build GRCh38
rs1029731192 Chr.13: 52504276-52504276 on Build GRCh38
rs927677938 Chr.13: 52504055-52504055 on Build GRCh38
rs1046775646 Chr.13: 52504129-52504129 on Build GRCh38
rs529214708 Chr.13: 52504388-52504388 on Build GRCh38

View Full Product Details