Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53380966-53381527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF525
Entrez Gene ID
170958
Gene Name
zinc finger protein 525
Gene Aliases
-
UniGene
Hs.352638
Gene Chromosome Location
Chr.19: 53365715-53386590 on Build GRCh38
Transcripts
NR_003699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6830942 c.480T>C p.I160I Chr.19: 53381059-53381059 on Build GRCh38
6830941 c.792T>C p.C264C Chr.19: 53381371-53381371 on Build GRCh38
3960425 c.442C>G p.L148V Chr.19: 53381021-53381021 on Build GRCh38
3971215 c.501T>C p.S167S Chr.19: 53381080-53381080 on Build GRCh38
341158 c.398A>G p.N133S Chr.19: 53380977-53380977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934438786 Chr.19: 53381465-53381465 on Build GRCh38
rs772669479 Chr.19: 53381383-53381383 on Build GRCh38
rs775597187 Chr.19: 53381327-53381327 on Build GRCh38
rs759827344 Chr.19: 53381098-53381098 on Build GRCh38
rs773058648 Chr.19: 53381196-53381196 on Build GRCh38
rs556692444 Chr.19: 53381194-53381194 on Build GRCh38
rs753369536 Chr.19: 53381345-53381345 on Build GRCh38
rs938451119 Chr.19: 53381026-53381026 on Build GRCh38
rs765834246 Chr.19: 53381328-53381329 on Build GRCh38
rs534837444 Chr.19: 53381097-53381097 on Build GRCh38
rs750332995 Chr.19: 53381428-53381428 on Build GRCh38
rs763774827 Chr.19: 53381413-53381413 on Build GRCh38
rs779882333 Chr.19: 53381013-53381013 on Build GRCh38
rs938644252 Chr.19: 53381168-53381168 on Build GRCh38
rs955667008 Chr.19: 53381002-53381002 on Build GRCh38
rs560595760 Chr.19: 53381223-53381223 on Build GRCh38
rs45625239 Chr.19: 53381350-53381350 on Build GRCh38
rs757065189 Chr.19: 53381424-53381424 on Build GRCh38
rs747939095 Chr.19: 53381310-53381310 on Build GRCh38
rs1056112711 Chr.19: 53381187-53381187 on Build GRCh38
rs746852204 Chr.19: 53381357-53381357 on Build GRCh38
rs549678748 Chr.19: 53381373-53381373 on Build GRCh38
rs183665728 Chr.19: 53381363-53381363 on Build GRCh38
rs753255604 Chr.19: 53381375-53381399 on Build GRCh38
rs553418942 Chr.19: 53381113-53381113 on Build GRCh38
rs931029029 Chr.19: 53381303-53381303 on Build GRCh38
rs987439827 Chr.19: 53381039-53381039 on Build GRCh38
rs774091401 Chr.19: 53381033-53381033 on Build GRCh38
rs764371545 Chr.19: 53381335-53381335 on Build GRCh38
rs113306675 Chr.19: 53381319-53381319 on Build GRCh38
rs758153460 Chr.19: 53381313-53381313 on Build GRCh38
rs765425326 Chr.19: 53381224-53381224 on Build GRCh38
rs766552036 Chr.19: 53381087-53381087 on Build GRCh38
rs758915553 Chr.19: 53381115-53381115 on Build GRCh38
rs111817845 Chr.19: 53381202-53381202 on Build GRCh38
rs757154677 Chr.19: 53381358-53381358 on Build GRCh38
rs1008513548 Chr.19: 53381269-53381269 on Build GRCh38
rs531281783 Chr.19: 53381372-53381372 on Build GRCh38
rs369898015 Chr.19: 53381339-53381339 on Build GRCh38
rs766427114 Chr.19: 53381419-53381419 on Build GRCh38
rs759033156 Chr.19: 53381403-53381403 on Build GRCh38
rs769776551 Chr.19: 53381364-53381364 on Build GRCh38
rs901841666 Chr.19: 53381175-53381175 on Build GRCh38
rs764612408 Chr.19: 53381256-53381256 on Build GRCh38
rs1051942833 Chr.19: 53381493-53381493 on Build GRCh38
rs758911176 Chr.19: 53381387-53381387 on Build GRCh38
rs151076278 Chr.19: 53381263-53381263 on Build GRCh38
rs4438413 Chr.19: 53381159-53381159 on Build GRCh38
rs745985076 Chr.19: 53381324-53381324 on Build GRCh38
rs762396967 Chr.19: 53381340-53381340 on Build GRCh38
rs761806969 Chr.19: 53381035-53381035 on Build GRCh38
rs778473198 Chr.19: 53381290-53381290 on Build GRCh38
rs749044040 Chr.19: 53381020-53381020 on Build GRCh38
rs763320505 Chr.19: 53381332-53381332 on Build GRCh38
rs778183122 Chr.19: 53381122-53381122 on Build GRCh38
rs752127054 Chr.19: 53381257-53381257 on Build GRCh38
rs765627658 Chr.19: 53381101-53381101 on Build GRCh38
rs562060597 Chr.19: 53381370-53381370 on Build GRCh38
rs762488069 Chr.19: 53381409-53381409 on Build GRCh38
rs773321553 Chr.19: 53381081-53381081 on Build GRCh38
rs928246020 Chr.19: 53381016-53381016 on Build GRCh38
rs967608880 Chr.19: 53381392-53381392 on Build GRCh38
rs754481513 Chr.19: 53381348-53381348 on Build GRCh38
rs770547420 Chr.19: 53381107-53381107 on Build GRCh38
rs751314213 Chr.19: 53381417-53381417 on Build GRCh38
rs4570987 Chr.19: 53381205-53381205 on Build GRCh38
rs10415690 Chr.19: 53381438-53381438 on Build GRCh38
rs529176483 Chr.19: 53381399-53381399 on Build GRCh38
rs774533262 Chr.19: 53381497-53381497 on Build GRCh38
rs764569207 Chr.19: 53381482-53381482 on Build GRCh38
rs778759729 Chr.19: 53381179-53381179 on Build GRCh38
rs778357704 Chr.19: 53381011-53381011 on Build GRCh38
rs779312372 Chr.19: 53381485-53381485 on Build GRCh38
rs188482460 Chr.19: 53381240-53381240 on Build GRCh38
rs977121285 Chr.19: 53381004-53381004 on Build GRCh38
rs545211475 Chr.19: 53381193-53381193 on Build GRCh38
rs567700001 Chr.19: 53381406-53381406 on Build GRCh38
rs779796113 Chr.19: 53381152-53381152 on Build GRCh38
rs1005513556 Chr.19: 53381126-53381126 on Build GRCh38
rs764645629 Chr.19: 53381405-53381405 on Build GRCh38
rs1049238983 Chr.19: 53381306-53381306 on Build GRCh38
rs768732883 Chr.19: 53381450-53381450 on Build GRCh38
rs755709150 Chr.19: 53381146-53381146 on Build GRCh38
rs768190457 Chr.19: 53381343-53381343 on Build GRCh38
rs776668890 Chr.19: 53381323-53381323 on Build GRCh38
rs749876398 Chr.19: 53381062-53381062 on Build GRCh38
rs777696065 Chr.19: 53381318-53381318 on Build GRCh38
rs773882504 Chr.19: 53381389-53381389 on Build GRCh38
rs779985548 Chr.19: 53381442-53381442 on Build GRCh38
rs531436305 Chr.19: 53381204-53381204 on Build GRCh38
rs770909666 Chr.19: 53381321-53381321 on Build GRCh38
rs553066327 Chr.19: 53381448-53381448 on Build GRCh38
rs184732580 Chr.19: 53381086-53381086 on Build GRCh38
rs749907571 Chr.19: 53381141-53381141 on Build GRCh38
rs771964552 Chr.19: 53381070-53381070 on Build GRCh38
rs752293161 Chr.19: 53381351-53381351 on Build GRCh38
rs748954573 Chr.19: 53381158-53381158 on Build GRCh38
rs776550310 Chr.19: 53381280-53381280 on Build GRCh38
rs754657851 Chr.19: 53381282-53381282 on Build GRCh38
rs758131655 Chr.19: 53381352-53381352 on Build GRCh38
rs781137094 Chr.19: 53381359-53381359 on Build GRCh38
rs770017359 Chr.19: 53381326-53381326 on Build GRCh38
rs796404061 Chr.19: 53381379-53381379 on Build GRCh38
rs977333151 Chr.19: 53381228-53381228 on Build GRCh38
rs768445270 Chr.19: 53381278-53381278 on Build GRCh38
rs762311066 Chr.19: 53381381-53381381 on Build GRCh38
rs768584625 Chr.19: 53381021-53381021 on Build GRCh38
rs777529257 Chr.19: 53381354-53381354 on Build GRCh38
rs888050685 Chr.19: 53381124-53381124 on Build GRCh38
rs775891067 Chr.19: 53381236-53381236 on Build GRCh38

View Full Product Details