Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23400071-23400619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC8P1
Entrez Gene ID
150244
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
Gene Aliases
ZDHHC8P
UniGene
Hs.386693
Gene Chromosome Location
Chr.22: 23390605-23402612 on Build GRCh38
Transcripts
NR_003950.1

SNP Detail

SNP ID Chromosome Location
rs13054327 Chr.22: 23400121-23400121 on Build GRCh38
rs865845235 Chr.22: 23400286-23400286 on Build GRCh38
rs915724687 Chr.22: 23400179-23400179 on Build GRCh38
rs780940633 Chr.22: 23400257-23400257 on Build GRCh38
rs375669030 Chr.22: 23400099-23400099 on Build GRCh38
rs743356 Chr.22: 23400522-23400522 on Build GRCh38
rs766280013 Chr.22: 23400376-23400376 on Build GRCh38
rs561847626 Chr.22: 23400297-23400297 on Build GRCh38
rs570601495 Chr.22: 23400327-23400327 on Build GRCh38
rs541237215 Chr.22: 23400575-23400575 on Build GRCh38
rs375270222 Chr.22: 23400293-23400293 on Build GRCh38
rs569725756 Chr.22: 23400412-23400412 on Build GRCh38
rs368610065 Chr.22: 23400334-23400334 on Build GRCh38
rs1015244943 Chr.22: 23400136-23400136 on Build GRCh38
rs577272659 Chr.22: 23400393-23400393 on Build GRCh38
rs541624088 Chr.22: 23400254-23400254 on Build GRCh38
rs374814932 Chr.22: 23400169-23400169 on Build GRCh38
rs527507739 Chr.22: 23400299-23400299 on Build GRCh38
rs966027346 Chr.22: 23400501-23400501 on Build GRCh38
rs1048508084 Chr.22: 23400381-23400381 on Build GRCh38
rs141296221 Chr.22: 23400252-23400252 on Build GRCh38
rs772986413 Chr.22: 23400185-23400185 on Build GRCh38
rs116402227 Chr.22: 23400204-23400204 on Build GRCh38
rs745550251 Chr.22: 23400270-23400270 on Build GRCh38
rs549752626 Chr.22: 23400343-23400343 on Build GRCh38
rs6003689 Chr.22: 23400392-23400392 on Build GRCh38
rs542120070 Chr.22: 23400488-23400488 on Build GRCh38
rs185613721 Chr.22: 23400197-23400197 on Build GRCh38
rs940180693 Chr.22: 23400591-23400591 on Build GRCh38
rs978648385 Chr.22: 23400191-23400191 on Build GRCh38
rs562595695 Chr.22: 23400228-23400228 on Build GRCh38
rs970204825 Chr.22: 23400447-23400447 on Build GRCh38
rs1027636193 Chr.22: 23400109-23400109 on Build GRCh38
rs1014320635 Chr.22: 23400409-23400409 on Build GRCh38
rs145498578 Chr.22: 23400345-23400345 on Build GRCh38
rs186133348 Chr.22: 23400385-23400385 on Build GRCh38
rs371419720 Chr.22: 23400146-23400146 on Build GRCh38
rs925810181 Chr.22: 23400347-23400347 on Build GRCh38
rs148030050 Chr.22: 23400352-23400352 on Build GRCh38
rs201942212 Chr.22: 23400159-23400160 on Build GRCh38
rs932783471 Chr.22: 23400378-23400378 on Build GRCh38
rs369243572 Chr.22: 23400511-23400511 on Build GRCh38
rs181614743 Chr.22: 23400310-23400310 on Build GRCh38
rs532751447 Chr.22: 23400342-23400342 on Build GRCh38
rs944022135 Chr.22: 23400360-23400360 on Build GRCh38
rs900016131 Chr.22: 23400106-23400106 on Build GRCh38
rs545851870 Chr.22: 23400446-23400446 on Build GRCh38
rs879944535 Chr.22: 23400287-23400287 on Build GRCh38
rs921280937 Chr.22: 23400526-23400526 on Build GRCh38
rs867416098 Chr.22: 23400566-23400566 on Build GRCh38
rs753166306 Chr.22: 23400203-23400203 on Build GRCh38
rs375697656 Chr.22: 23400117-23400117 on Build GRCh38
rs534165371 Chr.22: 23400391-23400391 on Build GRCh38
rs1048595865 Chr.22: 23400264-23400264 on Build GRCh38
rs758670539 Chr.22: 23400249-23400249 on Build GRCh38
rs984186923 Chr.22: 23400572-23400572 on Build GRCh38
rs743353 Chr.22: 23400351-23400351 on Build GRCh38
rs538007588 Chr.22: 23400189-23400189 on Build GRCh38
rs748799010 Chr.22: 23400384-23400384 on Build GRCh38
rs888688002 Chr.22: 23400267-23400267 on Build GRCh38
rs1041074389 Chr.22: 23400375-23400375 on Build GRCh38
rs140899128 Chr.22: 23400476-23400476 on Build GRCh38
rs743355 Chr.22: 23400489-23400489 on Build GRCh38
rs743354 Chr.22: 23400477-23400477 on Build GRCh38
rs368104961 Chr.22: 23400215-23400215 on Build GRCh38
rs959745712 Chr.22: 23400143-23400143 on Build GRCh38
rs969859447 Chr.22: 23400344-23400344 on Build GRCh38
rs539465768 Chr.22: 23400116-23400116 on Build GRCh38
rs959719241 Chr.22: 23400289-23400289 on Build GRCh38
rs370432470 Chr.22: 23400202-23400202 on Build GRCh38
rs992988373 Chr.22: 23400165-23400165 on Build GRCh38
rs190420532 Chr.22: 23400224-23400224 on Build GRCh38

View Full Product Details