Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185980338-185980913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC344887
Entrez Gene ID
344887
Gene Name
NmrA-like family domain containing 1 pseudogene
Gene Aliases
-
UniGene
Hs.128803
Gene Chromosome Location
Chr.3: 185959969-185980877 on Build GRCh38
Transcripts
NR_033752.2

SNP Detail

SNP ID Chromosome Location
rs13085763 Chr.3: 185980858-185980858 on Build GRCh38
rs534936256 Chr.3: 185980790-185980790 on Build GRCh38
rs570391480 Chr.3: 185980768-185980768 on Build GRCh38
rs1036516871 Chr.3: 185980591-185980591 on Build GRCh38
rs920950541 Chr.3: 185980507-185980507 on Build GRCh38
rs895109888 Chr.3: 185980569-185980569 on Build GRCh38
rs924081663 Chr.3: 185980845-185980845 on Build GRCh38
rs1012844039 Chr.3: 185980380-185980380 on Build GRCh38
rs920971686 Chr.3: 185980731-185980731 on Build GRCh38
rs771993467 Chr.3: 185980530-185980530 on Build GRCh38
rs906656368 Chr.3: 185980656-185980656 on Build GRCh38
rs553218159 Chr.3: 185980382-185980382 on Build GRCh38
rs151219672 Chr.3: 185980409-185980409 on Build GRCh38
rs189526516 Chr.3: 185980521-185980521 on Build GRCh38
rs954453397 Chr.3: 185980829-185980829 on Build GRCh38
rs965146369 Chr.3: 185980460-185980460 on Build GRCh38
rs897975837 Chr.3: 185980597-185980597 on Build GRCh38
rs558247205 Chr.3: 185980763-185980763 on Build GRCh38
rs574691366 Chr.3: 185980859-185980859 on Build GRCh38
rs972260562 Chr.3: 185980821-185980821 on Build GRCh38
rs185823915 Chr.3: 185980466-185980466 on Build GRCh38
rs60681086 Chr.3: 185980713-185980713 on Build GRCh38
rs894479458 Chr.3: 185980636-185980636 on Build GRCh38
rs780476580 Chr.3: 185980705-185980705 on Build GRCh38
rs1021922834 Chr.3: 185980399-185980399 on Build GRCh38
rs542036059 Chr.3: 185980884-185980884 on Build GRCh38
rs561321873 Chr.3: 185980493-185980493 on Build GRCh38
rs777126330 Chr.3: 185980807-185980807 on Build GRCh38
rs965178107 Chr.3: 185980719-185980719 on Build GRCh38
rs755970135 Chr.3: 185980424-185980424 on Build GRCh38
rs978290525 Chr.3: 185980512-185980512 on Build GRCh38
rs953816823 Chr.3: 185980752-185980752 on Build GRCh38
rs968663573 Chr.3: 185980425-185980425 on Build GRCh38
rs1047515498 Chr.3: 185980702-185980703 on Build GRCh38
rs185321299 Chr.3: 185980728-185980728 on Build GRCh38
rs868113612 Chr.3: 185980843-185980843 on Build GRCh38
rs998755948 Chr.3: 185980694-185980694 on Build GRCh38
rs150336102 Chr.3: 185980603-185980603 on Build GRCh38
rs565598465 Chr.3: 185980559-185980559 on Build GRCh38
rs1002927324 Chr.3: 185980602-185980602 on Build GRCh38
rs779690950 Chr.3: 185980429-185980429 on Build GRCh38
rs543448179 Chr.3: 185980537-185980537 on Build GRCh38
rs958205925 Chr.3: 185980855-185980855 on Build GRCh38
rs75941919 Chr.3: 185980667-185980667 on Build GRCh38
rs1057298278 Chr.3: 185980867-185980867 on Build GRCh38
rs933483539 Chr.3: 185980511-185980511 on Build GRCh38
rs1050702218 Chr.3: 185980534-185980534 on Build GRCh38
rs984226471 Chr.3: 185980767-185980767 on Build GRCh38
rs749281310 Chr.3: 185980431-185980431 on Build GRCh38
rs530186807 Chr.3: 185980662-185980662 on Build GRCh38
rs942278068 Chr.3: 185980538-185980538 on Build GRCh38
rs547651588 Chr.3: 185980644-185980644 on Build GRCh38
rs924428045 Chr.3: 185980441-185980441 on Build GRCh38
rs543143638 Chr.3: 185980474-185980474 on Build GRCh38
rs974885225 Chr.3: 185980498-185980498 on Build GRCh38
rs1013374229 Chr.3: 185980645-185980645 on Build GRCh38
rs181833148 Chr.3: 185980649-185980649 on Build GRCh38

View Full Product Details