Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80299545-80300093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC644936
Entrez Gene ID
644936
Gene Name
actin, beta pseudogene
Gene Aliases
-
UniGene
Hs.627173
Gene Chromosome Location
Chr.5: 80299085-80300478 on Build GRCh38
Transcripts
NR_004845.1

SNP Detail

SNP ID Chromosome Location
rs745700069 Chr.5: 80299849-80299849 on Build GRCh38
rs767045810 Chr.5: 80300017-80300017 on Build GRCh38
rs773810326 Chr.5: 80299898-80299898 on Build GRCh38
rs777935921 Chr.5: 80299719-80299720 on Build GRCh38
rs780200669 Chr.5: 80299719-80299719 on Build GRCh38
rs549331107 Chr.5: 80299692-80299692 on Build GRCh38
rs764747506 Chr.5: 80299776-80299776 on Build GRCh38
rs982935353 Chr.5: 80299588-80299588 on Build GRCh38
rs768823404 Chr.5: 80299741-80299741 on Build GRCh38
rs938194569 Chr.5: 80299571-80299571 on Build GRCh38
rs1043068407 Chr.5: 80299764-80299765 on Build GRCh38
rs757236963 Chr.5: 80299858-80299860 on Build GRCh38
rs779169249 Chr.5: 80299951-80299951 on Build GRCh38
rs761183729 Chr.5: 80300025-80300025 on Build GRCh38
rs749257414 Chr.5: 80299745-80299745 on Build GRCh38
rs760679168 Chr.5: 80299763-80299763 on Build GRCh38
rs776516484 Chr.5: 80299845-80299845 on Build GRCh38
rs765608787 Chr.5: 80300035-80300035 on Build GRCh38
rs754023341 Chr.5: 80299933-80299933 on Build GRCh38
rs758618511 Chr.5: 80300044-80300044 on Build GRCh38
rs180758298 Chr.5: 80299844-80299844 on Build GRCh38
rs757674284 Chr.5: 80299693-80299693 on Build GRCh38
rs750703688 Chr.5: 80299818-80299818 on Build GRCh38
rs773646116 Chr.5: 80300023-80300023 on Build GRCh38
rs766660788 Chr.5: 80300031-80300031 on Build GRCh38
rs566010384 Chr.5: 80299786-80299786 on Build GRCh38
rs571357517 Chr.5: 80299889-80299889 on Build GRCh38
rs574066216 Chr.5: 80300008-80300008 on Build GRCh38
rs770866105 Chr.5: 80299765-80299765 on Build GRCh38
rs752373516 Chr.5: 80299779-80299779 on Build GRCh38
rs752936603 Chr.5: 80300036-80300036 on Build GRCh38
rs78387030 Chr.5: 80300000-80300000 on Build GRCh38
rs781032114 Chr.5: 80299969-80299969 on Build GRCh38
rs759056133 Chr.5: 80299648-80299648 on Build GRCh38
rs775073718 Chr.5: 80299864-80299864 on Build GRCh38
rs868058753 Chr.5: 80299851-80299851 on Build GRCh38
rs778887361 Chr.5: 80299861-80299863 on Build GRCh38
rs33987570 Chr.5: 80299586-80299586 on Build GRCh38
rs756498958 Chr.5: 80299910-80299910 on Build GRCh38
rs758531296 Chr.5: 80299954-80299954 on Build GRCh38
rs746294908 Chr.5: 80299742-80299742 on Build GRCh38
rs947373282 Chr.5: 80299759-80299759 on Build GRCh38
rs767899569 Chr.5: 80299804-80299804 on Build GRCh38
rs796508434 Chr.5: 80299593-80299593 on Build GRCh38
rs778303157 Chr.5: 80299691-80299691 on Build GRCh38
rs531541707 Chr.5: 80299698-80299698 on Build GRCh38
rs752899946 Chr.5: 80299699-80299699 on Build GRCh38
rs769601249 Chr.5: 80299852-80299852 on Build GRCh38
rs1026606500 Chr.5: 80299937-80299937 on Build GRCh38
rs749371931 Chr.5: 80299722-80299722 on Build GRCh38
rs750907445 Chr.5: 80299906-80299906 on Build GRCh38
rs773346881 Chr.5: 80299755-80299755 on Build GRCh38
rs928792620 Chr.5: 80299611-80299611 on Build GRCh38
rs762532747 Chr.5: 80299892-80299892 on Build GRCh38
rs759268809 Chr.5: 80299773-80299773 on Build GRCh38
rs557386802 Chr.5: 80300007-80300007 on Build GRCh38
rs780176357 Chr.5: 80299824-80299824 on Build GRCh38
rs781626527 Chr.5: 80300051-80300051 on Build GRCh38
rs764813045 Chr.5: 80299653-80299653 on Build GRCh38
rs745815266 Chr.5: 80299980-80299980 on Build GRCh38
rs757692625 Chr.5: 80299803-80299803 on Build GRCh38
rs777897254 Chr.5: 80300055-80300055 on Build GRCh38
rs560428540 Chr.5: 80299977-80299977 on Build GRCh38
rs75500114 Chr.5: 80300032-80300032 on Build GRCh38
rs766751209 Chr.5: 80299927-80299927 on Build GRCh38
rs752113164 Chr.5: 80299674-80299674 on Build GRCh38
rs188711576 Chr.5: 80299645-80299645 on Build GRCh38
rs755188033 Chr.5: 80299936-80299936 on Build GRCh38
rs752071689 Chr.5: 80299799-80299799 on Build GRCh38
rs762412357 Chr.5: 80299783-80299783 on Build GRCh38
rs765829704 Chr.5: 80299857-80299857 on Build GRCh38
rs763244575 Chr.5: 80299789-80299789 on Build GRCh38
rs535021813 Chr.5: 80299833-80299833 on Build GRCh38
rs748194051 Chr.5: 80299952-80299952 on Build GRCh38
rs267600711 Chr.5: 80299964-80299964 on Build GRCh38
rs768057144 Chr.5: 80299903-80299903 on Build GRCh38
rs756165563 Chr.5: 80299643-80299646 on Build GRCh38
rs4704659 Chr.5: 80299577-80299577 on Build GRCh38
rs769536532 Chr.5: 80299998-80299998 on Build GRCh38
rs749518453 Chr.5: 80299832-80299832 on Build GRCh38
rs758967902 Chr.5: 80299687-80299687 on Build GRCh38
rs1056223598 Chr.5: 80299575-80299575 on Build GRCh38
rs1039584668 Chr.5: 80299930-80299930 on Build GRCh38
rs113631138 Chr.5: 80299709-80299709 on Build GRCh38
rs768247299 Chr.5: 80300002-80300002 on Build GRCh38
rs1035838289 Chr.5: 80299986-80299986 on Build GRCh38
rs553232783 Chr.5: 80300073-80300073 on Build GRCh38
rs753606481 Chr.5: 80299647-80299647 on Build GRCh38
rs779081128 Chr.5: 80299836-80299836 on Build GRCh38
rs915898512 Chr.5: 80299720-80299720 on Build GRCh38
rs756475544 Chr.5: 80299822-80299822 on Build GRCh38
rs776225570 Chr.5: 80299768-80299768 on Build GRCh38

View Full Product Details