SNP ID | Chromosome Location |
---|---|
rs782646782 | Chr.7: 72954921-72954921 on Build GRCh38 |
rs544708269 | Chr.7: 72954897-72954897 on Build GRCh38 |
rs529334060 | Chr.7: 72954685-72954685 on Build GRCh38 |
rs562045450 | Chr.7: 72954627-72954627 on Build GRCh38 |
rs781889767 | Chr.7: 72954856-72954856 on Build GRCh38 |
rs372531970 | Chr.7: 72954841-72954841 on Build GRCh38 |
rs562980100 | Chr.7: 72954913-72954913 on Build GRCh38 |
rs782471579 | Chr.7: 72954881-72954881 on Build GRCh38 |
rs782696351 | Chr.7: 72954895-72954895 on Build GRCh38 |