Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 271440-271788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS11
Entrez Gene ID
8786
Gene Name
regulator of G-protein signaling 11
Gene Aliases
RS11
UniGene
Hs.65756
Gene Chromosome Location
Chr.16: 268301-275943 on Build GRCh38
Transcripts
NM_001286485.1 NM_001286486.1 NM_003834.2 NM_183337.2 XM_011522719.2 XM_011522720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765749 c.674T>G p.F225C Chr.16: 271553-271553 on Build GRCh38
5754850 c.667G>C p.A223P Chr.16: 271560-271560 on Build GRCh38
3969556 c.708G>A p.A236A Chr.16: 271440-271440 on Build GRCh38
5971786 c.681G>T p.K227N Chr.16: 271546-271546 on Build GRCh38
4556753 c.701G>A p.R234K Chr.16: 271447-271447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746136466 Chr.16: 271487-271487 on Build GRCh38
rs116186505 Chr.16: 271602-271602 on Build GRCh38
rs781255419 Chr.16: 271481-271481 on Build GRCh38
rs754911017 Chr.16: 271468-271468 on Build GRCh38
rs760360581 Chr.16: 271514-271514 on Build GRCh38
rs78828691 Chr.16: 271475-271475 on Build GRCh38
rs776492339 Chr.16: 271618-271618 on Build GRCh38
rs762717698 Chr.16: 271697-271697 on Build GRCh38
rs376124889 Chr.16: 271490-271490 on Build GRCh38
rs751312663 Chr.16: 271544-271544 on Build GRCh38
rs778919378 Chr.16: 271471-271471 on Build GRCh38
rs774993456 Chr.16: 271504-271504 on Build GRCh38
rs772970431 Chr.16: 271513-271513 on Build GRCh38
rs553529200 Chr.16: 271532-271532 on Build GRCh38
rs942170273 Chr.16: 271583-271583 on Build GRCh38
rs533495196 Chr.16: 271721-271721 on Build GRCh38
rs761131273 Chr.16: 271607-271607 on Build GRCh38
rs756272311 Chr.16: 271580-271580 on Build GRCh38
rs767976524 Chr.16: 271511-271511 on Build GRCh38
rs372183786 Chr.16: 271588-271588 on Build GRCh38
rs146310051 Chr.16: 271545-271545 on Build GRCh38
rs903384432 Chr.16: 271743-271743 on Build GRCh38
rs183388202 Chr.16: 271755-271755 on Build GRCh38
rs762763286 Chr.16: 271562-271562 on Build GRCh38
rs779268887 Chr.16: 271597-271597 on Build GRCh38
rs770805823 Chr.16: 271578-271579 on Build GRCh38
rs193239476 Chr.16: 271744-271744 on Build GRCh38
rs34176388 Chr.16: 271499-271500 on Build GRCh38
rs538004424 Chr.16: 271463-271463 on Build GRCh38
rs200711536 Chr.16: 271538-271538 on Build GRCh38
rs370501221 Chr.16: 271491-271491 on Build GRCh38
rs774364756 Chr.16: 271622-271622 on Build GRCh38
rs768978272 Chr.16: 271499-271499 on Build GRCh38
rs188444285 Chr.16: 271683-271683 on Build GRCh38
rs951869583 Chr.16: 271670-271670 on Build GRCh38
rs1000767682 Chr.16: 271747-271747 on Build GRCh38
rs866504438 Chr.16: 271469-271469 on Build GRCh38
rs753426799 Chr.16: 271467-271467 on Build GRCh38
rs762075062 Chr.16: 271507-271507 on Build GRCh38
rs183564596 Chr.16: 271527-271527 on Build GRCh38
rs769672916 Chr.16: 271472-271483 on Build GRCh38
rs984760874 Chr.16: 271677-271677 on Build GRCh38
rs148110594 Chr.16: 271606-271606 on Build GRCh38
rs143680653 Chr.16: 271466-271466 on Build GRCh38
rs1034054116 Chr.16: 271674-271674 on Build GRCh38
rs868491584 Chr.16: 271483-271483 on Build GRCh38
rs989922935 Chr.16: 271703-271703 on Build GRCh38
rs1054587799 Chr.16: 271749-271749 on Build GRCh38
rs117340695 Chr.16: 271627-271627 on Build GRCh38
rs770036167 Chr.16: 271489-271489 on Build GRCh38
rs1056210184 Chr.16: 271706-271706 on Build GRCh38
rs753336870 Chr.16: 271518-271518 on Build GRCh38
rs942118587 Chr.16: 271759-271759 on Build GRCh38
rs777585890 Chr.16: 271562-271562 on Build GRCh38
rs976422680 Chr.16: 271719-271719 on Build GRCh38
rs950133583 Chr.16: 271716-271716 on Build GRCh38
rs768842752 Chr.16: 271589-271589 on Build GRCh38
rs926530637 Chr.16: 271695-271695 on Build GRCh38
rs748732044 Chr.16: 271601-271601 on Build GRCh38
rs563082171 Chr.16: 271707-271707 on Build GRCh38
rs772865019 Chr.16: 271473-271482 on Build GRCh38
rs766148936 Chr.16: 271517-271517 on Build GRCh38
rs780260073 Chr.16: 271587-271587 on Build GRCh38
rs746829716 Chr.16: 271579-271579 on Build GRCh38
rs763919987 Chr.16: 271739-271740 on Build GRCh38
rs770637006 Chr.16: 271610-271610 on Build GRCh38
rs758278783 Chr.16: 271480-271480 on Build GRCh38

View Full Product Details