Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35163893-35164206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC45B
Entrez Gene ID
146862
Gene Name
unc-45 myosin chaperone B
Gene Aliases
CMYA4, CTRCT43, SMUNC45, UNC-45B, UNC45
UniGene
Hs.379636
Gene Chromosome Location
Chr.17: 35147793-35189345 on Build GRCh38
Transcripts
NM_001033576.1 NM_001267052.1 NM_001308281.1 NM_173167.3 XM_017024234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6523442 c.1072C>A p.L358I Chr.17: 35164087-35164087 on Build GRCh38
6080583 c.1008G>T p.G336G Chr.17: 35164023-35164023 on Build GRCh38
6080582 c.1087T>C p.Y363H Chr.17: 35164102-35164102 on Build GRCh38
1269778 c.1110G>A p.P370P Chr.17: 35164125-35164125 on Build GRCh38
1269779 c.1116C>T p.R372R Chr.17: 35164131-35164131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371932269 Chr.17: 35164167-35164167 on Build GRCh38
rs8079654 Chr.17: 35163972-35163972 on Build GRCh38
rs73288986 Chr.17: 35164050-35164050 on Build GRCh38
rs746531584 Chr.17: 35164122-35164122 on Build GRCh38
rs148057044 Chr.17: 35164132-35164132 on Build GRCh38
rs910831938 Chr.17: 35163927-35163927 on Build GRCh38
rs747941098 Chr.17: 35164115-35164115 on Build GRCh38
rs147497902 Chr.17: 35164162-35164162 on Build GRCh38
rs369464349 Chr.17: 35163974-35163974 on Build GRCh38
rs767643886 Chr.17: 35164141-35164141 on Build GRCh38
rs894287699 Chr.17: 35164160-35164160 on Build GRCh38
rs200248455 Chr.17: 35163955-35163955 on Build GRCh38
rs760206661 Chr.17: 35164056-35164056 on Build GRCh38
rs971006993 Chr.17: 35164075-35164075 on Build GRCh38
rs769010743 Chr.17: 35164170-35164170 on Build GRCh38
rs971845470 Chr.17: 35164083-35164083 on Build GRCh38
rs41389545 Chr.17: 35164145-35164145 on Build GRCh38
rs758516385 Chr.17: 35163984-35163984 on Build GRCh38
rs370514797 Chr.17: 35164036-35164036 on Build GRCh38
rs745572525 Chr.17: 35164129-35164129 on Build GRCh38
rs186108233 Chr.17: 35163921-35163921 on Build GRCh38
rs749115934 Chr.17: 35164026-35164026 on Build GRCh38
rs548212884 Chr.17: 35164166-35164166 on Build GRCh38
rs1048289026 Chr.17: 35164052-35164052 on Build GRCh38
rs1036860958 Chr.17: 35163976-35163976 on Build GRCh38
rs780890443 Chr.17: 35164163-35164163 on Build GRCh38
rs778765079 Chr.17: 35164159-35164159 on Build GRCh38
rs143612410 Chr.17: 35164130-35164130 on Build GRCh38
rs751614171 Chr.17: 35164092-35164092 on Build GRCh38
rs894425072 Chr.17: 35164059-35164059 on Build GRCh38
rs150418771 Chr.17: 35164101-35164101 on Build GRCh38
rs759929321 Chr.17: 35163971-35163971 on Build GRCh38
rs749621908 Chr.17: 35164168-35164168 on Build GRCh38
rs764641013 Chr.17: 35164089-35164089 on Build GRCh38
rs775920008 Chr.17: 35164085-35164085 on Build GRCh38
rs781343218 Chr.17: 35164023-35164023 on Build GRCh38
rs190150367 Chr.17: 35164184-35164184 on Build GRCh38
rs374608869 Chr.17: 35164131-35164131 on Build GRCh38
rs553976077 Chr.17: 35164014-35164014 on Build GRCh38
rs759982142 Chr.17: 35164082-35164082 on Build GRCh38
rs766093913 Chr.17: 35163958-35163958 on Build GRCh38
rs867174215 Chr.17: 35163942-35163942 on Build GRCh38
rs143327878 Chr.17: 35164155-35164155 on Build GRCh38
rs145196717 Chr.17: 35164103-35164103 on Build GRCh38
rs200620770 Chr.17: 35164124-35164124 on Build GRCh38
rs754299931 Chr.17: 35163968-35163968 on Build GRCh38
rs777783420 Chr.17: 35163985-35163985 on Build GRCh38
rs116362062 Chr.17: 35164114-35164114 on Build GRCh38
rs201918277 Chr.17: 35164137-35164137 on Build GRCh38
rs777712664 Chr.17: 35164120-35164120 on Build GRCh38
rs768722725 Chr.17: 35163977-35163977 on Build GRCh38
rs748091247 Chr.17: 35164045-35164045 on Build GRCh38
rs575944372 Chr.17: 35164084-35164084 on Build GRCh38
rs571014198 Chr.17: 35164051-35164051 on Build GRCh38
rs750588632 Chr.17: 35164111-35164111 on Build GRCh38
rs779255426 Chr.17: 35164142-35164142 on Build GRCh38
rs746904732 Chr.17: 35164158-35164158 on Build GRCh38
rs200767716 Chr.17: 35164035-35164035 on Build GRCh38
rs140605574 Chr.17: 35164069-35164069 on Build GRCh38
rs776382433 Chr.17: 35164070-35164070 on Build GRCh38
rs751629771 Chr.17: 35163999-35163999 on Build GRCh38
rs761898642 Chr.17: 35164140-35164140 on Build GRCh38
rs200190191 Chr.17: 35164125-35164125 on Build GRCh38
rs201319738 Chr.17: 35164078-35164078 on Build GRCh38
rs763412905 Chr.17: 35164087-35164087 on Build GRCh38
rs771429052 Chr.17: 35163947-35163947 on Build GRCh38
rs1012798414 Chr.17: 35164174-35164174 on Build GRCh38
rs758172033 Chr.17: 35164146-35164146 on Build GRCh38

View Full Product Details