Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5900190-5900776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCZ1
Entrez Gene ID
51622
Gene Name
CCZ1 homolog, vacuolar protein trafficking and biogenesis associated
Gene Aliases
C7orf28A, CCZ1A, CGI-43, H_DJ1163J12.2
UniGene
Hs.530000
Gene Chromosome Location
Chr.7: 5898710-5925974 on Build GRCh38
Transcripts
NM_015622.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366988 c.163A>G p.K55E Chr.7: 5900326-5900326 on Build GRCh38
5467177 c.172A>C p.K58Q Chr.7: 5900335-5900335 on Build GRCh38
150231 c.312+10T>C p.? Chr.7: 5900576-5900576 on Build GRCh38
1091056 c.174G>T p.K58N Chr.7: 5900337-5900337 on Build GRCh38
6337765 c.222A>C p.T74T Chr.7: 5900476-5900476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766773475 Chr.7: 5900389-5900390 on Build GRCh38
rs746151502 Chr.7: 5900525-5900525 on Build GRCh38
rs755573529 Chr.7: 5900455-5900455 on Build GRCh38
rs760252882 Chr.7: 5900562-5900562 on Build GRCh38
rs754346957 Chr.7: 5900427-5900427 on Build GRCh38
rs780015346 Chr.7: 5900368-5900368 on Build GRCh38
rs565160061 Chr.7: 5900544-5900544 on Build GRCh38
rs761342665 Chr.7: 5900408-5900408 on Build GRCh38
rs577078309 Chr.7: 5900584-5900584 on Build GRCh38
rs761291189 Chr.7: 5900540-5900540 on Build GRCh38
rs759953456 Chr.7: 5900391-5900391 on Build GRCh38
rs753184587 Chr.7: 5900274-5900274 on Build GRCh38
rs573071922 Chr.7: 5900506-5900506 on Build GRCh38
rs933620119 Chr.7: 5900454-5900454 on Build GRCh38
rs767124622 Chr.7: 5900429-5900429 on Build GRCh38
rs763439471 Chr.7: 5900397-5900397 on Build GRCh38
rs556305434 Chr.7: 5900396-5900396 on Build GRCh38
rs773684573 Chr.7: 5900406-5900406 on Build GRCh38
rs770332495 Chr.7: 5900395-5900395 on Build GRCh38
rs1022840346 Chr.7: 5900745-5900745 on Build GRCh38
rs775652575 Chr.7: 5900535-5900535 on Build GRCh38
rs757404030 Chr.7: 5900581-5900581 on Build GRCh38
rs747049315 Chr.7: 5900494-5900494 on Build GRCh38
rs754834055 Chr.7: 5900573-5900573 on Build GRCh38
rs544164883 Chr.7: 5900607-5900607 on Build GRCh38
rs780844649 Chr.7: 5900346-5900346 on Build GRCh38
rs374188325 Chr.7: 5900435-5900435 on Build GRCh38
rs749582193 Chr.7: 5900616-5900616 on Build GRCh38
rs958355467 Chr.7: 5900399-5900399 on Build GRCh38
rs755943262 Chr.7: 5900365-5900365 on Build GRCh38
rs1037471631 Chr.7: 5900220-5900220 on Build GRCh38
rs372092894 Chr.7: 5900536-5900536 on Build GRCh38
rs570846172 Chr.7: 5900384-5900384 on Build GRCh38
rs756539311 Chr.7: 5900246-5900250 on Build GRCh38
rs765803129 Chr.7: 5900273-5900273 on Build GRCh38
rs771232057 Chr.7: 5900389-5900389 on Build GRCh38
rs552435211 Chr.7: 5900313-5900313 on Build GRCh38
rs993349920 Chr.7: 5900249-5900250 on Build GRCh38
rs898869536 Chr.7: 5900232-5900232 on Build GRCh38
rs779069169 Chr.7: 5900454-5900455 on Build GRCh38
rs969373315 Chr.7: 5900336-5900336 on Build GRCh38
rs778935516 Chr.7: 5900489-5900489 on Build GRCh38
rs748253320 Chr.7: 5900469-5900469 on Build GRCh38
rs534172641 Chr.7: 5900221-5900221 on Build GRCh38
rs763572440 Chr.7: 5900295-5900295 on Build GRCh38
rs772185789 Chr.7: 5900376-5900376 on Build GRCh38
rs745953074 Chr.7: 5900605-5900605 on Build GRCh38
rs758536353 Chr.7: 5900514-5900516 on Build GRCh38
rs1026556901 Chr.7: 5900256-5900256 on Build GRCh38
rs988366485 Chr.7: 5900420-5900426 on Build GRCh38
rs771285108 Chr.7: 5900254-5900254 on Build GRCh38
rs753529767 Chr.7: 5900572-5900572 on Build GRCh38
rs777028289 Chr.7: 5900390-5900390 on Build GRCh38
rs538206685 Chr.7: 5900386-5900386 on Build GRCh38
rs1014012670 Chr.7: 5900739-5900739 on Build GRCh38
rs556314773 Chr.7: 5900441-5900441 on Build GRCh38
rs199548480 Chr.7: 5900505-5900505 on Build GRCh38
rs2528311 Chr.7: 5900401-5900401 on Build GRCh38
rs754816624 Chr.7: 5900478-5900478 on Build GRCh38
rs577244373 Chr.7: 5900210-5900210 on Build GRCh38
rs1047580686 Chr.7: 5900676-5900676 on Build GRCh38
rs562346627 Chr.7: 5900645-5900645 on Build GRCh38
rs2528312 Chr.7: 5900576-5900576 on Build GRCh38
rs766151411 Chr.7: 5900452-5900452 on Build GRCh38
rs895624682 Chr.7: 5900699-5900699 on Build GRCh38
rs760004762 Chr.7: 5900263-5900263 on Build GRCh38
rs1024959664 Chr.7: 5900296-5900300 on Build GRCh38
rs752730347 Chr.7: 5900580-5900580 on Build GRCh38
rs993650676 Chr.7: 5900654-5900654 on Build GRCh38
rs762503587 Chr.7: 5900291-5900291 on Build GRCh38
rs747102323 Chr.7: 5900387-5900387 on Build GRCh38
rs778126444 Chr.7: 5900599-5900602 on Build GRCh38
rs777081265 Chr.7: 5900258-5900258 on Build GRCh38
rs753669327 Chr.7: 5900462-5900462 on Build GRCh38
rs751222623 Chr.7: 5900312-5900312 on Build GRCh38
rs969915393 Chr.7: 5900751-5900751 on Build GRCh38
rs1007204842 Chr.7: 5900215-5900215 on Build GRCh38
rs370275453 Chr.7: 5900371-5900371 on Build GRCh38
rs767067493 Chr.7: 5900542-5900542 on Build GRCh38
rs770191244 Chr.7: 5900530-5900530 on Build GRCh38

View Full Product Details