Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 16376983-16377133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGST1
Entrez Gene ID
4257
Gene Name
microsomal glutathione S-transferase 1
Gene Aliases
GST12, MGST, MGST-I
UniGene
Hs.389700
Gene Chromosome Location
Chr.12: 16347142-16377189 on Build GRCh38
Transcripts
NM_001267598.1

SNP Detail

SNP ID Chromosome Location
rs536411183 Chr.12: 16377078-16377078 on Build GRCh38
rs925750076 Chr.12: 16377071-16377071 on Build GRCh38
rs913148057 Chr.12: 16377013-16377013 on Build GRCh38
rs895597577 Chr.12: 16377094-16377094 on Build GRCh38
rs1055451096 Chr.12: 16377089-16377089 on Build GRCh38
rs945915453 Chr.12: 16377046-16377046 on Build GRCh38
rs978710280 Chr.12: 16377059-16377059 on Build GRCh38
rs188368509 Chr.12: 16377074-16377074 on Build GRCh38

View Full Product Details