Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133205811-133206166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OBP2B
Entrez Gene ID
29989
Gene Name
odorant binding protein 2B
Gene Aliases
LCN14, OBPIIb
UniGene
Hs.449629
Gene Chromosome Location
Chr.9: 133205279-133223255 on Build GRCh38
Transcripts
NM_001288987.1 NM_014581.3 NR_110242.1 NR_110243.1 XM_006717086.3 XM_017014654.1 XM_017014655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
403982 c.503C>T p.P168L Chr.9: 133205928-133205928 on Build GRCh38
1701877 c.491G>A p.G164E Chr.9: 133205940-133205940 on Build GRCh38
3655245 c.505G>A p.E169K Chr.9: 133205926-133205926 on Build GRCh38
5912935 c.502C>T p.P168S Chr.9: 133205929-133205929 on Build GRCh38
607842 c.504C>A p.P168P Chr.9: 133205927-133205927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149232831 Chr.9: 133205975-133205975 on Build GRCh38
rs146582704 Chr.9: 133205902-133205902 on Build GRCh38
rs112844018 Chr.9: 133206102-133206102 on Build GRCh38
rs137892402 Chr.9: 133205875-133205875 on Build GRCh38
rs6597608 Chr.9: 133206004-133206004 on Build GRCh38
rs6597609 Chr.9: 133206079-133206079 on Build GRCh38
rs147134232 Chr.9: 133205896-133205896 on Build GRCh38
rs148158748 Chr.9: 133205969-133205969 on Build GRCh38

View Full Product Details