Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50749774-50749940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLD
Entrez Gene ID
1540
Gene Name
CYLD lysine 63 deubiquitinase
Gene Aliases
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
UniGene
Hs.578973
Gene Chromosome Location
Chr.16: 50742026-50801935 on Build GRCh38
Transcripts
NM_001042355.1 NM_001042412.1 NM_015247.2 XM_005255812.2 XM_006721149.1 XM_011522907.2 XM_011522908.1 XM_011522911.2 XM_017022977.1 XM_017022978.1 XM_017022979.1 XM_017022980.1 XM_017022981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2834409 c.117T>C p.L39L Chr.16: 50749815-50749815 on Build GRCh38
703521 c.212G>T p.G71V Chr.16: 50749910-50749910 on Build GRCh38
2834411 c.161C>T p.S54F Chr.16: 50749859-50749859 on Build GRCh38
2834408 c.101A>C p.Q34P Chr.16: 50749799-50749799 on Build GRCh38
7246622 c.126G>C p.P42P Chr.16: 50749824-50749824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756702476 Chr.16: 50749813-50749813 on Build GRCh38
rs755565463 Chr.16: 50749800-50749800 on Build GRCh38
rs769485126 Chr.16: 50749847-50749847 on Build GRCh38
rs202119806 Chr.16: 50749824-50749824 on Build GRCh38
rs538206791 Chr.16: 50749798-50749798 on Build GRCh38
rs761931123 Chr.16: 50749866-50749866 on Build GRCh38
rs754294680 Chr.16: 50749907-50749907 on Build GRCh38
rs770539079 Chr.16: 50749823-50749823 on Build GRCh38
rs754313539 Chr.16: 50749797-50749797 on Build GRCh38
rs779556463 Chr.16: 50749812-50749812 on Build GRCh38
rs760892570 Chr.16: 50749892-50749892 on Build GRCh38
rs749025501 Chr.16: 50749858-50749858 on Build GRCh38
rs1048602687 Chr.16: 50749842-50749842 on Build GRCh38
rs774243056 Chr.16: 50749862-50749862 on Build GRCh38
rs369619557 Chr.16: 50749906-50749906 on Build GRCh38
rs767537235 Chr.16: 50749868-50749868 on Build GRCh38
rs745498288 Chr.16: 50749836-50749836 on Build GRCh38
rs373198684 Chr.16: 50749911-50749911 on Build GRCh38
rs375455772 Chr.16: 50749856-50749856 on Build GRCh38
rs768708025 Chr.16: 50749860-50749860 on Build GRCh38
rs773427979 Chr.16: 50749891-50749891 on Build GRCh38
rs931057264 Chr.16: 50749832-50749832 on Build GRCh38

View Full Product Details