Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147614343-147614507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9
Entrez Gene ID
607
Gene Name
B-cell CLL/lymphoma 9
Gene Aliases
LGS
UniGene
Hs.415209
Gene Chromosome Location
Chr.1: 147541394-147626219 on Build GRCh38
Transcripts
NM_004326.3 XM_005272971.4 XM_005277417.3 XM_005277418.3 XM_017002026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1560018 c.371-1G>T p.? Chr.1: 147614426-147614426 on Build GRCh38
6121023 c.397T>A p.S133T Chr.1: 147614453-147614453 on Build GRCh38
5048406 c.385G>T p.D129Y Chr.1: 147614441-147614441 on Build GRCh38
4829918 c.437C>G p.S146* Chr.1: 147614493-147614493 on Build GRCh38
6610778 c.437C>A p.S146* Chr.1: 147614493-147614493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782768600 Chr.1: 147614477-147614477 on Build GRCh38
rs782756769 Chr.1: 147614468-147614468 on Build GRCh38
rs931449079 Chr.1: 147614470-147614470 on Build GRCh38
rs1028346494 Chr.1: 147614398-147614398 on Build GRCh38
rs781934608 Chr.1: 147614459-147614459 on Build GRCh38
rs587620626 Chr.1: 147614375-147614375 on Build GRCh38
rs782326842 Chr.1: 147614402-147614402 on Build GRCh38
rs974587299 Chr.1: 147614367-147614367 on Build GRCh38
rs781911464 Chr.1: 147614488-147614488 on Build GRCh38
rs138412167 Chr.1: 147614472-147614472 on Build GRCh38
rs376554346 Chr.1: 147614432-147614432 on Build GRCh38
rs782526867 Chr.1: 147614474-147614474 on Build GRCh38
rs202244151 Chr.1: 147614406-147614406 on Build GRCh38
rs894867400 Chr.1: 147614482-147614482 on Build GRCh38
rs782103104 Chr.1: 147614414-147614414 on Build GRCh38
rs973406031 Chr.1: 147614462-147614462 on Build GRCh38
rs782041372 Chr.1: 147614467-147614467 on Build GRCh38
rs920226086 Chr.1: 147614466-147614466 on Build GRCh38
rs781980201 Chr.1: 147614427-147614427 on Build GRCh38
rs782311693 Chr.1: 147614407-147614407 on Build GRCh38
rs782788303 Chr.1: 147614455-147614455 on Build GRCh38

View Full Product Details