Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5044354-5044516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSL6
Entrez Gene ID
11172
Gene Name
insulin like 6
Gene Aliases
RIF1
UniGene
Hs.632648
Gene Chromosome Location
Chr.9: 4992408-5185706 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_001322204.1 NM_004972.3

Gene Symbol
JAK2
Entrez Gene ID
3717
Gene Name
Janus kinase 2
Gene Aliases
JTK10, THCYT3
UniGene
Hs.656213
Gene Chromosome Location
Chr.9: 4985086-5128183 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_001322204.1 NM_004972.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6907590 c.403G>T p.G135* Chr.9: 5044455-5044455 on Build GRCh38
6928835 c.436G>C p.D146H Chr.9: 5044488-5044488 on Build GRCh38
455998 c.396T>G p.Y132* Chr.9: 5044448-5044448 on Build GRCh38
3375256 c.366T>C p.R122R Chr.9: 5044418-5044418 on Build GRCh38
6571143 c.433C>T p.L145F Chr.9: 5044485-5044485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777208458 Chr.9: 5044465-5044465 on Build GRCh38
rs370926996 Chr.9: 5044399-5044399 on Build GRCh38
rs56118985 Chr.9: 5044432-5044432 on Build GRCh38
rs769648312 Chr.9: 5044443-5044443 on Build GRCh38
rs765699090 Chr.9: 5044479-5044479 on Build GRCh38
rs761305442 Chr.9: 5044395-5044395 on Build GRCh38
rs760861063 Chr.9: 5044453-5044453 on Build GRCh38
rs114448906 Chr.9: 5044397-5044397 on Build GRCh38
rs200008036 Chr.9: 5044390-5044390 on Build GRCh38
rs147483622 Chr.9: 5044416-5044416 on Build GRCh38
rs750963233 Chr.9: 5044393-5044393 on Build GRCh38
rs757898652 Chr.9: 5044417-5044417 on Build GRCh38
rs913352383 Chr.9: 5044382-5044382 on Build GRCh38
rs140052813 Chr.9: 5044458-5044458 on Build GRCh38
rs897100091 Chr.9: 5044471-5044471 on Build GRCh38
rs746923735 Chr.9: 5044428-5044428 on Build GRCh38
rs977211315 Chr.9: 5044485-5044485 on Build GRCh38
rs774426615 Chr.9: 5044447-5044447 on Build GRCh38
rs778371211 Chr.9: 5044407-5044407 on Build GRCh38
rs368963113 Chr.9: 5044427-5044427 on Build GRCh38
rs774641791 Chr.9: 5044386-5044386 on Build GRCh38
rs376070326 Chr.9: 5044450-5044450 on Build GRCh38
rs761744348 Chr.9: 5044388-5044388 on Build GRCh38
rs777303047 Chr.9: 5044418-5044418 on Build GRCh38
rs745587711 Chr.9: 5044437-5044437 on Build GRCh38
rs763071761 Chr.9: 5044386-5044386 on Build GRCh38
rs1046359522 Chr.9: 5044391-5044391 on Build GRCh38
rs371826393 Chr.9: 5044449-5044449 on Build GRCh38

View Full Product Details