Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43530065-43530213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
UniGene
Hs.434059
Gene Chromosome Location
Chr.17: 43527843-43546432 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3787345 c.886+7C>T p.? Chr.17: 43530100-43530100 on Build GRCh38
187406 c.876C>T p.D292D Chr.17: 43530117-43530117 on Build GRCh38
5741004 c.848A>C p.E283A Chr.17: 43530145-43530145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979920984 Chr.17: 43530173-43530173 on Build GRCh38
rs141492045 Chr.17: 43530162-43530162 on Build GRCh38
rs776031033 Chr.17: 43530093-43530093 on Build GRCh38
rs866359165 Chr.17: 43530167-43530167 on Build GRCh38
rs754808570 Chr.17: 43530185-43530185 on Build GRCh38
rs529970715 Chr.17: 43530168-43530168 on Build GRCh38
rs759361652 Chr.17: 43530148-43530148 on Build GRCh38
rs750690248 Chr.17: 43530145-43530145 on Build GRCh38
rs971572480 Chr.17: 43530169-43530169 on Build GRCh38
rs770324500 Chr.17: 43530092-43530092 on Build GRCh38
rs760428912 Chr.17: 43530155-43530155 on Build GRCh38
rs374876992 Chr.17: 43530166-43530166 on Build GRCh38
rs769220923 Chr.17: 43530109-43530109 on Build GRCh38
rs1046551303 Chr.17: 43530088-43530088 on Build GRCh38
rs763506155 Chr.17: 43530108-43530108 on Build GRCh38
rs776505043 Chr.17: 43530083-43530083 on Build GRCh38
rs137969667 Chr.17: 43530116-43530116 on Build GRCh38

View Full Product Details