Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 127738393-127738523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYC
Entrez Gene ID
4609
Gene Name
v-myc avian myelocytomatosis viral oncogene homolog
Gene Aliases
MRTL, MYCC, bHLHe39, c-Myc
UniGene
Hs.202453
Gene Chromosome Location
Chr.8: 127736069-127741434 on Build GRCh38
Transcripts
NM_002467.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4651620 c.163C>T p.L55L Chr.8: 127738425-127738425 on Build GRCh38
6180017 c.153G>T p.K51N Chr.8: 127738415-127738415 on Build GRCh38
1286403 c.218C>T p.T73I Chr.8: 127738435-127738435 on Build GRCh38
4171760 c.184delT p.S62fs*41 Chr.8: 127738446-127738446 on Build GRCh38
5652069 c.268G>A p.V90I Chr.8: 127738485-127738485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121918685 Chr.8: 127738437-127738437 on Build GRCh38
rs745545434 Chr.8: 127738472-127738472 on Build GRCh38
rs1016220380 Chr.8: 127738490-127738490 on Build GRCh38
rs750664148 Chr.8: 127738434-127738434 on Build GRCh38
rs148228388 Chr.8: 127738489-127738489 on Build GRCh38
rs375402667 Chr.8: 127738458-127738458 on Build GRCh38
rs886556187 Chr.8: 127738474-127738474 on Build GRCh38
rs866188449 Chr.8: 127738485-127738485 on Build GRCh38
rs749753251 Chr.8: 127738445-127738445 on Build GRCh38
rs762965100 Chr.8: 127738496-127738496 on Build GRCh38
rs756091827 Chr.8: 127738435-127738435 on Build GRCh38
rs28933407 Chr.8: 127738431-127738431 on Build GRCh38
rs931443247 Chr.8: 127738452-127738452 on Build GRCh38
rs769788275 Chr.8: 127738424-127738426 on Build GRCh38
rs780202654 Chr.8: 127738444-127738444 on Build GRCh38
rs767700277 Chr.8: 127738428-127738428 on Build GRCh38
rs372115981 Chr.8: 127738421-127738421 on Build GRCh38
rs776077629 Chr.8: 127738471-127738471 on Build GRCh38
rs770609785 Chr.8: 127738463-127738463 on Build GRCh38
rs769148296 Chr.8: 127738484-127738484 on Build GRCh38

View Full Product Details