Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42290124-42290261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKB
Entrez Gene ID
3551
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Gene Aliases
IKK-beta, IKK2, IKKB, IMD15, NFKBIKB
UniGene
Hs.597664
Gene Chromosome Location
Chr.8: 42271302-42332653 on Build GRCh38
Transcripts
NM_001190720.2 NM_001242778.1 NM_001556.2 NR_033818.1 NR_033819.1 NR_040009.1 XM_005273490.2 XM_005273491.4 XM_005273492.3 XM_005273493.3 XM_005273494.2 XM_005273495.2 XM_005273496.3 XM_005273498.3 XM_011544517.2 XM_011544518.2 XM_011544519.2 XM_011544520.2 XM_011544521.2 XM_011544522.2 XM_017013396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312110 c.201G>T p.R67S Chr.8: 42290156-42290156 on Build GRCh38
3649318 c.284C>T p.A95V Chr.8: 42290239-42290239 on Build GRCh38
374629 c.245G>T p.G82V Chr.8: 42290200-42290200 on Build GRCh38
2961821 c.268G>A p.D90N Chr.8: 42290223-42290223 on Build GRCh38
144951 c.241G>C p.E81Q Chr.8: 42290196-42290196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201340172 Chr.8: 42290151-42290151 on Build GRCh38
rs763432993 Chr.8: 42290221-42290221 on Build GRCh38
rs771764280 Chr.8: 42290170-42290170 on Build GRCh38
rs200136227 Chr.8: 42290185-42290185 on Build GRCh38
rs201837296 Chr.8: 42290216-42290216 on Build GRCh38
rs778765246 Chr.8: 42290147-42290147 on Build GRCh38
rs745680522 Chr.8: 42290149-42290149 on Build GRCh38
rs200048037 Chr.8: 42290202-42290202 on Build GRCh38
rs775274100 Chr.8: 42290175-42290175 on Build GRCh38
rs773847619 Chr.8: 42290217-42290217 on Build GRCh38
rs201017671 Chr.8: 42290201-42290201 on Build GRCh38
rs746623468 Chr.8: 42290189-42290189 on Build GRCh38
rs768256200 Chr.8: 42290192-42290192 on Build GRCh38
rs373547989 Chr.8: 42290231-42290231 on Build GRCh38
rs766630242 Chr.8: 42290225-42290225 on Build GRCh38
rs912645794 Chr.8: 42290184-42290184 on Build GRCh38
rs931318595 Chr.8: 42290237-42290237 on Build GRCh38
rs1046213152 Chr.8: 42290198-42290198 on Build GRCh38

View Full Product Details