Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39583866-39584020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THBS1
Entrez Gene ID
7057
Gene Name
thrombospondin 1
Gene Aliases
THBS, THBS-1, TSP, TSP-1, TSP1
UniGene
Hs.164226
Gene Chromosome Location
Chr.15: 39581079-39598918 on Build GRCh38
Transcripts
NM_003246.3 XM_011521971.1

SNP Detail

SNP ID Chromosome Location
rs764233037 Chr.15: 39583992-39583992 on Build GRCh38
rs371984857 Chr.15: 39583952-39583952 on Build GRCh38
rs754102615 Chr.15: 39583994-39583994 on Build GRCh38
rs759439308 Chr.15: 39583941-39583941 on Build GRCh38
rs757234170 Chr.15: 39583995-39583995 on Build GRCh38
rs1009019543 Chr.15: 39583912-39583912 on Build GRCh38
rs765389942 Chr.15: 39583998-39583998 on Build GRCh38
rs954819187 Chr.15: 39583903-39583903 on Build GRCh38
rs760532925 Chr.15: 39583971-39583971 on Build GRCh38
rs368239596 Chr.15: 39583949-39583949 on Build GRCh38
rs992933398 Chr.15: 39583943-39583943 on Build GRCh38
rs938806229 Chr.15: 39583918-39583918 on Build GRCh38
rs549522881 Chr.15: 39583927-39583927 on Build GRCh38

View Full Product Details