Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 124370097-124370269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D1A
Entrez Gene ID
4068
Gene Name
SH2 domain containing 1A
Gene Aliases
DSHP, EBVS, IMD5, LYP, MTCP1, SAP, SAP/SH2D1A, XLP, XLPD, XLPD1
UniGene
Hs.349094
Gene Chromosome Location
Chr.X: 124346282-124373160 on Build GRCh38
Transcripts
NM_001114937.2 NM_002351.4

SNP Detail

SNP ID Chromosome Location
rs954824608 Chr.X: 124370193-124370193 on Build GRCh38
rs371553659 Chr.X: 124370199-124370199 on Build GRCh38
rs1056354956 Chr.X: 124370181-124370181 on Build GRCh38
rs760763168 Chr.X: 124370138-124370138 on Build GRCh38
rs753697568 Chr.X: 124370240-124370240 on Build GRCh38
rs767110636 Chr.X: 124370168-124370168 on Build GRCh38
rs111033627 Chr.X: 124370177-124370177 on Build GRCh38

View Full Product Details