Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170731.4 | NP_733927.1 | 1 - 2 | 647 | 104 |
GenBank mRNA | AB038670.1 | - | 1 - 2 | 317 | 104 |
AK289853.1 | - | 1 - 2 | 34 | 104 | |
AY054392.2 | - | 1 - 2 | 338 | 104 | |
AY442188.1 | - | 1 - 2 | 148 | 104 | |
EF689021.1 | - | 1 - 2 | 647 | 104 |