Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82025440-82025677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC45
Entrez Gene ID
201255
Gene Name
leucine rich repeat containing 45
Gene Aliases
-
UniGene
Hs.143774
Gene Chromosome Location
Chr.17: 82023252-82031151 on Build GRCh38
Transcripts
NM_144999.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1522773 c.658G>A p.V220M Chr.17: 82025504-82025504 on Build GRCh38
5498291 c.661+2T>C p.? Chr.17: 82025509-82025509 on Build GRCh38
2805930 c.637C>T p.P213S Chr.17: 82025483-82025483 on Build GRCh38
7219263 c.657C>G p.A219A Chr.17: 82025503-82025503 on Build GRCh38
5853815 c.637_638CC>TT p.P213F Chr.17: 82025483-82025484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780563447 Chr.17: 82025514-82025514 on Build GRCh38
rs922061143 Chr.17: 82025564-82025564 on Build GRCh38
rs983102313 Chr.17: 82025513-82025513 on Build GRCh38
rs775157579 Chr.17: 82025537-82025537 on Build GRCh38
rs769492480 Chr.17: 82025536-82025536 on Build GRCh38
rs543499106 Chr.17: 82025516-82025516 on Build GRCh38
rs528861638 Chr.17: 82025578-82025578 on Build GRCh38
rs747188204 Chr.17: 82025586-82025586 on Build GRCh38
rs557866291 Chr.17: 82025504-82025504 on Build GRCh38
rs779244331 Chr.17: 82025526-82025526 on Build GRCh38
rs935308233 Chr.17: 82025620-82025620 on Build GRCh38
rs751405012 Chr.17: 82025542-82025542 on Build GRCh38
rs769082284 Chr.17: 82025520-82025520 on Build GRCh38
rs141253038 Chr.17: 82025473-82025473 on Build GRCh38
rs767675042 Chr.17: 82025546-82025546 on Build GRCh38
rs772769787 Chr.17: 82025464-82025464 on Build GRCh38
rs72861735 Chr.17: 82025503-82025503 on Build GRCh38
rs981494489 Chr.17: 82025597-82025597 on Build GRCh38
rs561678970 Chr.17: 82025571-82025571 on Build GRCh38
rs746067182 Chr.17: 82025511-82025511 on Build GRCh38
rs909399964 Chr.17: 82025642-82025642 on Build GRCh38
rs751519095 Chr.17: 82025492-82025492 on Build GRCh38
rs202169133 Chr.17: 82025483-82025483 on Build GRCh38
rs559775815 Chr.17: 82025641-82025641 on Build GRCh38
rs768825198 Chr.17: 82025589-82025589 on Build GRCh38
rs946919260 Chr.17: 82025572-82025572 on Build GRCh38
rs541770183 Chr.17: 82025594-82025594 on Build GRCh38
rs748707634 Chr.17: 82025530-82025530 on Build GRCh38
rs763981616 Chr.17: 82025541-82025541 on Build GRCh38
rs117200853 Chr.17: 82025491-82025491 on Build GRCh38
rs888193199 Chr.17: 82025480-82025480 on Build GRCh38
rs756383330 Chr.17: 82025516-82025529 on Build GRCh38
rs374567972 Chr.17: 82025535-82025535 on Build GRCh38
rs1027583383 Chr.17: 82025509-82025509 on Build GRCh38
rs761860198 Chr.17: 82025543-82025543 on Build GRCh38
rs370914482 Chr.17: 82025534-82025534 on Build GRCh38
rs143513889 Chr.17: 82025489-82025489 on Build GRCh38
rs774386568 Chr.17: 82025590-82025590 on Build GRCh38
rs929599290 Chr.17: 82025623-82025623 on Build GRCh38
rs200533399 Chr.17: 82025512-82025512 on Build GRCh38
rs372034528 Chr.17: 82025488-82025488 on Build GRCh38
rs760050172 Chr.17: 82025468-82025468 on Build GRCh38
rs759232920 Chr.17: 82025481-82025481 on Build GRCh38

View Full Product Details