Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33658489-33658744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR3
Entrez Gene ID
3710
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
Gene Aliases
IP3R, IP3R3
UniGene
Hs.65758
Gene Chromosome Location
Chr.6: 33621333-33696574 on Build GRCh38
Transcripts
NM_002224.3 XM_011514577.2 XM_017010832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3873228 c.440C>T p.A147V Chr.6: 33658740-33658740 on Build GRCh38
94545 c.413G>A p.R138Q Chr.6: 33658713-33658713 on Build GRCh38
4693588 c.440C>A p.A147D Chr.6: 33658740-33658740 on Build GRCh38
1255110 c.398T>C p.L133P Chr.6: 33658698-33658698 on Build GRCh38
6688866 c.420G>A p.P140P Chr.6: 33658720-33658720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149427831 Chr.6: 33658636-33658636 on Build GRCh38
rs376148863 Chr.6: 33658653-33658653 on Build GRCh38
rs779388729 Chr.6: 33658664-33658664 on Build GRCh38
rs755291621 Chr.6: 33658657-33658657 on Build GRCh38
rs376573493 Chr.6: 33658637-33658637 on Build GRCh38
rs41267657 Chr.6: 33658623-33658623 on Build GRCh38
rs776487015 Chr.6: 33658702-33658703 on Build GRCh38
rs1036894172 Chr.6: 33658575-33658575 on Build GRCh38
rs368040077 Chr.6: 33658624-33658624 on Build GRCh38
rs373241230 Chr.6: 33658639-33658639 on Build GRCh38
rs746532129 Chr.6: 33658695-33658695 on Build GRCh38
rs995399396 Chr.6: 33658608-33658608 on Build GRCh38
rs771023240 Chr.6: 33658621-33658621 on Build GRCh38
rs769456797 Chr.6: 33658713-33658713 on Build GRCh38
rs112967729 Chr.6: 33658601-33658601 on Build GRCh38
rs149436819 Chr.6: 33658720-33658720 on Build GRCh38
rs34233805 Chr.6: 33658596-33658597 on Build GRCh38
rs1046168566 Chr.6: 33658548-33658548 on Build GRCh38
rs372579558 Chr.6: 33658650-33658650 on Build GRCh38
rs770925389 Chr.6: 33658702-33658702 on Build GRCh38
rs759577490 Chr.6: 33658712-33658712 on Build GRCh38
rs553827680 Chr.6: 33658560-33658560 on Build GRCh38
rs369008901 Chr.6: 33658635-33658635 on Build GRCh38
rs749396203 Chr.6: 33658638-33658638 on Build GRCh38
rs1047650561 Chr.6: 33658549-33658549 on Build GRCh38
rs770877795 Chr.6: 33658631-33658638 on Build GRCh38
rs908718454 Chr.6: 33658572-33658572 on Build GRCh38
rs140001792 Chr.6: 33658693-33658693 on Build GRCh38
rs929092786 Chr.6: 33658529-33658529 on Build GRCh38
rs756568696 Chr.6: 33658649-33658649 on Build GRCh38
rs146063738 Chr.6: 33658679-33658679 on Build GRCh38
rs779858992 Chr.6: 33658568-33658568 on Build GRCh38
rs1038587542 Chr.6: 33658607-33658607 on Build GRCh38
rs897100484 Chr.6: 33658587-33658587 on Build GRCh38
rs748323588 Chr.6: 33658602-33658602 on Build GRCh38
rs765148910 Chr.6: 33658628-33658628 on Build GRCh38
rs758119600 Chr.6: 33658543-33658543 on Build GRCh38
rs754160122 Chr.6: 33658651-33658651 on Build GRCh38
rs771890282 Chr.6: 33658692-33658692 on Build GRCh38

View Full Product Details