Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7317886-7318131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEURL4
Entrez Gene ID
84461
Gene Name
neuralized E3 ubiquitin protein ligase 4
Gene Aliases
-
UniGene
Hs.654794
Gene Chromosome Location
Chr.17: 7315632-7329319 on Build GRCh38
Transcripts
NM_001005408.1 NM_032442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707377 c.4006G>T p.E1336* Chr.17: 7318113-7318113 on Build GRCh38
6724832 c.4072A>G p.M1358V Chr.17: 7317921-7317921 on Build GRCh38
1196081 c.4022G>A p.C1341Y Chr.17: 7318097-7318097 on Build GRCh38
983946 c.4006G>A p.E1336K Chr.17: 7318113-7318113 on Build GRCh38
2977106 c.4094T>G p.L1365R Chr.17: 7317899-7317899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775511826 Chr.17: 7318034-7318034 on Build GRCh38
rs901360959 Chr.17: 7317951-7317952 on Build GRCh38
rs199774639 Chr.17: 7317914-7317914 on Build GRCh38
rs34321237 Chr.17: 7318042-7318042 on Build GRCh38
rs769222803 Chr.17: 7317945-7317945 on Build GRCh38
rs1127440 Chr.17: 7318066-7318066 on Build GRCh38
rs566593186 Chr.17: 7317983-7317983 on Build GRCh38
rs933124738 Chr.17: 7317951-7317951 on Build GRCh38
rs139635041 Chr.17: 7317974-7317974 on Build GRCh38
rs1048250809 Chr.17: 7318086-7318086 on Build GRCh38
rs116939857 Chr.17: 7317913-7317913 on Build GRCh38
rs772739856 Chr.17: 7317966-7317966 on Build GRCh38
rs774090894 Chr.17: 7318019-7318019 on Build GRCh38
rs879747595 Chr.17: 7317993-7317993 on Build GRCh38
rs201025474 Chr.17: 7317921-7317921 on Build GRCh38
rs776288799 Chr.17: 7318053-7318054 on Build GRCh38
rs761710913 Chr.17: 7318076-7318078 on Build GRCh38
rs755435664 Chr.17: 7317936-7317936 on Build GRCh38
rs369997022 Chr.17: 7318049-7318049 on Build GRCh38
rs930325093 Chr.17: 7317906-7317906 on Build GRCh38
rs750610230 Chr.17: 7317915-7317915 on Build GRCh38
rs780420992 Chr.17: 7317931-7317931 on Build GRCh38
rs1037659090 Chr.17: 7318058-7318058 on Build GRCh38
rs767405463 Chr.17: 7318030-7318030 on Build GRCh38
rs758557088 Chr.17: 7318053-7318053 on Build GRCh38
rs767977252 Chr.17: 7317985-7317985 on Build GRCh38
rs748663327 Chr.17: 7317943-7317943 on Build GRCh38
rs1003658404 Chr.17: 7318096-7318096 on Build GRCh38
rs765372222 Chr.17: 7318048-7318048 on Build GRCh38
rs766503620 Chr.17: 7318056-7318056 on Build GRCh38
rs756636059 Chr.17: 7318092-7318092 on Build GRCh38
rs199799903 Chr.17: 7317937-7317937 on Build GRCh38
rs781620119 Chr.17: 7318085-7318085 on Build GRCh38
rs748614049 Chr.17: 7318087-7318087 on Build GRCh38
rs993515268 Chr.17: 7318083-7318083 on Build GRCh38
rs538715477 Chr.17: 7318044-7318044 on Build GRCh38
rs755236528 Chr.17: 7318078-7318078 on Build GRCh38

View Full Product Details