Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 133151627-133151854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH10
Entrez Gene ID
57575
Gene Name
protocadherin 10
Gene Aliases
OL-PCDH, PCDH19
UniGene
Hs.192859
Gene Chromosome Location
Chr.4: 133149290-133208606 on Build GRCh38
Transcripts
NM_020815.2 NM_032961.2 XM_011532150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428165 c.1554C>G p.I518M Chr.4: 133151694-133151694 on Build GRCh38
5048121 c.1624C>T p.Q542* Chr.4: 133151764-133151764 on Build GRCh38
4711479 c.1528A>T p.M510L Chr.4: 133151668-133151668 on Build GRCh38
273078 c.1641C>T p.D547D Chr.4: 133151781-133151781 on Build GRCh38
732590 c.1571A>G p.Y524C Chr.4: 133151711-133151711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772246043 Chr.4: 133151823-133151823 on Build GRCh38
rs147051817 Chr.4: 133151666-133151666 on Build GRCh38
rs1051884766 Chr.4: 133151726-133151726 on Build GRCh38
rs1009600867 Chr.4: 133151784-133151784 on Build GRCh38
rs746069561 Chr.4: 133151685-133151685 on Build GRCh38
rs770920439 Chr.4: 133151674-133151674 on Build GRCh38
rs893781694 Chr.4: 133151781-133151781 on Build GRCh38
rs753159365 Chr.4: 133151654-133151654 on Build GRCh38
rs765802021 Chr.4: 133151733-133151733 on Build GRCh38
rs773122619 Chr.4: 133151832-133151832 on Build GRCh38
rs778277377 Chr.4: 133151785-133151785 on Build GRCh38
rs200246899 Chr.4: 133151650-133151650 on Build GRCh38
rs138396564 Chr.4: 133151694-133151694 on Build GRCh38
rs970893347 Chr.4: 133151809-133151809 on Build GRCh38
rs757845067 Chr.4: 133151798-133151798 on Build GRCh38
rs769577344 Chr.4: 133151831-133151831 on Build GRCh38
rs756563130 Chr.4: 133151782-133151782 on Build GRCh38
rs756657017 Chr.4: 133151660-133151660 on Build GRCh38
rs767317232 Chr.4: 133151761-133151761 on Build GRCh38
rs200266347 Chr.4: 133151688-133151688 on Build GRCh38
rs552224221 Chr.4: 133151696-133151696 on Build GRCh38
rs745990864 Chr.4: 133151815-133151815 on Build GRCh38
rs568804747 Chr.4: 133151752-133151752 on Build GRCh38
rs769667444 Chr.4: 133151703-133151703 on Build GRCh38
rs773032336 Chr.4: 133151711-133151711 on Build GRCh38
rs780274523 Chr.4: 133151827-133151827 on Build GRCh38
rs778449540 Chr.4: 133151661-133151661 on Build GRCh38
rs754297421 Chr.4: 133151793-133151793 on Build GRCh38
rs1032040669 Chr.4: 133151686-133151686 on Build GRCh38
rs762454585 Chr.4: 133151718-133151718 on Build GRCh38
rs754743505 Chr.4: 133151648-133151648 on Build GRCh38
rs779119714 Chr.4: 133151675-133151675 on Build GRCh38
rs142476647 Chr.4: 133151738-133151738 on Build GRCh38
rs781679488 Chr.4: 133151649-133151649 on Build GRCh38
rs778841098 Chr.4: 133151799-133151799 on Build GRCh38
rs531255597 Chr.4: 133151772-133151772 on Build GRCh38

View Full Product Details