Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70275026-70275251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYOX1
Entrez Gene ID
51449
Gene Name
prenylcysteine oxidase 1
Gene Aliases
PCL1
UniGene
Hs.567502
Gene Chromosome Location
Chr.2: 70258099-70281185 on Build GRCh38
Transcripts
NM_016297.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6742169 c.611A>G p.Q204R Chr.2: 70275075-70275075 on Build GRCh38
276682 c.614A>C p.K205T Chr.2: 70275078-70275078 on Build GRCh38
1641966 c.600T>C p.L200L Chr.2: 70275064-70275064 on Build GRCh38
1180912 c.686delC p.T229fs*22 Chr.2: 70275150-70275150 on Build GRCh38
4757054 c.619G>A p.G207S Chr.2: 70275083-70275083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759458331 Chr.2: 70275170-70275170 on Build GRCh38
rs768822629 Chr.2: 70275139-70275139 on Build GRCh38
rs759337796 Chr.2: 70275105-70275105 on Build GRCh38
rs763108431 Chr.2: 70275074-70275074 on Build GRCh38
rs751259078 Chr.2: 70275125-70275125 on Build GRCh38
rs777113568 Chr.2: 70275117-70275117 on Build GRCh38
rs1050443770 Chr.2: 70275059-70275059 on Build GRCh38
rs60495451 Chr.2: 70275090-70275090 on Build GRCh38
rs762775879 Chr.2: 70275181-70275181 on Build GRCh38
rs765234614 Chr.2: 70275108-70275108 on Build GRCh38
rs772426700 Chr.2: 70275157-70275157 on Build GRCh38
rs112438565 Chr.2: 70275192-70275192 on Build GRCh38
rs994053596 Chr.2: 70275126-70275126 on Build GRCh38
rs138112690 Chr.2: 70275103-70275103 on Build GRCh38
rs767797442 Chr.2: 70275092-70275092 on Build GRCh38
rs761942957 Chr.2: 70275151-70275151 on Build GRCh38
rs756691186 Chr.2: 70275185-70275185 on Build GRCh38
rs200274702 Chr.2: 70275159-70275159 on Build GRCh38
rs775797895 Chr.2: 70275058-70275058 on Build GRCh38
rs948077676 Chr.2: 70275053-70275053 on Build GRCh38
rs780253234 Chr.2: 70275209-70275209 on Build GRCh38
rs532860601 Chr.2: 70275128-70275128 on Build GRCh38
rs751937451 Chr.2: 70275191-70275192 on Build GRCh38
rs199918295 Chr.2: 70275150-70275150 on Build GRCh38
rs368822515 Chr.2: 70275112-70275112 on Build GRCh38
rs780668774 Chr.2: 70275131-70275131 on Build GRCh38
rs757990304 Chr.2: 70275114-70275114 on Build GRCh38
rs113179619 Chr.2: 70275229-70275229 on Build GRCh38
rs372696710 Chr.2: 70275129-70275129 on Build GRCh38
rs1027128308 Chr.2: 70275158-70275158 on Build GRCh38
rs764933256 Chr.2: 70275171-70275171 on Build GRCh38
rs767171249 Chr.2: 70275190-70275190 on Build GRCh38
rs201962424 Chr.2: 70275135-70275135 on Build GRCh38
rs761512877 Chr.2: 70275227-70275227 on Build GRCh38
rs769995781 Chr.2: 70275130-70275130 on Build GRCh38
rs141857998 Chr.2: 70275076-70275076 on Build GRCh38
rs186275811 Chr.2: 70275082-70275082 on Build GRCh38
rs756571921 Chr.2: 70275206-70275206 on Build GRCh38
rs997857716 Chr.2: 70275218-70275218 on Build GRCh38
rs775535323 Chr.2: 70275180-70275180 on Build GRCh38
rs762098210 Chr.2: 70275087-70275087 on Build GRCh38
rs954873930 Chr.2: 70275228-70275228 on Build GRCh38
rs764045294 Chr.2: 70275182-70275182 on Build GRCh38
rs200248412 Chr.2: 70275054-70275054 on Build GRCh38

View Full Product Details