Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106901445-106901706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN2
Entrez Gene ID
9075
Gene Name
claudin 2
Gene Aliases
-
UniGene
Hs.522746
Gene Chromosome Location
Chr.X: 106900164-106930861 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171706.1 NM_138382.2

Gene Symbol
RIPPLY1
Entrez Gene ID
92129
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
UniGene
Hs.334726
Gene Chromosome Location
Chr.X: 106900063-106903331 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171706.1 NM_138382.2

SNP Detail

SNP ID Chromosome Location
rs755166835 Chr.X: 106901533-106901533 on Build GRCh38
rs905639182 Chr.X: 106901652-106901652 on Build GRCh38
rs762242561 Chr.X: 106901507-106901507 on Build GRCh38
rs893939105 Chr.X: 106901591-106901591 on Build GRCh38
rs147935658 Chr.X: 106901675-106901675 on Build GRCh38
rs774712878 Chr.X: 106901506-106901506 on Build GRCh38
rs749492848 Chr.X: 106901623-106901623 on Build GRCh38
rs1001587558 Chr.X: 106901658-106901658 on Build GRCh38
rs749040159 Chr.X: 106901612-106901612 on Build GRCh38
rs777555072 Chr.X: 106901535-106901535 on Build GRCh38
rs375779868 Chr.X: 106901581-106901581 on Build GRCh38
rs889207288 Chr.X: 106901528-106901528 on Build GRCh38
rs760725358 Chr.X: 106901525-106901525 on Build GRCh38
rs766412926 Chr.X: 106901527-106901527 on Build GRCh38
rs751427848 Chr.X: 106901550-106901550 on Build GRCh38
rs190216177 Chr.X: 106901521-106901521 on Build GRCh38
rs193137729 Chr.X: 106901568-106901568 on Build GRCh38
rs148800101 Chr.X: 106901501-106901501 on Build GRCh38
rs780021319 Chr.X: 106901573-106901573 on Build GRCh38
rs769049678 Chr.X: 106901505-106901505 on Build GRCh38
rs144879961 Chr.X: 106901597-106901597 on Build GRCh38
rs960400341 Chr.X: 106901662-106901662 on Build GRCh38
rs200487665 Chr.X: 106901485-106901485 on Build GRCh38
rs779303961 Chr.X: 106901649-106901649 on Build GRCh38
rs750454244 Chr.X: 106901522-106901522 on Build GRCh38
rs1052694188 Chr.X: 106901589-106901589 on Build GRCh38
rs1011661890 Chr.X: 106901538-106901538 on Build GRCh38
rs757516784 Chr.X: 106901624-106901624 on Build GRCh38
rs1036375477 Chr.X: 106901647-106901647 on Build GRCh38
rs574917949 Chr.X: 106901632-106901632 on Build GRCh38

View Full Product Details