Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116616818-116617051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH4A
Entrez Gene ID
345895
Gene Name
radial spoke head 4 homolog A
Gene Aliases
CILD11, RSHL3, RSPH6B, dJ412I7.1
UniGene
Hs.160380
Gene Chromosome Location
Chr.6: 116598184-116633051 on Build GRCh38
Transcripts
NM_001010892.2 NM_001161664.1 XM_017010826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243354 c.314C>T p.A105V Chr.6: 116616937-116616937 on Build GRCh38
6318119 c.226G>T p.G76* Chr.6: 116616849-116616849 on Build GRCh38
4825473 c.278C>G p.S93C Chr.6: 116616901-116616901 on Build GRCh38
3157321 c.286C>G p.P96A Chr.6: 116616909-116616909 on Build GRCh38
3619692 c.373C>T p.P125S Chr.6: 116616996-116616996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530756069 Chr.6: 116616891-116616891 on Build GRCh38
rs577977924 Chr.6: 116616965-116616965 on Build GRCh38
rs895120131 Chr.6: 116616867-116616867 on Build GRCh38
rs764572415 Chr.6: 116616995-116616996 on Build GRCh38
rs201187909 Chr.6: 116616910-116616910 on Build GRCh38
rs118204042 Chr.6: 116616948-116616948 on Build GRCh38
rs768436105 Chr.6: 116617016-116617016 on Build GRCh38
rs371869815 Chr.6: 116616865-116616865 on Build GRCh38
rs555565982 Chr.6: 116616888-116616888 on Build GRCh38
rs976414886 Chr.6: 116617020-116617020 on Build GRCh38
rs755522050 Chr.6: 116616902-116616902 on Build GRCh38
rs553472573 Chr.6: 116616847-116616847 on Build GRCh38
rs773878067 Chr.6: 116616870-116616870 on Build GRCh38
rs759417066 Chr.6: 116616878-116616878 on Build GRCh38
rs1018616962 Chr.6: 116616993-116616993 on Build GRCh38
rs202006186 Chr.6: 116616883-116616883 on Build GRCh38
rs775559853 Chr.6: 116616982-116616982 on Build GRCh38
rs1008393343 Chr.6: 116616876-116616876 on Build GRCh38
rs766758184 Chr.6: 116617019-116617019 on Build GRCh38
rs766527231 Chr.6: 116616893-116616893 on Build GRCh38
rs145831200 Chr.6: 116616860-116616860 on Build GRCh38
rs756442852 Chr.6: 116616924-116616924 on Build GRCh38
rs1030816346 Chr.6: 116616936-116616936 on Build GRCh38
rs374708076 Chr.6: 116617017-116617017 on Build GRCh38
rs139745647 Chr.6: 116616930-116616930 on Build GRCh38
rs778370377 Chr.6: 116616963-116616963 on Build GRCh38
rs781780861 Chr.6: 116616909-116616909 on Build GRCh38
rs767490154 Chr.6: 116616882-116616882 on Build GRCh38
rs769325632 Chr.6: 116616843-116616843 on Build GRCh38
rs748926290 Chr.6: 116616940-116616940 on Build GRCh38
rs763466678 Chr.6: 116616967-116616968 on Build GRCh38
rs369824862 Chr.6: 116617010-116617010 on Build GRCh38
rs763188022 Chr.6: 116617018-116617018 on Build GRCh38
rs775574466 Chr.6: 116616859-116616861 on Build GRCh38
rs562856574 Chr.6: 116616964-116616964 on Build GRCh38
rs760588703 Chr.6: 116616885-116616885 on Build GRCh38
rs751712749 Chr.6: 116616895-116616895 on Build GRCh38
rs750605299 Chr.6: 116616889-116616889 on Build GRCh38

View Full Product Details