Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 51320755-51320996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXDNL
Entrez Gene ID
137902
Gene Name
peroxidasin like
Gene Aliases
PMR1, PRM1, VPO2
UniGene
Hs.444882
Gene Chromosome Location
Chr.8: 51319575-51811031 on Build GRCh38
Transcripts
NM_144651.4 XM_005251168.3 XM_006716420.3 XM_011517456.2 XM_011517457.2 XM_011517458.2 XM_017013041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3951712 c.4260+10G>T p.? Chr.8: 51320774-51320774 on Build GRCh38
4588139 c.4248C>T p.H1416H Chr.8: 51320796-51320796 on Build GRCh38
1263613 c.4233A>G p.K1411K Chr.8: 51320811-51320811 on Build GRCh38
5874770 c.4193T>C p.V1398A Chr.8: 51320851-51320851 on Build GRCh38
341530 c.4176G>A p.Q1392Q Chr.8: 51320868-51320868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866068105 Chr.8: 51320855-51320855 on Build GRCh38
rs574393200 Chr.8: 51320869-51320869 on Build GRCh38
rs770443496 Chr.8: 51320820-51320820 on Build GRCh38
rs748474859 Chr.8: 51320856-51320856 on Build GRCh38
rs778198681 Chr.8: 51320876-51320876 on Build GRCh38
rs771582296 Chr.8: 51320922-51320922 on Build GRCh38
rs180687856 Chr.8: 51320805-51320805 on Build GRCh38
rs762061589 Chr.8: 51320862-51320862 on Build GRCh38
rs781468410 Chr.8: 51320792-51320792 on Build GRCh38
rs760487068 Chr.8: 51320932-51320932 on Build GRCh38
rs899776538 Chr.8: 51320968-51320968 on Build GRCh38
rs775228222 Chr.8: 51320923-51320923 on Build GRCh38
rs201816215 Chr.8: 51320821-51320821 on Build GRCh38
rs370461650 Chr.8: 51320796-51320796 on Build GRCh38
rs537952252 Chr.8: 51320822-51320822 on Build GRCh38
rs373372683 Chr.8: 51320948-51320948 on Build GRCh38
rs761899792 Chr.8: 51320944-51320944 on Build GRCh38
rs371945543 Chr.8: 51320938-51320938 on Build GRCh38
rs868605102 Chr.8: 51320837-51320837 on Build GRCh38
rs973601661 Chr.8: 51320849-51320849 on Build GRCh38
rs7827446 Chr.8: 51320848-51320848 on Build GRCh38
rs778332200 Chr.8: 51320900-51320900 on Build GRCh38
rs368775744 Chr.8: 51320945-51320945 on Build GRCh38
rs781318486 Chr.8: 51320806-51320806 on Build GRCh38
rs758484347 Chr.8: 51320830-51320830 on Build GRCh38
rs751704591 Chr.8: 51320807-51320808 on Build GRCh38
rs541402353 Chr.8: 51320910-51320910 on Build GRCh38
rs749802319 Chr.8: 51320878-51320878 on Build GRCh38
rs755364527 Chr.8: 51320782-51320782 on Build GRCh38
rs916192165 Chr.8: 51320838-51320838 on Build GRCh38
rs937437549 Chr.8: 51320956-51320956 on Build GRCh38
rs748750828 Chr.8: 51320817-51320817 on Build GRCh38
rs376862990 Chr.8: 51320939-51320939 on Build GRCh38
rs202101425 Chr.8: 51320875-51320875 on Build GRCh38
rs929428392 Chr.8: 51320872-51320872 on Build GRCh38
rs766435712 Chr.8: 51320846-51320846 on Build GRCh38
rs751791668 Chr.8: 51320847-51320847 on Build GRCh38
rs760575154 Chr.8: 51320823-51320823 on Build GRCh38
rs751630629 Chr.8: 51320947-51320947 on Build GRCh38
rs868032388 Chr.8: 51320863-51320863 on Build GRCh38
rs890532443 Chr.8: 51320926-51320926 on Build GRCh38
rs977211245 Chr.8: 51320811-51320811 on Build GRCh38
rs371941977 Chr.8: 51320829-51320829 on Build GRCh38
rs755986290 Chr.8: 51320828-51320828 on Build GRCh38
rs781589832 Chr.8: 51320854-51320854 on Build GRCh38
rs368422364 Chr.8: 51320879-51320879 on Build GRCh38
rs773741030 Chr.8: 51320807-51320807 on Build GRCh38

View Full Product Details